Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 11998662-11999258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPA2
Entrez Gene ID
3613
Gene Name
inositol monophosphatase 2
Gene Aliases
-
UniGene
Hs.743311
Gene Chromosome Location
Chr.18: 11981428-12030886 on Build GRCh38
Transcripts
NM_014214.2 XM_011525661.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
986584 c.97-2A>C p.? Chr.18: 11999052-11999052 on Build GRCh38
3932797 c.102C>G p.I34M Chr.18: 11999059-11999059 on Build GRCh38
148396 c.159T>C p.L53L Chr.18: 11999116-11999116 on Build GRCh38
4130913 c.119A>T p.E40V Chr.18: 11999076-11999076 on Build GRCh38
3524224 c.224C>T p.S75L Chr.18: 11999181-11999181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963867589 Chr.18: 11998899-11998899 on Build GRCh38
rs586585 Chr.18: 11998735-11998735 on Build GRCh38
rs779664047 Chr.18: 11999212-11999212 on Build GRCh38
rs963472837 Chr.18: 11999103-11999103 on Build GRCh38
rs781208697 Chr.18: 11999053-11999053 on Build GRCh38
rs754812678 Chr.18: 11999149-11999150 on Build GRCh38
rs766443589 Chr.18: 11999008-11999008 on Build GRCh38
rs910407031 Chr.18: 11998925-11998925 on Build GRCh38
rs529603887 Chr.18: 11998733-11998733 on Build GRCh38
rs972367439 Chr.18: 11998905-11998905 on Build GRCh38
rs755567980 Chr.18: 11999209-11999209 on Build GRCh38
rs1009049331 Chr.18: 11999002-11999002 on Build GRCh38
rs998068092 Chr.18: 11998979-11998979 on Build GRCh38
rs369305882 Chr.18: 11999143-11999143 on Build GRCh38
rs45442994 Chr.18: 11999039-11999039 on Build GRCh38
rs1051805182 Chr.18: 11998984-11998984 on Build GRCh38
rs547087703 Chr.18: 11998689-11998689 on Build GRCh38
rs1052932563 Chr.18: 11998951-11998951 on Build GRCh38
rs1007646412 Chr.18: 11998847-11998847 on Build GRCh38
rs1030308190 Chr.18: 11998784-11998784 on Build GRCh38
rs953450834 Chr.18: 11998809-11998809 on Build GRCh38
rs948510153 Chr.18: 11998944-11998944 on Build GRCh38
rs749561254 Chr.18: 11998942-11998942 on Build GRCh38
rs192327840 Chr.18: 11998765-11998765 on Build GRCh38
rs767293538 Chr.18: 11999197-11999197 on Build GRCh38
rs73946208 Chr.18: 11998928-11998928 on Build GRCh38
rs935506342 Chr.18: 11998791-11998791 on Build GRCh38
rs780203010 Chr.18: 11999039-11999039 on Build GRCh38
rs749002337 Chr.18: 11999071-11999071 on Build GRCh38
rs748544516 Chr.18: 11999109-11999109 on Build GRCh38
rs925835789 Chr.18: 11998948-11998948 on Build GRCh38
rs760665367 Chr.18: 11999131-11999131 on Build GRCh38
rs778562558 Chr.18: 11999216-11999216 on Build GRCh38
rs901098450 Chr.18: 11998974-11998974 on Build GRCh38
rs569906849 Chr.18: 11998894-11998894 on Build GRCh38
rs764029423 Chr.18: 11999031-11999031 on Build GRCh38
rs201071409 Chr.18: 11999105-11999105 on Build GRCh38
rs878890066 Chr.18: 11998940-11998940 on Build GRCh38
rs200026931 Chr.18: 11998947-11998947 on Build GRCh38
rs928075270 Chr.18: 11999186-11999186 on Build GRCh38
rs780168052 Chr.18: 11998941-11998941 on Build GRCh38
rs777607026 Chr.18: 11998943-11998943 on Build GRCh38
rs971926801 Chr.18: 11998700-11998700 on Build GRCh38
rs1024036576 Chr.18: 11999091-11999091 on Build GRCh38
rs534858144 Chr.18: 11999021-11999021 on Build GRCh38
rs760833026 Chr.18: 11999006-11999006 on Build GRCh38
rs1043188530 Chr.18: 11998946-11998946 on Build GRCh38
rs961932207 Chr.18: 11998692-11998692 on Build GRCh38
rs915613413 Chr.18: 11999194-11999194 on Build GRCh38
rs753301581 Chr.18: 11999238-11999238 on Build GRCh38
rs748456705 Chr.18: 11999228-11999228 on Build GRCh38
rs11545506 Chr.18: 11999116-11999116 on Build GRCh38
rs982584329 Chr.18: 11999138-11999138 on Build GRCh38
rs773785260 Chr.18: 11999121-11999121 on Build GRCh38
rs374737686 Chr.18: 11999085-11999085 on Build GRCh38
rs1027545190 Chr.18: 11998709-11998709 on Build GRCh38
rs960285738 Chr.18: 11999178-11999178 on Build GRCh38
rs952674419 Chr.18: 11998716-11998716 on Build GRCh38
rs1052649914 Chr.18: 11998856-11998856 on Build GRCh38
rs759877969 Chr.18: 11999157-11999157 on Build GRCh38
rs558849421 Chr.18: 11998952-11998952 on Build GRCh38
rs982240364 Chr.18: 11998773-11998773 on Build GRCh38
rs779571987 Chr.18: 11999067-11999067 on Build GRCh38
rs199839687 Chr.18: 11999187-11999187 on Build GRCh38
rs184740260 Chr.18: 11999166-11999166 on Build GRCh38
rs765654972 Chr.18: 11999128-11999128 on Build GRCh38
rs559700702 Chr.18: 11998928-11998932 on Build GRCh38
rs772551509 Chr.18: 11999231-11999231 on Build GRCh38
rs112775784 Chr.18: 11998913-11998913 on Build GRCh38
rs201336998 Chr.18: 11999205-11999205 on Build GRCh38
rs377725722 Chr.18: 11998923-11998925 on Build GRCh38
rs897637832 Chr.18: 11999036-11999036 on Build GRCh38
rs776694209 Chr.18: 11999154-11999154 on Build GRCh38
rs998814612 Chr.18: 11998726-11998726 on Build GRCh38
rs570648197 Chr.18: 11998991-11998991 on Build GRCh38
rs894072738 Chr.18: 11998908-11998908 on Build GRCh38
rs748912477 Chr.18: 11999213-11999213 on Build GRCh38
rs774737574 Chr.18: 11999106-11999106 on Build GRCh38
rs74696534 Chr.18: 11998947-11998947 on Build GRCh38
rs200522493 Chr.18: 11999038-11999038 on Build GRCh38
rs796170557 Chr.18: 11998926-11998933 on Build GRCh38
rs751528431 Chr.18: 11999085-11999086 on Build GRCh38
rs368231623 Chr.18: 11999030-11999030 on Build GRCh38
rs755657106 Chr.18: 11999064-11999064 on Build GRCh38
rs946987188 Chr.18: 11998929-11998938 on Build GRCh38
rs565442781 Chr.18: 11998732-11998732 on Build GRCh38
rs781044507 Chr.18: 11999238-11999239 on Build GRCh38
rs745311348 Chr.18: 11999063-11999063 on Build GRCh38
rs915452626 Chr.18: 11999227-11999227 on Build GRCh38
rs373209322 Chr.18: 11998826-11998826 on Build GRCh38
rs984092213 Chr.18: 11998718-11998718 on Build GRCh38
rs111952278 Chr.18: 11999120-11999120 on Build GRCh38
rs751350184 Chr.18: 11999170-11999170 on Build GRCh38
rs62099893 Chr.18: 11998950-11998950 on Build GRCh38

View Full Product Details