Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9962679-9962965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL5A3
Entrez Gene ID
50509
Gene Name
collagen type V alpha 3
Gene Aliases
-
UniGene
Hs.235368
Gene Chromosome Location
Chr.19: 9959561-10010532 on Build GRCh38
Transcripts
NM_015719.3 XM_011528042.2 XM_017026849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
41481 c.4851+10C>A p.? Chr.19: 9962809-9962809 on Build GRCh38
310265 c.4821C>T p.G1607G Chr.19: 9962849-9962849 on Build GRCh38
230363 c.4846A>G p.K1616E Chr.19: 9962824-9962824 on Build GRCh38
474084 c.4825T>A p.Y1609N Chr.19: 9962845-9962845 on Build GRCh38
3891396 c.4845G>A p.G1615G Chr.19: 9962825-9962825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888781280 Chr.19: 9962775-9962775 on Build GRCh38
rs543905269 Chr.19: 9962794-9962794 on Build GRCh38
rs942307695 Chr.19: 9962760-9962760 on Build GRCh38
rs149577329 Chr.19: 9962735-9962735 on Build GRCh38
rs1022102593 Chr.19: 9962938-9962938 on Build GRCh38
rs112995727 Chr.19: 9962776-9962776 on Build GRCh38
rs753270591 Chr.19: 9962896-9962896 on Build GRCh38
rs748953537 Chr.19: 9962829-9962829 on Build GRCh38
rs760485484 Chr.19: 9962837-9962837 on Build GRCh38
rs769774899 Chr.19: 9962830-9962831 on Build GRCh38
rs113051582 Chr.19: 9962893-9962893 on Build GRCh38
rs572351428 Chr.19: 9962724-9962724 on Build GRCh38
rs1037823059 Chr.19: 9962798-9962798 on Build GRCh38
rs745622461 Chr.19: 9962819-9962821 on Build GRCh38
rs1050339084 Chr.19: 9962763-9962763 on Build GRCh38
rs756521310 Chr.19: 9962908-9962908 on Build GRCh38
rs771639973 Chr.19: 9962935-9962935 on Build GRCh38
rs775470366 Chr.19: 9962835-9962835 on Build GRCh38
rs148658694 Chr.19: 9962880-9962880 on Build GRCh38
rs778517875 Chr.19: 9962925-9962925 on Build GRCh38
rs1011588597 Chr.19: 9962932-9962932 on Build GRCh38
rs747596416 Chr.19: 9962887-9962887 on Build GRCh38
rs372438430 Chr.19: 9962939-9962939 on Build GRCh38
rs758640951 Chr.19: 9962793-9962793 on Build GRCh38
rs776450230 Chr.19: 9962788-9962809 on Build GRCh38
rs561098782 Chr.19: 9962765-9962765 on Build GRCh38
rs756469748 Chr.19: 9962814-9962814 on Build GRCh38
rs373180563 Chr.19: 9962833-9962833 on Build GRCh38
rs911151476 Chr.19: 9962723-9962723 on Build GRCh38
rs1038330430 Chr.19: 9962816-9962816 on Build GRCh38
rs745418389 Chr.19: 9962930-9962930 on Build GRCh38
rs58180798 Chr.19: 9962900-9962901 on Build GRCh38
rs148762424 Chr.19: 9962803-9962803 on Build GRCh38
rs776227666 Chr.19: 9962855-9962855 on Build GRCh38
rs765421530 Chr.19: 9962784-9962784 on Build GRCh38
rs760500778 Chr.19: 9962780-9962780 on Build GRCh38
rs377581980 Chr.19: 9962891-9962891 on Build GRCh38
rs371142637 Chr.19: 9962921-9962921 on Build GRCh38
rs770506606 Chr.19: 9962830-9962830 on Build GRCh38
rs368933139 Chr.19: 9962832-9962832 on Build GRCh38
rs111952540 Chr.19: 9962808-9962808 on Build GRCh38
rs756964632 Chr.19: 9962923-9962923 on Build GRCh38
rs112341845 Chr.19: 9962802-9962802 on Build GRCh38
rs772405599 Chr.19: 9962933-9962933 on Build GRCh38
rs751745540 Chr.19: 9962881-9962881 on Build GRCh38
rs777951601 Chr.19: 9962818-9962818 on Build GRCh38
rs377236149 Chr.19: 9962809-9962809 on Build GRCh38
rs547819367 Chr.19: 9962857-9962857 on Build GRCh38
rs775210362 Chr.19: 9962778-9962778 on Build GRCh38
rs376649429 Chr.19: 9962919-9962919 on Build GRCh38
rs750684787 Chr.19: 9962788-9962788 on Build GRCh38
rs550247987 Chr.19: 9962708-9962708 on Build GRCh38

View Full Product Details