Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 86046348-86046788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLM1
Entrez Gene ID
51280
Gene Name
golgi membrane protein 1
Gene Aliases
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3
UniGene
Hs.494337
Gene Chromosome Location
Chr.9: 86026141-86100201 on Build GRCh38
Transcripts
NM_016548.3 NM_177937.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3659297 c.389A>T p.N130I Chr.9: 86046548-86046548 on Build GRCh38
6115968 c.395G>C p.G132A Chr.9: 86046542-86046542 on Build GRCh38
4491324 c.378C>T p.T126T Chr.9: 86046559-86046559 on Build GRCh38
6452014 c.416T>C p.L139P Chr.9: 86046521-86046521 on Build GRCh38
1110753 c.393C>T p.Y131Y Chr.9: 86046544-86046544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765721826 Chr.9: 86046727-86046727 on Build GRCh38
rs12350451 Chr.9: 86046571-86046571 on Build GRCh38
rs370485983 Chr.9: 86046469-86046469 on Build GRCh38
rs776343347 Chr.9: 86046421-86046421 on Build GRCh38
rs369300245 Chr.9: 86046423-86046423 on Build GRCh38
rs771621652 Chr.9: 86046608-86046608 on Build GRCh38
rs896004386 Chr.9: 86046725-86046725 on Build GRCh38
rs1020031046 Chr.9: 86046713-86046713 on Build GRCh38
rs763717503 Chr.9: 86046619-86046619 on Build GRCh38
rs202205078 Chr.9: 86046701-86046702 on Build GRCh38
rs116356059 Chr.9: 86046467-86046467 on Build GRCh38
rs184738517 Chr.9: 86046685-86046685 on Build GRCh38
rs560186396 Chr.9: 86046709-86046709 on Build GRCh38
rs773076505 Chr.9: 86046516-86046516 on Build GRCh38
rs140984957 Chr.9: 86046538-86046538 on Build GRCh38
rs768757816 Chr.9: 86046584-86046584 on Build GRCh38
rs776654774 Chr.9: 86046610-86046610 on Build GRCh38
rs760184860 Chr.9: 86046554-86046554 on Build GRCh38
rs773305977 Chr.9: 86046430-86046430 on Build GRCh38
rs1048533224 Chr.9: 86046636-86046636 on Build GRCh38
rs532020120 Chr.9: 86046710-86046710 on Build GRCh38
rs201724840 Chr.9: 86046449-86046449 on Build GRCh38
rs534059912 Chr.9: 86046474-86046474 on Build GRCh38
rs774461037 Chr.9: 86046601-86046601 on Build GRCh38
rs373928322 Chr.9: 86046544-86046544 on Build GRCh38
rs188938440 Chr.9: 86046431-86046431 on Build GRCh38
rs756528880 Chr.9: 86046464-86046464 on Build GRCh38
rs780359666 Chr.9: 86046502-86046502 on Build GRCh38
rs747260980 Chr.9: 86046577-86046577 on Build GRCh38
rs774239440 Chr.9: 86046532-86046532 on Build GRCh38
rs896837384 Chr.9: 86046411-86046411 on Build GRCh38
rs764443794 Chr.9: 86046557-86046557 on Build GRCh38
rs1018221735 Chr.9: 86046696-86046696 on Build GRCh38
rs759053998 Chr.9: 86046539-86046539 on Build GRCh38
rs779024724 Chr.9: 86046475-86046475 on Build GRCh38
rs542456452 Chr.9: 86046673-86046673 on Build GRCh38
rs772092142 Chr.9: 86046484-86046486 on Build GRCh38
rs957192171 Chr.9: 86046442-86046442 on Build GRCh38
rs750894457 Chr.9: 86046567-86046567 on Build GRCh38
rs777023265 Chr.9: 86046621-86046621 on Build GRCh38
rs897126080 Chr.9: 86046413-86046413 on Build GRCh38
rs373948424 Chr.9: 86046451-86046451 on Build GRCh38
rs1018084205 Chr.9: 86046440-86046440 on Build GRCh38
rs769644477 Chr.9: 86046513-86046513 on Build GRCh38
rs878955913 Chr.9: 86046468-86046468 on Build GRCh38
rs777767888 Chr.9: 86046588-86046588 on Build GRCh38
rs754191893 Chr.9: 86046558-86046558 on Build GRCh38
rs760439741 Chr.9: 86046618-86046618 on Build GRCh38
rs368914992 Chr.9: 86046450-86046450 on Build GRCh38
rs994604518 Chr.9: 86046415-86046415 on Build GRCh38
rs779940523 Chr.9: 86046575-86046575 on Build GRCh38
rs141970031 Chr.9: 86046505-86046505 on Build GRCh38
rs779364702 Chr.9: 86046560-86046560 on Build GRCh38
rs991691031 Chr.9: 86046746-86046746 on Build GRCh38
rs772226448 Chr.9: 86046497-86046497 on Build GRCh38
rs943945427 Chr.9: 86046368-86046368 on Build GRCh38
rs540002481 Chr.9: 86046705-86046705 on Build GRCh38
rs199583252 Chr.9: 86046543-86046543 on Build GRCh38
rs370080649 Chr.9: 86046452-86046452 on Build GRCh38
rs562304890 Chr.9: 86046674-86046674 on Build GRCh38
rs930996803 Chr.9: 86046631-86046631 on Build GRCh38
rs539802579 Chr.9: 86046580-86046580 on Build GRCh38
rs745928797 Chr.9: 86046490-86046490 on Build GRCh38
rs192198513 Chr.9: 86046599-86046599 on Build GRCh38
rs757813183 Chr.9: 86046559-86046559 on Build GRCh38
rs928231990 Chr.9: 86046530-86046530 on Build GRCh38
rs147300680 Chr.9: 86046525-86046525 on Build GRCh38
rs116583603 Chr.9: 86046386-86046386 on Build GRCh38
rs74457723 Chr.9: 86046729-86046729 on Build GRCh38
rs148043716 Chr.9: 86046607-86046607 on Build GRCh38
rs775080404 Chr.9: 86046720-86046721 on Build GRCh38
rs1055307616 Chr.9: 86046542-86046542 on Build GRCh38
rs772328860 Chr.9: 86046419-86046419 on Build GRCh38
rs7041576 Chr.9: 86046429-86046429 on Build GRCh38
rs932053677 Chr.9: 86046587-86046587 on Build GRCh38
rs749068693 Chr.9: 86046596-86046596 on Build GRCh38
rs566374621 Chr.9: 86046381-86046381 on Build GRCh38

View Full Product Details