Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121186697-121187147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB14
Entrez Gene ID
51552
Gene Name
RAB14, member RAS oncogene family
Gene Aliases
FBP, RAB-14
UniGene
Hs.371563
Gene Chromosome Location
Chr.9: 121178137-121202087 on Build GRCh38
Transcripts
NM_016322.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3664055 c.349A>T p.T117S Chr.9: 121186955-121186955 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201249611 Chr.9: 121187042-121187042 on Build GRCh38
rs1011953240 Chr.9: 121186779-121186779 on Build GRCh38
rs1030898804 Chr.9: 121186879-121186879 on Build GRCh38
rs111350313 Chr.9: 121187091-121187091 on Build GRCh38
rs576800708 Chr.9: 121186845-121186845 on Build GRCh38
rs909356176 Chr.9: 121186923-121186923 on Build GRCh38
rs911057054 Chr.9: 121187047-121187047 on Build GRCh38
rs971876551 Chr.9: 121187043-121187043 on Build GRCh38
rs748221019 Chr.9: 121186932-121186932 on Build GRCh38
rs781389261 Chr.9: 121186925-121186925 on Build GRCh38
rs1056622780 Chr.9: 121186757-121186757 on Build GRCh38
rs751871775 Chr.9: 121186860-121186860 on Build GRCh38
rs1004143678 Chr.9: 121186756-121186756 on Build GRCh38
rs967699736 Chr.9: 121186832-121186832 on Build GRCh38
rs777710614 Chr.9: 121187023-121187023 on Build GRCh38
rs992151997 Chr.9: 121186858-121186858 on Build GRCh38
rs755300623 Chr.9: 121186912-121186912 on Build GRCh38
rs756171797 Chr.9: 121186939-121186939 on Build GRCh38
rs748240673 Chr.9: 121187065-121187065 on Build GRCh38
rs1000984776 Chr.9: 121186767-121186767 on Build GRCh38
rs543014750 Chr.9: 121186740-121186740 on Build GRCh38
rs548413890 Chr.9: 121187104-121187104 on Build GRCh38
rs894926952 Chr.9: 121186761-121186761 on Build GRCh38
rs145269731 Chr.9: 121186846-121186846 on Build GRCh38
rs766507742 Chr.9: 121186742-121186742 on Build GRCh38
rs955301824 Chr.9: 121186915-121186915 on Build GRCh38
rs559539631 Chr.9: 121187065-121187065 on Build GRCh38
rs185836182 Chr.9: 121186798-121186798 on Build GRCh38
rs920650412 Chr.9: 121187073-121187073 on Build GRCh38
rs367866798 Chr.9: 121186911-121186911 on Build GRCh38
rs974833665 Chr.9: 121187072-121187072 on Build GRCh38

View Full Product Details