Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 68838496-68839044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNB1
Entrez Gene ID
1947
Gene Name
ephrin B1
Gene Aliases
CFND, CFNS, EFB1, EFL3, EPLG2, Elk-L, LERK2
UniGene
Hs.144700
Gene Chromosome Location
Chr.X: 68828997-68842164 on Build GRCh38
Transcripts
NM_004429.4

SNP Detail

SNP ID Chromosome Location
rs781514536 Chr.X: 68838827-68838827 on Build GRCh38
rs752714814 Chr.X: 68838782-68838782 on Build GRCh38
rs927176447 Chr.X: 68838935-68838935 on Build GRCh38
rs778669520 Chr.X: 68838768-68838768 on Build GRCh38
rs1028526303 Chr.X: 68838558-68838558 on Build GRCh38
rs104894804 Chr.X: 68838684-68838684 on Build GRCh38
rs771228253 Chr.X: 68838652-68838652 on Build GRCh38
rs868481776 Chr.X: 68838618-68838618 on Build GRCh38
rs1020239313 Chr.X: 68838677-68838677 on Build GRCh38
rs753571763 Chr.X: 68838925-68838925 on Build GRCh38
rs765712163 Chr.X: 68838693-68838693 on Build GRCh38
rs749220715 Chr.X: 68838576-68838576 on Build GRCh38
rs772921295 Chr.X: 68838904-68838904 on Build GRCh38
rs954074835 Chr.X: 68838567-68838567 on Build GRCh38
rs760376529 Chr.X: 68838919-68838919 on Build GRCh38
rs754924952 Chr.X: 68838767-68838767 on Build GRCh38
rs995935535 Chr.X: 68838533-68838533 on Build GRCh38
rs747219481 Chr.X: 68838814-68838814 on Build GRCh38
rs759216120 Chr.X: 68838943-68838943 on Build GRCh38
rs772218757 Chr.X: 68838583-68838583 on Build GRCh38
rs917291771 Chr.X: 68838980-68838980 on Build GRCh38
rs747725805 Chr.X: 68838569-68838569 on Build GRCh38
rs104894801 Chr.X: 68838649-68838649 on Build GRCh38
rs370637906 Chr.X: 68838733-68838733 on Build GRCh38
rs779023711 Chr.X: 68838579-68838579 on Build GRCh38
rs757940803 Chr.X: 68838910-68838910 on Build GRCh38
rs771211723 Chr.X: 68838821-68838821 on Build GRCh38
rs767055938 Chr.X: 68838700-68838700 on Build GRCh38
rs746168241 Chr.X: 68838863-68838863 on Build GRCh38
rs1019590407 Chr.X: 68838665-68838665 on Build GRCh38
rs777846246 Chr.X: 68838803-68838803 on Build GRCh38
rs367996552 Chr.X: 68838699-68838699 on Build GRCh38
rs375090760 Chr.X: 68838923-68838923 on Build GRCh38
rs145603720 Chr.X: 68838621-68838621 on Build GRCh38
rs938555803 Chr.X: 68838951-68838951 on Build GRCh38
rs755885387 Chr.X: 68838731-68838731 on Build GRCh38
rs777382965 Chr.X: 68838941-68838944 on Build GRCh38
rs758345370 Chr.X: 68838783-68838783 on Build GRCh38
rs770225645 Chr.X: 68838879-68838879 on Build GRCh38
rs797045549 Chr.X: 68838663-68838663 on Build GRCh38
rs104894796 Chr.X: 68838820-68838820 on Build GRCh38
rs986747386 Chr.X: 68838580-68838580 on Build GRCh38
rs776085390 Chr.X: 68838694-68838694 on Build GRCh38
rs886041800 Chr.X: 68838707-68838707 on Build GRCh38
rs771693614 Chr.X: 68838903-68838903 on Build GRCh38
rs148907346 Chr.X: 68838680-68838680 on Build GRCh38
rs774333672 Chr.X: 68838582-68838582 on Build GRCh38
rs978792532 Chr.X: 68838810-68838810 on Build GRCh38
rs766186701 Chr.X: 68838924-68838924 on Build GRCh38
rs1057519032 Chr.X: 68838759-68838759 on Build GRCh38
rs754940451 Chr.X: 68838578-68838578 on Build GRCh38
rs1001112997 Chr.X: 68838544-68838544 on Build GRCh38
rs748103349 Chr.X: 68838685-68838685 on Build GRCh38
rs1056028 Chr.X: 68838650-68838650 on Build GRCh38
rs775791339 Chr.X: 68838887-68838887 on Build GRCh38
rs763525185 Chr.X: 68838893-68838893 on Build GRCh38
rs1057214451 Chr.X: 68838954-68838954 on Build GRCh38
rs750035783 Chr.X: 68838725-68838725 on Build GRCh38
rs768219945 Chr.X: 68838977-68838977 on Build GRCh38
rs748909631 Chr.X: 68838534-68838534 on Build GRCh38
rs891427646 Chr.X: 68838979-68838979 on Build GRCh38
rs143648191 Chr.X: 68838750-68838750 on Build GRCh38

View Full Product Details