Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76738980-76739570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHROOM3
Entrez Gene ID
57619
Gene Name
shroom family member 3
Gene Aliases
APXL3, MSTP013, SHRM, ShrmL
UniGene
Hs.683808
Gene Chromosome Location
Chr.4: 76435100-76783253 on Build GRCh38
Transcripts
NM_020859.3 XM_005263163.4 XM_005263164.4 XM_005263165.4 XM_006714282.3 XM_011532157.2 XM_011532158.2 XM_011532159.1 XM_011532160.1 XM_011532161.2 XM_017008488.1 XM_017008489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7298972 c.1029C>T p.G343G Chr.4: 76739202-76739202 on Build GRCh38
5481901 c.845G>A p.R282H Chr.4: 76739018-76739018 on Build GRCh38
3409497 c.1055G>A p.R352Q Chr.4: 76739228-76739228 on Build GRCh38
1225677 c.889G>A p.E297K Chr.4: 76739062-76739062 on Build GRCh38
213415 c.1211G>T p.W404L Chr.4: 76739384-76739384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572629061 Chr.4: 76739519-76739519 on Build GRCh38
rs998058121 Chr.4: 76739185-76739185 on Build GRCh38
rs1004266045 Chr.4: 76739127-76739127 on Build GRCh38
rs759067719 Chr.4: 76739522-76739522 on Build GRCh38
rs769701354 Chr.4: 76739087-76739087 on Build GRCh38
rs760768267 Chr.4: 76739112-76739112 on Build GRCh38
rs776605073 Chr.4: 76739017-76739017 on Build GRCh38
rs374543213 Chr.4: 76739040-76739040 on Build GRCh38
rs775627642 Chr.4: 76739390-76739390 on Build GRCh38
rs764886278 Chr.4: 76739020-76739020 on Build GRCh38
rs143726434 Chr.4: 76739404-76739404 on Build GRCh38
rs754121390 Chr.4: 76739105-76739105 on Build GRCh38
rs747906408 Chr.4: 76739370-76739370 on Build GRCh38
rs752580613 Chr.4: 76739451-76739451 on Build GRCh38
rs111521571 Chr.4: 76739067-76739067 on Build GRCh38
rs201586601 Chr.4: 76739091-76739091 on Build GRCh38
rs142944791 Chr.4: 76739525-76739525 on Build GRCh38
rs201389959 Chr.4: 76739340-76739340 on Build GRCh38
rs61745983 Chr.4: 76739397-76739397 on Build GRCh38
rs533735013 Chr.4: 76739336-76739336 on Build GRCh38
rs889643632 Chr.4: 76739439-76739439 on Build GRCh38
rs377324397 Chr.4: 76739025-76739025 on Build GRCh38
rs747894530 Chr.4: 76739437-76739437 on Build GRCh38
rs765626467 Chr.4: 76739237-76739237 on Build GRCh38
rs765190647 Chr.4: 76739444-76739444 on Build GRCh38
rs761601802 Chr.4: 76739349-76739349 on Build GRCh38
rs1043202472 Chr.4: 76739320-76739320 on Build GRCh38
rs751339105 Chr.4: 76739545-76739545 on Build GRCh38
rs202185868 Chr.4: 76739047-76739047 on Build GRCh38
rs777319771 Chr.4: 76739139-76739139 on Build GRCh38
rs777388129 Chr.4: 76739061-76739061 on Build GRCh38
rs150617668 Chr.4: 76739508-76739508 on Build GRCh38
rs867503578 Chr.4: 76739441-76739441 on Build GRCh38
rs142581367 Chr.4: 76739053-76739053 on Build GRCh38
rs141874410 Chr.4: 76739436-76739436 on Build GRCh38
rs779579252 Chr.4: 76739503-76739503 on Build GRCh38
rs774963603 Chr.4: 76739405-76739405 on Build GRCh38
rs374712873 Chr.4: 76739473-76739473 on Build GRCh38
rs757837533 Chr.4: 76739250-76739250 on Build GRCh38
rs755595009 Chr.4: 76739359-76739359 on Build GRCh38
rs760368145 Chr.4: 76739418-76739418 on Build GRCh38
rs754292324 Chr.4: 76739039-76739039 on Build GRCh38
rs778673286 Chr.4: 76739293-76739293 on Build GRCh38
rs756959889 Chr.4: 76739260-76739260 on Build GRCh38
rs745964950 Chr.4: 76739321-76739321 on Build GRCh38
rs144518459 Chr.4: 76739421-76739421 on Build GRCh38
rs554492056 Chr.4: 76739227-76739227 on Build GRCh38
rs752977945 Chr.4: 76739118-76739118 on Build GRCh38
rs746996342 Chr.4: 76739011-76739011 on Build GRCh38
rs199685557 Chr.4: 76739113-76739113 on Build GRCh38
rs758148359 Chr.4: 76739371-76739371 on Build GRCh38
rs138985148 Chr.4: 76739291-76739291 on Build GRCh38
rs777555163 Chr.4: 76739458-76739458 on Build GRCh38
rs373643546 Chr.4: 76739548-76739548 on Build GRCh38
rs775139022 Chr.4: 76739092-76739092 on Build GRCh38
rs781294814 Chr.4: 76739072-76739072 on Build GRCh38
rs753783570 Chr.4: 76739363-76739363 on Build GRCh38
rs781290519 Chr.4: 76739372-76739372 on Build GRCh38
rs760134794 Chr.4: 76739232-76739232 on Build GRCh38
rs769596316 Chr.4: 76739179-76739179 on Build GRCh38
rs750480368 Chr.4: 76739031-76739031 on Build GRCh38
rs367773952 Chr.4: 76739086-76739086 on Build GRCh38
rs754893564 Chr.4: 76739365-76739365 on Build GRCh38
rs962270438 Chr.4: 76739138-76739138 on Build GRCh38
rs200716219 Chr.4: 76739240-76739240 on Build GRCh38
rs774834974 Chr.4: 76739348-76739348 on Build GRCh38
rs372029011 Chr.4: 76739290-76739290 on Build GRCh38
rs757969587 Chr.4: 76739056-76739056 on Build GRCh38
rs772628720 Chr.4: 76739507-76739507 on Build GRCh38
rs746043483 Chr.4: 76739376-76739376 on Build GRCh38
rs147426728 Chr.4: 76739529-76739529 on Build GRCh38
rs759433745 Chr.4: 76739018-76739018 on Build GRCh38
rs371504856 Chr.4: 76739048-76739048 on Build GRCh38
rs542307605 Chr.4: 76739464-76739464 on Build GRCh38
rs879284299 Chr.4: 76739512-76739512 on Build GRCh38
rs748600991 Chr.4: 76739506-76739506 on Build GRCh38
rs145159967 Chr.4: 76739274-76739274 on Build GRCh38
rs771119974 Chr.4: 76739316-76739316 on Build GRCh38
rs950302332 Chr.4: 76739277-76739277 on Build GRCh38
rs373907641 Chr.4: 76739541-76739541 on Build GRCh38
rs138986562 Chr.4: 76739524-76739524 on Build GRCh38
rs775873096 Chr.4: 76739327-76739327 on Build GRCh38
rs780849253 Chr.4: 76739170-76739170 on Build GRCh38
rs765691133 Chr.4: 76739117-76739117 on Build GRCh38
rs201725731 Chr.4: 76739360-76739360 on Build GRCh38
rs745862820 Chr.4: 76739175-76739175 on Build GRCh38
rs1024731294 Chr.4: 76739538-76739538 on Build GRCh38
rs768450600 Chr.4: 76739199-76739199 on Build GRCh38
rs776679144 Chr.4: 76739231-76739231 on Build GRCh38
rs541262431 Chr.4: 76739532-76739532 on Build GRCh38
rs764489203 Chr.4: 76739247-76739247 on Build GRCh38
rs756834979 Chr.4: 76739167-76739167 on Build GRCh38
rs758201409 Chr.4: 76739452-76739452 on Build GRCh38
rs771064814 Chr.4: 76739229-76739229 on Build GRCh38
rs766905572 Chr.4: 76739038-76739038 on Build GRCh38
rs61999292 Chr.4: 76739146-76739146 on Build GRCh38
rs148120221 Chr.4: 76739413-76739413 on Build GRCh38
rs201509477 Chr.4: 76739261-76739261 on Build GRCh38
rs560258820 Chr.4: 76739022-76739022 on Build GRCh38
rs917544216 Chr.4: 76739275-76739275 on Build GRCh38
rs967636557 Chr.4: 76739535-76739535 on Build GRCh38
rs756338847 Chr.4: 76739465-76739465 on Build GRCh38
rs769861836 Chr.4: 76739322-76739322 on Build GRCh38
rs372730468 Chr.4: 76739103-76739103 on Build GRCh38
rs201142744 Chr.4: 76739366-76739366 on Build GRCh38
rs772746533 Chr.4: 76739416-76739416 on Build GRCh38
rs756654865 Chr.4: 76739032-76739032 on Build GRCh38
rs956089417 Chr.4: 76739249-76739249 on Build GRCh38
rs771615558 Chr.4: 76739008-76739008 on Build GRCh38
rs770966223 Chr.4: 76739012-76739012 on Build GRCh38
rs200211229 Chr.4: 76739028-76739028 on Build GRCh38
rs776728183 Chr.4: 76739318-76739318 on Build GRCh38
rs754732664 Chr.4: 76739292-76739292 on Build GRCh38
rs191638826 Chr.4: 76739180-76739180 on Build GRCh38
rs768811288 Chr.4: 76739476-76739476 on Build GRCh38
rs146652221 Chr.4: 76739228-76739228 on Build GRCh38
rs374883048 Chr.4: 76739335-76739335 on Build GRCh38
rs79007254 Chr.4: 76739414-76739414 on Build GRCh38
rs551747532 Chr.4: 76739128-76739128 on Build GRCh38
rs344140 Chr.4: 76739009-76739009 on Build GRCh38
rs753205250 Chr.4: 76739239-76739239 on Build GRCh38
rs200056000 Chr.4: 76739350-76739350 on Build GRCh38
rs960118361 Chr.4: 76739510-76739510 on Build GRCh38
rs762145571 Chr.4: 76739102-76739102 on Build GRCh38
rs780038507 Chr.4: 76739471-76739471 on Build GRCh38
rs749869702 Chr.4: 76739273-76739273 on Build GRCh38
rs746483211 Chr.4: 76739062-76739062 on Build GRCh38
rs747288947 Chr.4: 76739311-76739311 on Build GRCh38
rs140762344 Chr.4: 76739114-76739114 on Build GRCh38

View Full Product Details