Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 188104845-188105435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIML2
Entrez Gene ID
205860
Gene Name
tripartite motif family like 2
Gene Aliases
SPRYD6
UniGene
Hs.276429
Gene Chromosome Location
Chr.4: 188091255-188109603 on Build GRCh38
Transcripts
NM_001303419.1 NM_173553.2 XM_011531737.2 XM_011531738.1 XM_011531739.1 XM_017007882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1429086 c.68C>T p.S23L Chr.4: 188104904-188104904 on Build GRCh38
4938132 c.193T>C p.L65L Chr.4: 188104929-188104929 on Build GRCh38
3396553 c.60G>T p.L20F Chr.4: 188104912-188104912 on Build GRCh38
5509362 c.7T>C p.C3R Chr.4: 188105212-188105212 on Build GRCh38
3602775 c.69G>A p.S23S Chr.4: 188104903-188104903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746224498 Chr.4: 188104985-188104985 on Build GRCh38
rs755335038 Chr.4: 188105149-188105149 on Build GRCh38
rs1032999369 Chr.4: 188105283-188105283 on Build GRCh38
rs752699324 Chr.4: 188105345-188105345 on Build GRCh38
rs778312039 Chr.4: 188105285-188105285 on Build GRCh38
rs971542196 Chr.4: 188105376-188105376 on Build GRCh38
rs553463960 Chr.4: 188105183-188105183 on Build GRCh38
rs923104069 Chr.4: 188105292-188105292 on Build GRCh38
rs571936655 Chr.4: 188105110-188105110 on Build GRCh38
rs771002495 Chr.4: 188105363-188105363 on Build GRCh38
rs758373008 Chr.4: 188105301-188105301 on Build GRCh38
rs528979615 Chr.4: 188104917-188104917 on Build GRCh38
rs975898737 Chr.4: 188105290-188105290 on Build GRCh38
rs763871243 Chr.4: 188105276-188105276 on Build GRCh38
rs752356233 Chr.4: 188105165-188105165 on Build GRCh38
rs540495175 Chr.4: 188105173-188105173 on Build GRCh38
rs903875764 Chr.4: 188105048-188105048 on Build GRCh38
rs779802083 Chr.4: 188105308-188105308 on Build GRCh38
rs763021367 Chr.4: 188105237-188105237 on Build GRCh38
rs748883522 Chr.4: 188105171-188105171 on Build GRCh38
rs773154565 Chr.4: 188105233-188105233 on Build GRCh38
rs773425392 Chr.4: 188105339-188105339 on Build GRCh38
rs768016477 Chr.4: 188105158-188105158 on Build GRCh38
rs537752263 Chr.4: 188105116-188105116 on Build GRCh38
rs1055249731 Chr.4: 188105094-188105094 on Build GRCh38
rs746866427 Chr.4: 188105329-188105329 on Build GRCh38
rs777369731 Chr.4: 188104936-188104936 on Build GRCh38
rs761816764 Chr.4: 188104881-188104881 on Build GRCh38
rs752006007 Chr.4: 188104909-188104909 on Build GRCh38
rs758977958 Chr.4: 188104904-188104904 on Build GRCh38
rs1013763958 Chr.4: 188105103-188105103 on Build GRCh38
rs762047906 Chr.4: 188104980-188104980 on Build GRCh38
rs781619330 Chr.4: 188105332-188105334 on Build GRCh38
rs750848428 Chr.4: 188104893-188104893 on Build GRCh38
rs753439996 Chr.4: 188105277-188105277 on Build GRCh38
rs747458824 Chr.4: 188105257-188105258 on Build GRCh38
rs765317542 Chr.4: 188104884-188104884 on Build GRCh38
rs759396523 Chr.4: 188104973-188104973 on Build GRCh38
rs780250981 Chr.4: 188104907-188104907 on Build GRCh38
rs535248684 Chr.4: 188105095-188105095 on Build GRCh38
rs753914974 Chr.4: 188104878-188104878 on Build GRCh38
rs759121188 Chr.4: 188104981-188104981 on Build GRCh38
rs773382468 Chr.4: 188105131-188105131 on Build GRCh38
rs371016184 Chr.4: 188104975-188104975 on Build GRCh38
rs769924905 Chr.4: 188105375-188105376 on Build GRCh38
rs919019901 Chr.4: 188105381-188105381 on Build GRCh38
rs749492501 Chr.4: 188105349-188105349 on Build GRCh38
rs1040643785 Chr.4: 188105155-188105155 on Build GRCh38
rs770947404 Chr.4: 188104949-188104949 on Build GRCh38
rs762580812 Chr.4: 188105102-188105102 on Build GRCh38
rs917423470 Chr.4: 188105016-188105016 on Build GRCh38
rs755811683 Chr.4: 188105274-188105274 on Build GRCh38
rs951745782 Chr.4: 188105184-188105184 on Build GRCh38
rs778845906 Chr.4: 188105178-188105178 on Build GRCh38
rs780923129 Chr.4: 188104913-188104913 on Build GRCh38
rs959255554 Chr.4: 188105316-188105316 on Build GRCh38
rs756736330 Chr.4: 188105280-188105280 on Build GRCh38
rs564751603 Chr.4: 188105387-188105387 on Build GRCh38
rs376306481 Chr.4: 188104968-188104968 on Build GRCh38
rs775295928 Chr.4: 188105207-188105207 on Build GRCh38
rs367563033 Chr.4: 188105168-188105168 on Build GRCh38
rs13139393 Chr.4: 188105370-188105370 on Build GRCh38
rs776779573 Chr.4: 188104976-188104976 on Build GRCh38
rs766380698 Chr.4: 188105255-188105255 on Build GRCh38
rs1040141125 Chr.4: 188104966-188104966 on Build GRCh38
rs992093223 Chr.4: 188104988-188104988 on Build GRCh38
rs376243917 Chr.4: 188105040-188105040 on Build GRCh38
rs758220829 Chr.4: 188105352-188105352 on Build GRCh38
rs1000854579 Chr.4: 188105080-188105080 on Build GRCh38
rs185517486 Chr.4: 188105147-188105147 on Build GRCh38
rs374542494 Chr.4: 188105134-188105134 on Build GRCh38
rs769905335 Chr.4: 188105332-188105332 on Build GRCh38
rs748392100 Chr.4: 188105359-188105359 on Build GRCh38
rs773370470 Chr.4: 188105399-188105400 on Build GRCh38
rs113404223 Chr.4: 188104931-188104931 on Build GRCh38
rs148885182 Chr.4: 188105154-188105154 on Build GRCh38
rs143492361 Chr.4: 188105170-188105170 on Build GRCh38
rs371443404 Chr.4: 188105204-188105204 on Build GRCh38
rs1036079689 Chr.4: 188105020-188105020 on Build GRCh38
rs767959404 Chr.4: 188105270-188105270 on Build GRCh38
rs780234921 Chr.4: 188104878-188104878 on Build GRCh38
rs761036503 Chr.4: 188105400-188105400 on Build GRCh38
rs28615703 Chr.4: 188105017-188105017 on Build GRCh38
rs529693724 Chr.4: 188104930-188104930 on Build GRCh38
rs754671947 Chr.4: 188104912-188104912 on Build GRCh38
rs753157392 Chr.4: 188105272-188105272 on Build GRCh38
rs573176023 Chr.4: 188105104-188105104 on Build GRCh38
rs748338339 Chr.4: 188105220-188105220 on Build GRCh38
rs755224766 Chr.4: 188105260-188105260 on Build GRCh38
rs116745036 Chr.4: 188105108-188105108 on Build GRCh38
rs771808902 Chr.4: 188105203-188105203 on Build GRCh38
rs745769155 Chr.4: 188105299-188105299 on Build GRCh38
rs967424830 Chr.4: 188105213-188105213 on Build GRCh38
rs1048682385 Chr.4: 188105105-188105105 on Build GRCh38
rs1042335639 Chr.4: 188105341-188105341 on Build GRCh38
rs984415910 Chr.4: 188105229-188105229 on Build GRCh38
rs112058466 Chr.4: 188105377-188105377 on Build GRCh38
rs374236853 Chr.4: 188104940-188104940 on Build GRCh38
rs774872763 Chr.4: 188105258-188105258 on Build GRCh38
rs1000222836 Chr.4: 188105260-188105260 on Build GRCh38
rs764361846 Chr.4: 188104867-188104867 on Build GRCh38
rs770105323 Chr.4: 188105223-188105223 on Build GRCh38

View Full Product Details