Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28147917-28148464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZKSCAN8
Entrez Gene ID
7745
Gene Name
zinc finger with KRAB and SCAN domains 8
Gene Aliases
LD5-1, ZNF192, ZSCAN40
UniGene
Hs.57679
Gene Chromosome Location
Chr.6: 28141644-28159472 on Build GRCh38
Transcripts
NM_001278119.1 NM_001278121.1 NM_001278122.1 NM_006298.3 XM_011514870.2 XM_011514873.2 XM_011514874.2 XM_017011266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3994783 c.7G>A p.E3K Chr.6: 28148414-28148414 on Build GRCh38
3994784 c.7G>A p.E3K Chr.6: 28148414-28148414 on Build GRCh38
6036795 c.34T>G p.S12A Chr.6: 28148441-28148441 on Build GRCh38
6036794 c.34T>G p.S12A Chr.6: 28148441-28148441 on Build GRCh38
1077026 c.24T>G p.P8P Chr.6: 28148431-28148431 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751761827 Chr.6: 28148358-28148358 on Build GRCh38
rs377289231 Chr.6: 28148403-28148403 on Build GRCh38
rs570060627 Chr.6: 28148233-28148233 on Build GRCh38
rs1007195957 Chr.6: 28148117-28148117 on Build GRCh38
rs112053323 Chr.6: 28148332-28148332 on Build GRCh38
rs902466711 Chr.6: 28147973-28147973 on Build GRCh38
rs756612582 Chr.6: 28148381-28148381 on Build GRCh38
rs374416230 Chr.6: 28148376-28148376 on Build GRCh38
rs749096573 Chr.6: 28148440-28148440 on Build GRCh38
rs914956249 Chr.6: 28148270-28148270 on Build GRCh38
rs959538586 Chr.6: 28148248-28148248 on Build GRCh38
rs900343526 Chr.6: 28148365-28148365 on Build GRCh38
rs765276486 Chr.6: 28148399-28148400 on Build GRCh38
rs112583577 Chr.6: 28148074-28148074 on Build GRCh38
rs754186096 Chr.6: 28148385-28148385 on Build GRCh38
rs1008807541 Chr.6: 28147994-28147994 on Build GRCh38
rs369053905 Chr.6: 28147944-28147944 on Build GRCh38
rs1024035388 Chr.6: 28148228-28148228 on Build GRCh38
rs984163847 Chr.6: 28148225-28148225 on Build GRCh38
rs935522600 Chr.6: 28148310-28148310 on Build GRCh38
rs4713148 Chr.6: 28148143-28148143 on Build GRCh38
rs961634109 Chr.6: 28148202-28148202 on Build GRCh38
rs891546386 Chr.6: 28148040-28148040 on Build GRCh38
rs745845523 Chr.6: 28148429-28148429 on Build GRCh38
rs753765315 Chr.6: 28148364-28148367 on Build GRCh38
rs938418168 Chr.6: 28147977-28147977 on Build GRCh38
rs769759921 Chr.6: 28148430-28148430 on Build GRCh38
rs754955132 Chr.6: 28148257-28148257 on Build GRCh38
rs1025721183 Chr.6: 28148169-28148169 on Build GRCh38
rs570479704 Chr.6: 28148082-28148082 on Build GRCh38
rs1042698673 Chr.6: 28147971-28147971 on Build GRCh38
rs1040286585 Chr.6: 28148353-28148353 on Build GRCh38
rs993959572 Chr.6: 28148168-28148168 on Build GRCh38
rs139971461 Chr.6: 28148319-28148319 on Build GRCh38
rs750924319 Chr.6: 28148378-28148378 on Build GRCh38
rs369500730 Chr.6: 28148405-28148405 on Build GRCh38
rs569837662 Chr.6: 28148214-28148214 on Build GRCh38
rs988300602 Chr.6: 28148337-28148337 on Build GRCh38
rs1040256217 Chr.6: 28148026-28148026 on Build GRCh38
rs535751156 Chr.6: 28148239-28148239 on Build GRCh38
rs755270774 Chr.6: 28148399-28148399 on Build GRCh38
rs778635581 Chr.6: 28148318-28148318 on Build GRCh38
rs145136963 Chr.6: 28148296-28148296 on Build GRCh38
rs200124524 Chr.6: 28148382-28148382 on Build GRCh38
rs868192779 Chr.6: 28148151-28148151 on Build GRCh38
rs759650086 Chr.6: 28148397-28148399 on Build GRCh38
rs763025748 Chr.6: 28148163-28148163 on Build GRCh38
rs761994134 Chr.6: 28148362-28148362 on Build GRCh38
rs565972276 Chr.6: 28148299-28148299 on Build GRCh38
rs149980108 Chr.6: 28148111-28148111 on Build GRCh38
rs900314492 Chr.6: 28148034-28148034 on Build GRCh38
rs780725960 Chr.6: 28148412-28148412 on Build GRCh38
rs990984788 Chr.6: 28148254-28148254 on Build GRCh38
rs944625078 Chr.6: 28148330-28148330 on Build GRCh38
rs143435480 Chr.6: 28148437-28148437 on Build GRCh38

View Full Product Details