Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 78944865-78945454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRAK1BP1
Entrez Gene ID
134728
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Gene Aliases
AIP70, SIMPL
UniGene
Hs.656212
Gene Chromosome Location
Chr.6: 78867486-78979411 on Build GRCh38
Transcripts
NM_017934.5 XM_005248729.4 XM_011535918.2 XM_017010989.1 XM_017010990.1

Gene Symbol
PHIP
Entrez Gene ID
55023
Gene Name
pleckstrin homology domain interacting protein
Gene Aliases
BRWD2, DCAF14, WDR11, ndrp
UniGene
Hs.511817
Gene Chromosome Location
Chr.6: 78934419-79078294 on Build GRCh38
Transcripts
NM_017934.5 XM_005248729.4 XM_011535918.2 XM_017010989.1 XM_017010990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3875666 c.4706G>A p.G1569D Chr.6: 78945422-78945422 on Build GRCh38
5579314 c.4682C>T p.P1561L Chr.6: 78945446-78945446 on Build GRCh38
1081843 c.4810T>C p.S1604P Chr.6: 78945318-78945318 on Build GRCh38
1081844 c.4790C>T p.A1597V Chr.6: 78945338-78945338 on Build GRCh38
421228 c.4700G>C p.S1567T Chr.6: 78945428-78945428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369317750 Chr.6: 78945314-78945314 on Build GRCh38
rs1037024018 Chr.6: 78945153-78945153 on Build GRCh38
rs758217287 Chr.6: 78944991-78944991 on Build GRCh38
rs946461902 Chr.6: 78945051-78945051 on Build GRCh38
rs747531541 Chr.6: 78945294-78945294 on Build GRCh38
rs758863569 Chr.6: 78945277-78945277 on Build GRCh38
rs183087779 Chr.6: 78944939-78944939 on Build GRCh38
rs766905699 Chr.6: 78944964-78944964 on Build GRCh38
rs889932182 Chr.6: 78945188-78945188 on Build GRCh38
rs192244789 Chr.6: 78945184-78945184 on Build GRCh38
rs556842979 Chr.6: 78945421-78945421 on Build GRCh38
rs577476778 Chr.6: 78945284-78945284 on Build GRCh38
rs745460278 Chr.6: 78945276-78945276 on Build GRCh38
rs760455292 Chr.6: 78945368-78945368 on Build GRCh38
rs199510464 Chr.6: 78945382-78945382 on Build GRCh38
rs554532155 Chr.6: 78945093-78945093 on Build GRCh38
rs775416929 Chr.6: 78945379-78945384 on Build GRCh38
rs752037326 Chr.6: 78945405-78945405 on Build GRCh38
rs879391783 Chr.6: 78945083-78945083 on Build GRCh38
rs374117949 Chr.6: 78945345-78945345 on Build GRCh38
rs898504619 Chr.6: 78945159-78945159 on Build GRCh38
rs992674304 Chr.6: 78945181-78945181 on Build GRCh38
rs954357446 Chr.6: 78944990-78944990 on Build GRCh38
rs1020298689 Chr.6: 78945003-78945003 on Build GRCh38
rs925110447 Chr.6: 78944925-78944925 on Build GRCh38
rs568815174 Chr.6: 78945052-78945052 on Build GRCh38
rs936610248 Chr.6: 78945262-78945262 on Build GRCh38
rs1052143218 Chr.6: 78944993-78944993 on Build GRCh38
rs753693081 Chr.6: 78945376-78945376 on Build GRCh38
rs1019804960 Chr.6: 78945288-78945288 on Build GRCh38
rs758065132 Chr.6: 78945386-78945386 on Build GRCh38
rs5877636 Chr.6: 78944968-78944969 on Build GRCh38
rs773509249 Chr.6: 78945335-78945335 on Build GRCh38
rs76962186 Chr.6: 78945166-78945166 on Build GRCh38
rs756812200 Chr.6: 78945379-78945379 on Build GRCh38
rs777273493 Chr.6: 78945312-78945312 on Build GRCh38
rs888800175 Chr.6: 78945060-78945060 on Build GRCh38
rs980677440 Chr.6: 78945205-78945205 on Build GRCh38
rs866648472 Chr.6: 78945304-78945304 on Build GRCh38
rs925192468 Chr.6: 78945248-78945248 on Build GRCh38
rs756688401 Chr.6: 78945269-78945269 on Build GRCh38
rs753450619 Chr.6: 78945258-78945258 on Build GRCh38
rs969804634 Chr.6: 78945191-78945191 on Build GRCh38
rs937863161 Chr.6: 78945082-78945082 on Build GRCh38
rs769741723 Chr.6: 78945334-78945334 on Build GRCh38
rs878942622 Chr.6: 78945097-78945097 on Build GRCh38
rs375420570 Chr.6: 78945038-78945038 on Build GRCh38
rs896382501 Chr.6: 78945147-78945147 on Build GRCh38
rs146537039 Chr.6: 78945020-78945020 on Build GRCh38
rs769344926 Chr.6: 78945302-78945302 on Build GRCh38
rs778829949 Chr.6: 78944903-78944903 on Build GRCh38
rs750095261 Chr.6: 78945384-78945384 on Build GRCh38
rs926441361 Chr.6: 78945078-78945078 on Build GRCh38
rs781748135 Chr.6: 78945424-78945424 on Build GRCh38
rs1008798905 Chr.6: 78945197-78945197 on Build GRCh38
rs1056356002 Chr.6: 78945096-78945096 on Build GRCh38
rs397740182 Chr.6: 78944973-78944974 on Build GRCh38
rs951454145 Chr.6: 78945167-78945167 on Build GRCh38
rs869145756 Chr.6: 78944974-78944975 on Build GRCh38
rs537777229 Chr.6: 78945068-78945068 on Build GRCh38
rs748557453 Chr.6: 78945425-78945425 on Build GRCh38
rs956317852 Chr.6: 78945005-78945005 on Build GRCh38
rs764056825 Chr.6: 78945369-78945369 on Build GRCh38
rs747312156 Chr.6: 78945338-78945338 on Build GRCh38
rs775318448 Chr.6: 78945348-78945348 on Build GRCh38
rs888753822 Chr.6: 78945400-78945400 on Build GRCh38
rs187916911 Chr.6: 78945049-78945049 on Build GRCh38
rs9688399 Chr.6: 78944938-78944938 on Build GRCh38
rs779841096 Chr.6: 78945392-78945392 on Build GRCh38
rs778673906 Chr.6: 78945270-78945270 on Build GRCh38
rs543814235 Chr.6: 78945295-78945295 on Build GRCh38
rs116682867 Chr.6: 78945148-78945148 on Build GRCh38

View Full Product Details