Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149810800-149811396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCMT1
Entrez Gene ID
5110
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase
Gene Aliases
PIMT
UniGene
Hs.279257
Gene Chromosome Location
Chr.6: 149749695-149811421 on Build GRCh38
Transcripts
NM_001252049.1 NM_001252050.1 NM_001252051.1 NM_001252052.1 NM_005389.2 XM_011535868.2

SNP Detail

SNP ID Chromosome Location
rs904958636 Chr.6: 149811119-149811119 on Build GRCh38
rs4552 Chr.6: 149811183-149811183 on Build GRCh38
rs984496547 Chr.6: 149811363-149811363 on Build GRCh38
rs1018985998 Chr.6: 149811102-149811102 on Build GRCh38
rs563861446 Chr.6: 149811366-149811366 on Build GRCh38
rs541565200 Chr.6: 149811210-149811210 on Build GRCh38
rs374644536 Chr.6: 149811044-149811044 on Build GRCh38
rs1013322090 Chr.6: 149811066-149811066 on Build GRCh38
rs3209824 Chr.6: 149810880-149810880 on Build GRCh38
rs1045442408 Chr.6: 149811330-149811330 on Build GRCh38
rs1019389435 Chr.6: 149811238-149811238 on Build GRCh38
rs926992916 Chr.6: 149811336-149811336 on Build GRCh38
rs925897247 Chr.6: 149811197-149811197 on Build GRCh38
rs542219855 Chr.6: 149810854-149810854 on Build GRCh38
rs193012643 Chr.6: 149811225-149811225 on Build GRCh38
rs551889168 Chr.6: 149810836-149810836 on Build GRCh38
rs995019218 Chr.6: 149810991-149810991 on Build GRCh38
rs975927332 Chr.6: 149810841-149810841 on Build GRCh38
rs975287272 Chr.6: 149811265-149811265 on Build GRCh38
rs1002437605 Chr.6: 149810944-149810944 on Build GRCh38
rs949778015 Chr.6: 149810917-149810917 on Build GRCh38
rs371044437 Chr.6: 149811005-149811005 on Build GRCh38
rs1804171 Chr.6: 149811307-149811307 on Build GRCh38
rs565630056 Chr.6: 149810842-149810842 on Build GRCh38
rs949824945 Chr.6: 149811318-149811318 on Build GRCh38
rs544054488 Chr.6: 149811362-149811362 on Build GRCh38
rs1031975619 Chr.6: 149811181-149811181 on Build GRCh38
rs185340125 Chr.6: 149810972-149810972 on Build GRCh38
rs7818 Chr.6: 149810956-149810956 on Build GRCh38
rs935846903 Chr.6: 149810825-149810825 on Build GRCh38
rs905516984 Chr.6: 149810938-149810938 on Build GRCh38
rs189051612 Chr.6: 149811085-149811085 on Build GRCh38
rs562572933 Chr.6: 149810890-149810890 on Build GRCh38
rs891553756 Chr.6: 149810896-149810896 on Build GRCh38
rs1045392979 Chr.6: 149810933-149810933 on Build GRCh38
rs756157655 Chr.6: 149811259-149811259 on Build GRCh38
rs574953393 Chr.6: 149811309-149811309 on Build GRCh38
rs558719308 Chr.6: 149811152-149811152 on Build GRCh38
rs965213515 Chr.6: 149811254-149811257 on Build GRCh38
rs139545012 Chr.6: 149811041-149811041 on Build GRCh38
rs765127276 Chr.6: 149810862-149810862 on Build GRCh38
rs536862649 Chr.6: 149811014-149811014 on Build GRCh38
rs767661380 Chr.6: 149811112-149811112 on Build GRCh38
rs766536636 Chr.6: 149811341-149811341 on Build GRCh38
rs964700006 Chr.6: 149811182-149811182 on Build GRCh38
rs774766706 Chr.6: 149811371-149811372 on Build GRCh38
rs1033925877 Chr.6: 149810955-149810955 on Build GRCh38

View Full Product Details