Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99908672-99909213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM4
Entrez Gene ID
89122
Gene Name
tripartite motif containing 4
Gene Aliases
RNF87
UniGene
Hs.50749
Gene Chromosome Location
Chr.7: 99890407-99919600 on Build GRCh38
Transcripts
NM_033017.3 NM_033091.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5467271 c.608A>C p.K203T Chr.7: 99908694-99908694 on Build GRCh38
7169116 c.640G>C p.E214Q Chr.7: 99908740-99908740 on Build GRCh38
3382033 c.684G>A p.T228T Chr.7: 99908696-99908696 on Build GRCh38
6808194 c.540G>A p.L180L Chr.7: 99908762-99908762 on Build GRCh38
4810361 c.568C>A p.L190M Chr.7: 99908734-99908734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868327923 Chr.7: 99908777-99908777 on Build GRCh38
rs914558925 Chr.7: 99909128-99909133 on Build GRCh38
rs147278816 Chr.7: 99908778-99908778 on Build GRCh38
rs563588351 Chr.7: 99908960-99908960 on Build GRCh38
rs547751972 Chr.7: 99909157-99909157 on Build GRCh38
rs893277277 Chr.7: 99909184-99909184 on Build GRCh38
rs893393521 Chr.7: 99909018-99909018 on Build GRCh38
rs762296876 Chr.7: 99908821-99908821 on Build GRCh38
rs57960671 Chr.7: 99909158-99909159 on Build GRCh38
rs191638127 Chr.7: 99909160-99909160 on Build GRCh38
rs781285227 Chr.7: 99908842-99908842 on Build GRCh38
rs1016735526 Chr.7: 99909133-99909133 on Build GRCh38
rs113050838 Chr.7: 99909102-99909347 on Build GRCh38
rs781090856 Chr.7: 99908775-99908775 on Build GRCh38
rs953479263 Chr.7: 99909105-99909105 on Build GRCh38
rs991636097 Chr.7: 99909137-99909137 on Build GRCh38
rs574772627 Chr.7: 99909135-99909135 on Build GRCh38
rs767897065 Chr.7: 99908707-99908707 on Build GRCh38
rs937857330 Chr.7: 99909145-99909146 on Build GRCh38
rs986910334 Chr.7: 99909003-99909006 on Build GRCh38
rs1036689187 Chr.7: 99908697-99908697 on Build GRCh38
rs201446854 Chr.7: 99909159-99909159 on Build GRCh38
rs144840385 Chr.7: 99908980-99908980 on Build GRCh38
rs1054597344 Chr.7: 99909017-99909017 on Build GRCh38
rs1029532747 Chr.7: 99909101-99909101 on Build GRCh38
rs371305209 Chr.7: 99908801-99908801 on Build GRCh38
rs140032652 Chr.7: 99909133-99909134 on Build GRCh38
rs1036918803 Chr.7: 99909178-99909180 on Build GRCh38
rs757828171 Chr.7: 99908826-99908826 on Build GRCh38
rs752417758 Chr.7: 99908847-99908847 on Build GRCh38
rs867877343 Chr.7: 99908736-99908736 on Build GRCh38
rs909821835 Chr.7: 99908882-99908882 on Build GRCh38
rs138155062 Chr.7: 99908704-99908704 on Build GRCh38
rs879130499 Chr.7: 99908723-99908723 on Build GRCh38
rs149554361 Chr.7: 99908744-99908744 on Build GRCh38
rs1002148428 Chr.7: 99909012-99909012 on Build GRCh38
rs535824192 Chr.7: 99909161-99909161 on Build GRCh38
rs549969743 Chr.7: 99908999-99908999 on Build GRCh38
rs949722819 Chr.7: 99909158-99909158 on Build GRCh38
rs947342258 Chr.7: 99909134-99909137 on Build GRCh38
rs1036700355 Chr.7: 99908949-99908949 on Build GRCh38
rs770997658 Chr.7: 99908781-99908781 on Build GRCh38
rs937068113 Chr.7: 99909164-99909164 on Build GRCh38
rs530079849 Chr.7: 99908840-99908840 on Build GRCh38
rs995224062 Chr.7: 99908969-99908969 on Build GRCh38
rs962315666 Chr.7: 99908951-99908951 on Build GRCh38
rs745931126 Chr.7: 99908793-99908793 on Build GRCh38
rs66953435 Chr.7: 99909177-99909177 on Build GRCh38
rs972616090 Chr.7: 99909145-99909145 on Build GRCh38
rs901882832 Chr.7: 99909060-99909060 on Build GRCh38
rs148577892 Chr.7: 99909078-99909078 on Build GRCh38
rs1020255495 Chr.7: 99909040-99909040 on Build GRCh38
rs776042163 Chr.7: 99908712-99908712 on Build GRCh38
rs187483585 Chr.7: 99909131-99909131 on Build GRCh38
rs1015675791 Chr.7: 99908940-99908940 on Build GRCh38
rs764590844 Chr.7: 99908808-99908808 on Build GRCh38
rs764796513 Chr.7: 99908824-99908824 on Build GRCh38
rs918231462 Chr.7: 99908970-99908970 on Build GRCh38
rs896444916 Chr.7: 99909160-99909163 on Build GRCh38
rs760994998 Chr.7: 99908714-99908714 on Build GRCh38
rs754985807 Chr.7: 99908762-99908762 on Build GRCh38
rs927343474 Chr.7: 99909134-99909145 on Build GRCh38
rs757996629 Chr.7: 99908753-99908753 on Build GRCh38
rs754233219 Chr.7: 99908728-99908728 on Build GRCh38
rs956481029 Chr.7: 99909089-99909089 on Build GRCh38
rs746994341 Chr.7: 99908760-99908760 on Build GRCh38
rs905593911 Chr.7: 99909001-99909001 on Build GRCh38
rs200389520 Chr.7: 99908755-99908755 on Build GRCh38
rs149813794 Chr.7: 99908920-99908920 on Build GRCh38
rs985310448 Chr.7: 99908879-99908879 on Build GRCh38
rs758310378 Chr.7: 99908822-99908822 on Build GRCh38
rs765941379 Chr.7: 99908837-99908837 on Build GRCh38
rs545481671 Chr.7: 99909101-99909105 on Build GRCh38
rs764956366 Chr.7: 99908738-99908738 on Build GRCh38
rs957611685 Chr.7: 99908780-99908780 on Build GRCh38
rs376720616 Chr.7: 99908789-99908789 on Build GRCh38
rs774621750 Chr.7: 99908700-99908702 on Build GRCh38
rs896380886 Chr.7: 99908907-99908907 on Build GRCh38
rs756003291 Chr.7: 99908859-99908859 on Build GRCh38
rs541353597 Chr.7: 99908735-99908735 on Build GRCh38
rs139121700 Chr.7: 99908794-99908794 on Build GRCh38
rs556375779 Chr.7: 99908986-99908986 on Build GRCh38
rs3220811 Chr.7: 99909127-99909128 on Build GRCh38
rs775952000 Chr.7: 99908799-99908799 on Build GRCh38
rs776923095 Chr.7: 99908813-99908813 on Build GRCh38
rs764394942 Chr.7: 99908715-99908715 on Build GRCh38
rs1044518969 Chr.7: 99909134-99909139 on Build GRCh38
rs757233379 Chr.7: 99909035-99909035 on Build GRCh38
rs759462306 Chr.7: 99909157-99909158 on Build GRCh38

View Full Product Details