Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 74849000-74849521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PI15
Entrez Gene ID
51050
Gene Name
peptidase inhibitor 15
Gene Aliases
CRISP8, P24TI, P25TI
UniGene
Hs.98558
Gene Chromosome Location
Chr.8: 74824537-74855044 on Build GRCh38
Transcripts
NM_001324403.1 NM_015886.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4929282 c.706T>A p.Y236N Chr.8: 74849182-74849182 on Build GRCh38
6745730 c.666A>G p.P222P Chr.8: 74849142-74849142 on Build GRCh38
5998017 c.755A>G p.N252S Chr.8: 74849231-74849231 on Build GRCh38
1314200 c.686G>T p.C229F Chr.8: 74849162-74849162 on Build GRCh38
35448 c.733T>A p.C245S Chr.8: 74849209-74849209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536342698 Chr.8: 74849250-74849250 on Build GRCh38
rs745795218 Chr.8: 74849096-74849096 on Build GRCh38
rs373126603 Chr.8: 74849226-74849226 on Build GRCh38
rs779915149 Chr.8: 74849128-74849128 on Build GRCh38
rs549148253 Chr.8: 74849041-74849041 on Build GRCh38
rs755792224 Chr.8: 74849076-74849076 on Build GRCh38
rs746717516 Chr.8: 74849136-74849136 on Build GRCh38
rs903267948 Chr.8: 74849208-74849208 on Build GRCh38
rs764990189 Chr.8: 74849405-74849405 on Build GRCh38
rs756806200 Chr.8: 74849080-74849080 on Build GRCh38
rs141226749 Chr.8: 74849303-74849304 on Build GRCh38
rs937963495 Chr.8: 74849313-74849313 on Build GRCh38
rs138348649 Chr.8: 74849322-74849322 on Build GRCh38
rs77715425 Chr.8: 74849160-74849160 on Build GRCh38
rs1015447145 Chr.8: 74849359-74849359 on Build GRCh38
rs773374926 Chr.8: 74849149-74849149 on Build GRCh38
rs571878620 Chr.8: 74849111-74849111 on Build GRCh38
rs200082533 Chr.8: 74849295-74849295 on Build GRCh38
rs758362523 Chr.8: 74849239-74849239 on Build GRCh38
rs201645030 Chr.8: 74849094-74849094 on Build GRCh38
rs772987829 Chr.8: 74849485-74849485 on Build GRCh38
rs781195240 Chr.8: 74849305-74849306 on Build GRCh38
rs774416854 Chr.8: 74849297-74849297 on Build GRCh38
rs955203998 Chr.8: 74849042-74849042 on Build GRCh38
rs937662038 Chr.8: 74849450-74849450 on Build GRCh38
rs141213589 Chr.8: 74849184-74849184 on Build GRCh38
rs146938685 Chr.8: 74849189-74849189 on Build GRCh38
rs955315919 Chr.8: 74849339-74849339 on Build GRCh38
rs764727741 Chr.8: 74849206-74849207 on Build GRCh38
rs371348737 Chr.8: 74849268-74849268 on Build GRCh38
rs767843743 Chr.8: 74849178-74849178 on Build GRCh38
rs763335450 Chr.8: 74849098-74849098 on Build GRCh38
rs962452650 Chr.8: 74849394-74849394 on Build GRCh38
rs753159184 Chr.8: 74849079-74849079 on Build GRCh38
rs991809065 Chr.8: 74849467-74849467 on Build GRCh38
rs200826695 Chr.8: 74849225-74849225 on Build GRCh38
rs749892777 Chr.8: 74849236-74849236 on Build GRCh38
rs768314134 Chr.8: 74849260-74849260 on Build GRCh38
rs375982020 Chr.8: 74849235-74849235 on Build GRCh38
rs117775004 Chr.8: 74849296-74849296 on Build GRCh38
rs763852974 Chr.8: 74849217-74849217 on Build GRCh38
rs982637330 Chr.8: 74849059-74849059 on Build GRCh38
rs752945036 Chr.8: 74849067-74849067 on Build GRCh38
rs888329151 Chr.8: 74849340-74849340 on Build GRCh38
rs368284096 Chr.8: 74849066-74849066 on Build GRCh38
rs1055985073 Chr.8: 74849157-74849157 on Build GRCh38
rs760077771 Chr.8: 74849161-74849161 on Build GRCh38
rs896102142 Chr.8: 74849159-74849159 on Build GRCh38
rs79847015 Chr.8: 74849300-74849300 on Build GRCh38
rs753416574 Chr.8: 74849079-74849079 on Build GRCh38
rs772651702 Chr.8: 74849302-74849302 on Build GRCh38
rs770086861 Chr.8: 74849137-74849137 on Build GRCh38
rs775124486 Chr.8: 74849024-74849024 on Build GRCh38
rs796328156 Chr.8: 74849304-74849305 on Build GRCh38
rs763073877 Chr.8: 74849158-74849158 on Build GRCh38
rs982606221 Chr.8: 74849346-74849346 on Build GRCh38
rs866370079 Chr.8: 74849231-74849231 on Build GRCh38
rs777248249 Chr.8: 74849078-74849078 on Build GRCh38
rs151180210 Chr.8: 74849155-74849155 on Build GRCh38
rs749511723 Chr.8: 74849285-74849285 on Build GRCh38
rs775834418 Chr.8: 74849305-74849305 on Build GRCh38
rs561303991 Chr.8: 74849056-74849056 on Build GRCh38
rs34728422 Chr.8: 74849060-74849060 on Build GRCh38
rs926281518 Chr.8: 74849430-74849430 on Build GRCh38
rs555074510 Chr.8: 74849259-74849259 on Build GRCh38
rs760895558 Chr.8: 74849199-74849199 on Build GRCh38

View Full Product Details