Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98949415-98949984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSR2
Entrez Gene ID
116039
Gene Name
odd-skipped related transciption factor 2
Gene Aliases
-
UniGene
Hs.253247
Gene Chromosome Location
Chr.8: 98944403-98952104 on Build GRCh38
Transcripts
NM_001142462.2 NM_001286841.1 NM_053001.3 XM_005250778.4 XM_011516825.2 XM_011516826.2 XM_011516827.2 XM_017013018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6735929 c.588C>T p.D196D Chr.8: 98949540-98949540 on Build GRCh38
2792756 c.1010G>A p.R337Q Chr.8: 98949599-98949599 on Build GRCh38
54553 c.524A>G p.K175R Chr.8: 98949476-98949476 on Build GRCh38
5029336 c.551C>G p.S184C Chr.8: 98949503-98949503 on Build GRCh38
2792752 c.947C>T p.T316M Chr.8: 98949536-98949536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374378552 Chr.8: 98949528-98949528 on Build GRCh38
rs945249536 Chr.8: 98949786-98949786 on Build GRCh38
rs576653383 Chr.8: 98949665-98949665 on Build GRCh38
rs770191972 Chr.8: 98949817-98949817 on Build GRCh38
rs771173600 Chr.8: 98949585-98949585 on Build GRCh38
rs936856912 Chr.8: 98949948-98949948 on Build GRCh38
rs961657979 Chr.8: 98949450-98949450 on Build GRCh38
rs370918198 Chr.8: 98949614-98949614 on Build GRCh38
rs917603848 Chr.8: 98949845-98949845 on Build GRCh38
rs767562127 Chr.8: 98949437-98949437 on Build GRCh38
rs778807175 Chr.8: 98949483-98949483 on Build GRCh38
rs898857518 Chr.8: 98949846-98949846 on Build GRCh38
rs559637087 Chr.8: 98949743-98949743 on Build GRCh38
rs554677790 Chr.8: 98949468-98949468 on Build GRCh38
rs975881431 Chr.8: 98949680-98949680 on Build GRCh38
rs181820566 Chr.8: 98949864-98949864 on Build GRCh38
rs139966789 Chr.8: 98949646-98949646 on Build GRCh38
rs965901764 Chr.8: 98949960-98949960 on Build GRCh38
rs370468254 Chr.8: 98949850-98949850 on Build GRCh38
rs778734770 Chr.8: 98949633-98949633 on Build GRCh38
rs528385044 Chr.8: 98949796-98949796 on Build GRCh38
rs1042151076 Chr.8: 98949890-98949890 on Build GRCh38
rs750189447 Chr.8: 98949643-98949643 on Build GRCh38
rs937787701 Chr.8: 98949826-98949826 on Build GRCh38
rs1027222358 Chr.8: 98949564-98949564 on Build GRCh38
rs767688551 Chr.8: 98949615-98949615 on Build GRCh38
rs778045899 Chr.8: 98949442-98949442 on Build GRCh38
rs574548072 Chr.8: 98949538-98949538 on Build GRCh38
rs1054867005 Chr.8: 98949839-98949839 on Build GRCh38
rs545686617 Chr.8: 98949738-98949738 on Build GRCh38
rs992026775 Chr.8: 98949802-98949802 on Build GRCh38
rs757268256 Chr.8: 98949622-98949622 on Build GRCh38
rs924989747 Chr.8: 98949938-98949938 on Build GRCh38
rs375217129 Chr.8: 98949806-98949806 on Build GRCh38
rs560352225 Chr.8: 98949742-98949742 on Build GRCh38
rs763588260 Chr.8: 98949554-98949554 on Build GRCh38
rs1046231500 Chr.8: 98949798-98949798 on Build GRCh38
rs959319175 Chr.8: 98949687-98949687 on Build GRCh38
rs370955340 Chr.8: 98949435-98949435 on Build GRCh38
rs1049959657 Chr.8: 98949954-98949954 on Build GRCh38
rs989393996 Chr.8: 98949770-98949770 on Build GRCh38
rs769169543 Chr.8: 98949455-98949455 on Build GRCh38
rs1019800282 Chr.8: 98949955-98949955 on Build GRCh38
rs745486458 Chr.8: 98949504-98949504 on Build GRCh38
rs756996874 Chr.8: 98949474-98949474 on Build GRCh38
rs543533750 Chr.8: 98949618-98949618 on Build GRCh38
rs771797861 Chr.8: 98949522-98949522 on Build GRCh38
rs748480984 Chr.8: 98949735-98949735 on Build GRCh38
rs913811439 Chr.8: 98949785-98949785 on Build GRCh38
rs752923643 Chr.8: 98949616-98949616 on Build GRCh38
rs773582439 Chr.8: 98949553-98949553 on Build GRCh38
rs376720519 Chr.8: 98949608-98949608 on Build GRCh38
rs753757117 Chr.8: 98949647-98949648 on Build GRCh38
rs751730409 Chr.8: 98949434-98949434 on Build GRCh38
rs768292533 Chr.8: 98949549-98949549 on Build GRCh38
rs755014549 Chr.8: 98949606-98949606 on Build GRCh38
rs994503546 Chr.8: 98949875-98949875 on Build GRCh38

View Full Product Details