Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57672264-57672855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC5
Entrez Gene ID
25921
Gene Name
zinc finger DHHC-type containing 5
Gene Aliases
DHHC5, ZNF375
UniGene
Hs.27239
Gene Chromosome Location
Chr.11: 57667836-57701187 on Build GRCh38
Transcripts
NM_015457.2 XM_011544899.1 XM_011544900.1 XM_011544901.1 XM_017017498.1

SNP Detail

SNP ID Chromosome Location
rs181866738 Chr.11: 57672529-57672529 on Build GRCh38
rs1025403505 Chr.11: 57672327-57672327 on Build GRCh38
rs975321285 Chr.11: 57672594-57672594 on Build GRCh38
rs1024632973 Chr.11: 57672369-57672369 on Build GRCh38
rs758784427 Chr.11: 57672631-57672631 on Build GRCh38
rs967424490 Chr.11: 57672471-57672471 on Build GRCh38
rs560581553 Chr.11: 57672618-57672618 on Build GRCh38
rs534917191 Chr.11: 57672816-57672816 on Build GRCh38
rs888208673 Chr.11: 57672540-57672540 on Build GRCh38
rs999602568 Chr.11: 57672625-57672625 on Build GRCh38
rs911063704 Chr.11: 57672759-57672759 on Build GRCh38
rs977117201 Chr.11: 57672763-57672763 on Build GRCh38
rs1034584639 Chr.11: 57672781-57672781 on Build GRCh38
rs1001755294 Chr.11: 57672401-57672405 on Build GRCh38
rs535152146 Chr.11: 57672638-57672638 on Build GRCh38
rs929662288 Chr.11: 57672811-57672811 on Build GRCh38
rs572203699 Chr.11: 57672663-57672664 on Build GRCh38
rs3741076 Chr.11: 57672646-57672646 on Build GRCh38
rs543215720 Chr.11: 57672350-57672350 on Build GRCh38
rs986799570 Chr.11: 57672522-57672522 on Build GRCh38
rs963988888 Chr.11: 57672760-57672760 on Build GRCh38
rs780989770 Chr.11: 57672446-57672446 on Build GRCh38
rs989234100 Chr.11: 57672750-57672750 on Build GRCh38
rs143672960 Chr.11: 57672687-57672687 on Build GRCh38
rs757607622 Chr.11: 57672541-57672543 on Build GRCh38
rs1037874001 Chr.11: 57672588-57672588 on Build GRCh38
rs753042674 Chr.11: 57672466-57672466 on Build GRCh38
rs540946209 Chr.11: 57672612-57672612 on Build GRCh38
rs745512679 Chr.11: 57672791-57672791 on Build GRCh38
rs185338437 Chr.11: 57672597-57672597 on Build GRCh38
rs995669707 Chr.11: 57672289-57672289 on Build GRCh38
rs971672772 Chr.11: 57672371-57672371 on Build GRCh38
rs904827332 Chr.11: 57672729-57672729 on Build GRCh38
rs895364673 Chr.11: 57672592-57672592 on Build GRCh38
rs1013280125 Chr.11: 57672363-57672363 on Build GRCh38
rs957895854 Chr.11: 57672430-57672430 on Build GRCh38
rs1006368418 Chr.11: 57672576-57672576 on Build GRCh38
rs779433762 Chr.11: 57672402-57672402 on Build GRCh38
rs959627249 Chr.11: 57672293-57672293 on Build GRCh38
rs1046872755 Chr.11: 57672517-57672517 on Build GRCh38
rs942616036 Chr.11: 57672458-57672458 on Build GRCh38
rs990794385 Chr.11: 57672303-57672303 on Build GRCh38
rs923756902 Chr.11: 57672416-57672416 on Build GRCh38
rs929740529 Chr.11: 57672505-57672505 on Build GRCh38
rs942621944 Chr.11: 57672566-57672566 on Build GRCh38
rs886792838 Chr.11: 57672330-57672330 on Build GRCh38
rs971158980 Chr.11: 57672619-57672619 on Build GRCh38
rs893362849 Chr.11: 57672827-57672827 on Build GRCh38
rs576966622 Chr.11: 57672548-57672548 on Build GRCh38
rs911114551 Chr.11: 57672457-57672457 on Build GRCh38
rs118112861 Chr.11: 57672644-57672644 on Build GRCh38
rs922296859 Chr.11: 57672479-57672479 on Build GRCh38
rs957532833 Chr.11: 57672405-57672405 on Build GRCh38

View Full Product Details