Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59123786-59124314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM111B
Entrez Gene ID
374393
Gene Name
family with sequence similarity 111 member B
Gene Aliases
CANP, POIKTMP
UniGene
Hs.186579
Gene Chromosome Location
Chr.11: 59107185-59127415 on Build GRCh38
Transcripts
NM_001142703.1 NM_001142704.1 NM_198947.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5050928 c.114A>G p.T38T Chr.11: 59124211-59124211 on Build GRCh38
4833469 c.175G>A p.E59K Chr.11: 59124272-59124272 on Build GRCh38
7366946 c.190G>A p.E64K Chr.11: 59124287-59124287 on Build GRCh38
1354957 c.191A>C p.E64A Chr.11: 59124288-59124288 on Build GRCh38
5372893 c.152G>A p.S51N Chr.11: 59124249-59124249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754136839 Chr.11: 59124131-59124133 on Build GRCh38
rs369485601 Chr.11: 59124234-59124234 on Build GRCh38
rs950675463 Chr.11: 59124055-59124055 on Build GRCh38
rs148727780 Chr.11: 59124283-59124283 on Build GRCh38
rs770514961 Chr.11: 59124225-59124225 on Build GRCh38
rs558041395 Chr.11: 59123865-59123865 on Build GRCh38
rs774916118 Chr.11: 59124263-59124263 on Build GRCh38
rs995551608 Chr.11: 59123970-59123970 on Build GRCh38
rs564290979 Chr.11: 59123997-59124001 on Build GRCh38
rs543937326 Chr.11: 59124031-59124031 on Build GRCh38
rs866912203 Chr.11: 59123946-59123946 on Build GRCh38
rs779147467 Chr.11: 59124145-59124147 on Build GRCh38
rs555039161 Chr.11: 59123976-59123976 on Build GRCh38
rs752830956 Chr.11: 59124170-59124175 on Build GRCh38
rs932922219 Chr.11: 59123920-59123920 on Build GRCh38
rs575386520 Chr.11: 59124183-59124183 on Build GRCh38
rs373130212 Chr.11: 59124073-59124073 on Build GRCh38
rs377469298 Chr.11: 59124044-59124045 on Build GRCh38
rs201011410 Chr.11: 59124258-59124258 on Build GRCh38
rs897129656 Chr.11: 59123965-59123965 on Build GRCh38
rs765615695 Chr.11: 59124181-59124181 on Build GRCh38
rs144378733 Chr.11: 59124261-59124261 on Build GRCh38
rs990971068 Chr.11: 59124125-59124125 on Build GRCh38
rs563928683 Chr.11: 59124123-59124123 on Build GRCh38
rs777620417 Chr.11: 59124216-59124216 on Build GRCh38
rs1005164559 Chr.11: 59124078-59124078 on Build GRCh38
rs762038298 Chr.11: 59124179-59124179 on Build GRCh38
rs764519594 Chr.11: 59124172-59124172 on Build GRCh38
rs1055103116 Chr.11: 59123875-59123875 on Build GRCh38
rs1039531586 Chr.11: 59123979-59123979 on Build GRCh38
rs773999761 Chr.11: 59124230-59124230 on Build GRCh38
rs201851649 Chr.11: 59124140-59124140 on Build GRCh38
rs555482770 Chr.11: 59124062-59124062 on Build GRCh38
rs533825570 Chr.11: 59123981-59123981 on Build GRCh38
rs796371171 Chr.11: 59124155-59124155 on Build GRCh38
rs780941789 Chr.11: 59124199-59124199 on Build GRCh38
rs755138806 Chr.11: 59124189-59124189 on Build GRCh38
rs377492266 Chr.11: 59124293-59124293 on Build GRCh38
rs1030375263 Chr.11: 59124009-59124009 on Build GRCh38
rs766666078 Chr.11: 59124286-59124286 on Build GRCh38
rs755225765 Chr.11: 59124131-59124132 on Build GRCh38
rs774173620 Chr.11: 59123828-59123828 on Build GRCh38
rs891303077 Chr.11: 59123948-59123948 on Build GRCh38
rs1008259111 Chr.11: 59123950-59123950 on Build GRCh38
rs540652183 Chr.11: 59124278-59124278 on Build GRCh38
rs780434423 Chr.11: 59124186-59124186 on Build GRCh38
rs765718160 Chr.11: 59124279-59124279 on Build GRCh38
rs371217106 Chr.11: 59124020-59124023 on Build GRCh38
rs934299979 Chr.11: 59124190-59124190 on Build GRCh38
rs77354659 Chr.11: 59124018-59124021 on Build GRCh38
rs373557170 Chr.11: 59124285-59124285 on Build GRCh38
rs762634551 Chr.11: 59124133-59124133 on Build GRCh38
rs372755718 Chr.11: 59124170-59124170 on Build GRCh38
rs564481369 Chr.11: 59124219-59124219 on Build GRCh38
rs761718649 Chr.11: 59123923-59123923 on Build GRCh38
rs767546691 Chr.11: 59124020-59124020 on Build GRCh38
rs971257351 Chr.11: 59124039-59124039 on Build GRCh38
rs186601522 Chr.11: 59123989-59123989 on Build GRCh38
rs10896925 Chr.11: 59123819-59123819 on Build GRCh38
rs140302898 Chr.11: 59123951-59123951 on Build GRCh38
rs201747467 Chr.11: 59124185-59124185 on Build GRCh38
rs576080631 Chr.11: 59124001-59124001 on Build GRCh38
rs751851641 Chr.11: 59124290-59124290 on Build GRCh38
rs1016494812 Chr.11: 59124083-59124083 on Build GRCh38
rs146690106 Chr.11: 59123908-59123908 on Build GRCh38
rs916729631 Chr.11: 59124142-59124142 on Build GRCh38
rs981221524 Chr.11: 59123811-59123811 on Build GRCh38
rs963884963 Chr.11: 59124097-59124097 on Build GRCh38
rs997592513 Chr.11: 59124008-59124008 on Build GRCh38
rs549327780 Chr.11: 59124201-59124201 on Build GRCh38
rs1014250422 Chr.11: 59124275-59124275 on Build GRCh38
rs746249220 Chr.11: 59124166-59124166 on Build GRCh38
rs376281103 Chr.11: 59124187-59124187 on Build GRCh38
rs754403024 Chr.11: 59124177-59124177 on Build GRCh38
rs759010155 Chr.11: 59124256-59124256 on Build GRCh38
rs989807959 Chr.11: 59124137-59124137 on Build GRCh38
rs769659482 Chr.11: 59124209-59124209 on Build GRCh38

View Full Product Details