Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66848899-66849493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PC
Entrez Gene ID
5091
Gene Name
pyruvate carboxylase
Gene Aliases
PCB
UniGene
Hs.89890
Gene Chromosome Location
Chr.11: 66848522-66958418 on Build GRCh38
Transcripts
NM_000920.3 NM_001040716.1 NM_022172.2 XM_005274031.4 XM_005274032.4 XM_006718578.3 XM_006718579.3 XM_011545086.2 XM_011545087.2 XM_017017868.1 XM_017017869.1 XM_017017870.1 XM_017017871.1 XM_017017872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5592377 c.3478G>A p.V1160I Chr.11: 66848958-66848958 on Build GRCh38
7079766 c.3452C>T p.S1151L Chr.11: 66848984-66848984 on Build GRCh38
5643670 c.3336C>T p.I1112I Chr.11: 66849100-66849100 on Build GRCh38
4922022 c.3211G>T p.A1071S Chr.11: 66849307-66849307 on Build GRCh38
1356327 c.3444G>A p.V1148V Chr.11: 66848992-66848992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746822762 Chr.11: 66849176-66849176 on Build GRCh38
rs982225457 Chr.11: 66849216-66849216 on Build GRCh38
rs527559823 Chr.11: 66849319-66849319 on Build GRCh38
rs377466972 Chr.11: 66849202-66849202 on Build GRCh38
rs758911778 Chr.11: 66849115-66849115 on Build GRCh38
rs145167276 Chr.11: 66849052-66849052 on Build GRCh38
rs529411023 Chr.11: 66849157-66849157 on Build GRCh38
rs751432105 Chr.11: 66848961-66848961 on Build GRCh38
rs369100780 Chr.11: 66849198-66849198 on Build GRCh38
rs988531701 Chr.11: 66849042-66849042 on Build GRCh38
rs992252687 Chr.11: 66849088-66849088 on Build GRCh38
rs147594996 Chr.11: 66848939-66848939 on Build GRCh38
rs753201697 Chr.11: 66849058-66849058 on Build GRCh38
rs531640812 Chr.11: 66849284-66849284 on Build GRCh38
rs149054698 Chr.11: 66848963-66848963 on Build GRCh38
rs747611524 Chr.11: 66849378-66849378 on Build GRCh38
rs748523908 Chr.11: 66849008-66849008 on Build GRCh38
rs376631492 Chr.11: 66849238-66849238 on Build GRCh38
rs1032917874 Chr.11: 66849189-66849189 on Build GRCh38
rs756309608 Chr.11: 66849167-66849167 on Build GRCh38
rs779825176 Chr.11: 66849343-66849343 on Build GRCh38
rs761184685 Chr.11: 66849391-66849391 on Build GRCh38
rs149583113 Chr.11: 66849306-66849306 on Build GRCh38
rs764717082 Chr.11: 66849108-66849108 on Build GRCh38
rs200217380 Chr.11: 66849056-66849056 on Build GRCh38
rs780090371 Chr.11: 66849219-66849219 on Build GRCh38
rs922296605 Chr.11: 66849292-66849292 on Build GRCh38
rs760307612 Chr.11: 66849394-66849395 on Build GRCh38
rs769462039 Chr.11: 66849280-66849280 on Build GRCh38
rs756436290 Chr.11: 66849213-66849213 on Build GRCh38
rs748037775 Chr.11: 66849036-66849044 on Build GRCh38
rs777927070 Chr.11: 66849027-66849027 on Build GRCh38
rs998353945 Chr.11: 66849128-66849128 on Build GRCh38
rs147550265 Chr.11: 66849347-66849347 on Build GRCh38
rs754942279 Chr.11: 66849239-66849239 on Build GRCh38
rs771894796 Chr.11: 66849215-66849216 on Build GRCh38
rs143777875 Chr.11: 66848964-66848964 on Build GRCh38
rs745626388 Chr.11: 66849276-66849276 on Build GRCh38
rs745677173 Chr.11: 66849338-66849338 on Build GRCh38
rs367795372 Chr.11: 66849410-66849410 on Build GRCh38
rs940669509 Chr.11: 66849195-66849195 on Build GRCh38
rs755635285 Chr.11: 66848967-66848967 on Build GRCh38
rs776196411 Chr.11: 66849100-66849100 on Build GRCh38
rs770969239 Chr.11: 66849034-66849034 on Build GRCh38
rs889047866 Chr.11: 66849217-66849217 on Build GRCh38
rs781063439 Chr.11: 66849337-66849337 on Build GRCh38
rs781571109 Chr.11: 66849139-66849139 on Build GRCh38
rs374396467 Chr.11: 66849205-66849205 on Build GRCh38
rs774487294 Chr.11: 66849086-66849086 on Build GRCh38
rs763623526 Chr.11: 66848923-66848923 on Build GRCh38
rs762965114 Chr.11: 66848943-66848943 on Build GRCh38
rs556299918 Chr.11: 66849443-66849443 on Build GRCh38
rs764770524 Chr.11: 66849186-66849186 on Build GRCh38
rs757220525 Chr.11: 66849329-66849329 on Build GRCh38
rs1017530164 Chr.11: 66849428-66849428 on Build GRCh38
rs34726770 Chr.11: 66849350-66849350 on Build GRCh38
rs762322442 Chr.11: 66849199-66849199 on Build GRCh38
rs770705087 Chr.11: 66849177-66849177 on Build GRCh38
rs1033650744 Chr.11: 66849407-66849407 on Build GRCh38
rs747239033 Chr.11: 66849065-66849065 on Build GRCh38
rs747244374 Chr.11: 66849031-66849031 on Build GRCh38
rs777830995 Chr.11: 66849247-66849247 on Build GRCh38
rs199640363 Chr.11: 66848979-66848979 on Build GRCh38
rs750712278 Chr.11: 66849210-66849210 on Build GRCh38
rs762530027 Chr.11: 66849291-66849291 on Build GRCh38
rs377509358 Chr.11: 66849168-66849168 on Build GRCh38
rs776825183 Chr.11: 66849037-66849037 on Build GRCh38
rs374682909 Chr.11: 66849269-66849269 on Build GRCh38
rs962105375 Chr.11: 66849413-66849413 on Build GRCh38
rs761578681 Chr.11: 66849049-66849049 on Build GRCh38
rs982178539 Chr.11: 66849051-66849051 on Build GRCh38
rs45441301 Chr.11: 66849201-66849201 on Build GRCh38
rs148661980 Chr.11: 66849348-66849348 on Build GRCh38
rs933360657 Chr.11: 66849430-66849430 on Build GRCh38
rs148492494 Chr.11: 66849095-66849095 on Build GRCh38
rs569765772 Chr.11: 66849452-66849452 on Build GRCh38
rs769512871 Chr.11: 66849341-66849341 on Build GRCh38
rs553914941 Chr.11: 66849040-66849040 on Build GRCh38
rs576126989 Chr.11: 66848959-66848959 on Build GRCh38
rs947199729 Chr.11: 66849460-66849460 on Build GRCh38
rs765735026 Chr.11: 66849057-66849057 on Build GRCh38
rs984166572 Chr.11: 66849427-66849427 on Build GRCh38
rs909244617 Chr.11: 66849106-66849106 on Build GRCh38
rs757532320 Chr.11: 66849066-66849066 on Build GRCh38
rs774375052 Chr.11: 66849048-66849048 on Build GRCh38
rs368783983 Chr.11: 66849309-66849309 on Build GRCh38
rs759110508 Chr.11: 66849103-66849103 on Build GRCh38
rs960938078 Chr.11: 66849079-66849079 on Build GRCh38
rs138238125 Chr.11: 66849260-66849260 on Build GRCh38
rs745961257 Chr.11: 66849072-66849072 on Build GRCh38
rs116518022 Chr.11: 66849045-66849045 on Build GRCh38
rs772960728 Chr.11: 66849172-66849172 on Build GRCh38
rs751541124 Chr.11: 66849326-66849326 on Build GRCh38
rs753961343 Chr.11: 66849226-66849226 on Build GRCh38
rs372116771 Chr.11: 66849310-66849310 on Build GRCh38
rs773784306 Chr.11: 66849384-66849384 on Build GRCh38
rs536414447 Chr.11: 66849442-66849442 on Build GRCh38
rs933670600 Chr.11: 66849293-66849293 on Build GRCh38
rs759169537 Chr.11: 66849181-66849181 on Build GRCh38
rs34376611 Chr.11: 66849324-66849324 on Build GRCh38
rs200166901 Chr.11: 66849046-66849046 on Build GRCh38
rs781515936 Chr.11: 66849071-66849071 on Build GRCh38
rs568925019 Chr.11: 66848922-66848922 on Build GRCh38
rs141118314 Chr.11: 66849050-66849050 on Build GRCh38
rs201796252 Chr.11: 66849001-66849001 on Build GRCh38
rs984302850 Chr.11: 66849235-66849235 on Build GRCh38
rs753363996 Chr.11: 66849055-66849055 on Build GRCh38
rs776326974 Chr.11: 66849179-66849179 on Build GRCh38
rs766636507 Chr.11: 66849404-66849404 on Build GRCh38
rs778839142 Chr.11: 66849325-66849325 on Build GRCh38
rs751977282 Chr.11: 66849127-66849127 on Build GRCh38
rs772718644 Chr.11: 66849380-66849380 on Build GRCh38
rs774982916 Chr.11: 66849305-66849305 on Build GRCh38
rs190223302 Chr.11: 66848953-66848953 on Build GRCh38
rs867013283 Chr.11: 66849303-66849303 on Build GRCh38
rs113994148 Chr.11: 66849026-66849027 on Build GRCh38
rs202085529 Chr.11: 66849109-66849109 on Build GRCh38
rs769930671 Chr.11: 66849076-66849076 on Build GRCh38
rs910028001 Chr.11: 66849256-66849256 on Build GRCh38
rs199585461 Chr.11: 66849010-66849010 on Build GRCh38
rs973437119 Chr.11: 66849470-66849470 on Build GRCh38
rs562596956 Chr.11: 66849209-66849209 on Build GRCh38
rs769801484 Chr.11: 66849099-66849099 on Build GRCh38
rs1003532375 Chr.11: 66849382-66849382 on Build GRCh38
rs1054233534 Chr.11: 66849323-66849323 on Build GRCh38
rs772973661 Chr.11: 66849216-66849216 on Build GRCh38
rs369947750 Chr.11: 66849063-66849063 on Build GRCh38
rs571513074 Chr.11: 66848983-66848983 on Build GRCh38
rs764471951 Chr.11: 66849121-66849121 on Build GRCh38
rs141978731 Chr.11: 66849094-66849094 on Build GRCh38
rs768048602 Chr.11: 66849304-66849304 on Build GRCh38
rs959557439 Chr.11: 66849234-66849234 on Build GRCh38
rs773764263 Chr.11: 66849357-66849357 on Build GRCh38
rs757506866 Chr.11: 66849130-66849130 on Build GRCh38
rs747701928 Chr.11: 66849090-66849090 on Build GRCh38
rs138990904 Chr.11: 66849053-66849053 on Build GRCh38
rs759649369 Chr.11: 66849416-66849416 on Build GRCh38
rs182440197 Chr.11: 66849060-66849060 on Build GRCh38
rs373619068 Chr.11: 66849218-66849218 on Build GRCh38
rs373287823 Chr.11: 66848974-66848974 on Build GRCh38
rs746903912 Chr.11: 66849251-66849251 on Build GRCh38
rs772410868 Chr.11: 66849020-66849020 on Build GRCh38
rs144087445 Chr.11: 66849320-66849320 on Build GRCh38

View Full Product Details