Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23030282-23030638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM1A
Entrez Gene ID
23028
Gene Name
lysine demethylase 1A
Gene Aliases
AOF2, BHC110, CPRF, KDM1, LSD1
UniGene
Hs.591518
Gene Chromosome Location
Chr.1: 23019443-23083691 on Build GRCh38
Transcripts
NM_001009999.2 NM_015013.3 XM_005245786.2 XM_006710472.3 XM_006710473.2 XM_006710474.3 XM_017000716.1 XM_017000717.1 XM_017000718.1 XM_017000719.1 XM_017000720.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
75101 c.454C>A p.P152T Chr.1: 23030571-23030571 on Build GRCh38
3485391 c.456C>T p.P152P Chr.1: 23030573-23030573 on Build GRCh38
1999579 c.465C>T p.P155P Chr.1: 23030582-23030582 on Build GRCh38
5927159 c.412G>A p.E138K Chr.1: 23030529-23030529 on Build GRCh38
1999575 c.422G>T p.R141I Chr.1: 23030539-23030539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006361029 Chr.1: 23030414-23030414 on Build GRCh38
rs572830037 Chr.1: 23030324-23030324 on Build GRCh38
rs981661791 Chr.1: 23030406-23030406 on Build GRCh38
rs971208893 Chr.1: 23030401-23030401 on Build GRCh38
rs938179141 Chr.1: 23030351-23030351 on Build GRCh38
rs1023145516 Chr.1: 23030394-23030394 on Build GRCh38
rs201780098 Chr.1: 23030537-23030537 on Build GRCh38
rs267598419 Chr.1: 23030596-23030596 on Build GRCh38
rs991200319 Chr.1: 23030388-23030388 on Build GRCh38
rs1001146530 Chr.1: 23030373-23030373 on Build GRCh38
rs1032652355 Chr.1: 23030379-23030379 on Build GRCh38
rs779826840 Chr.1: 23030573-23030573 on Build GRCh38
rs908397426 Chr.1: 23030465-23030465 on Build GRCh38
rs771350880 Chr.1: 23030489-23030489 on Build GRCh38
rs1021163055 Chr.1: 23030349-23030349 on Build GRCh38
rs769395218 Chr.1: 23030436-23030436 on Build GRCh38
rs1037439310 Chr.1: 23030459-23030459 on Build GRCh38
rs886205191 Chr.1: 23030412-23030412 on Build GRCh38
rs778266231 Chr.1: 23030461-23030461 on Build GRCh38
rs939909549 Chr.1: 23030557-23030557 on Build GRCh38
rs763306904 Chr.1: 23030616-23030617 on Build GRCh38
rs149557028 Chr.1: 23030576-23030576 on Build GRCh38
rs756077324 Chr.1: 23030437-23030437 on Build GRCh38
rs1008805478 Chr.1: 23030322-23030322 on Build GRCh38
rs749781563 Chr.1: 23030466-23030466 on Build GRCh38
rs746691186 Chr.1: 23030572-23030572 on Build GRCh38
rs184275508 Chr.1: 23030334-23030334 on Build GRCh38
rs189520263 Chr.1: 23030383-23030383 on Build GRCh38
rs370190039 Chr.1: 23030418-23030418 on Build GRCh38
rs10799788 Chr.1: 23030371-23030371 on Build GRCh38
rs112725574 Chr.1: 23030555-23030555 on Build GRCh38
rs201256608 Chr.1: 23030599-23030599 on Build GRCh38
rs774665623 Chr.1: 23030565-23030565 on Build GRCh38
rs748286629 Chr.1: 23030427-23030427 on Build GRCh38

View Full Product Details