Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167765447-167765975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPZL1
Entrez Gene ID
9019
Gene Name
myelin protein zero like 1
Gene Aliases
MPZL1b, PZR, PZR1b, PZRa, PZRb
UniGene
Hs.493919
Gene Chromosome Location
Chr.1: 167721950-167791919 on Build GRCh38
Transcripts
NM_001146191.1 NM_003953.5 NM_024569.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6714954 c.98C>T p.A33V Chr.1: 167765589-167765589 on Build GRCh38
5629016 c.122A>G p.Y41C Chr.1: 167765613-167765613 on Build GRCh38
463347 c.149A>C p.N50T Chr.1: 167765640-167765640 on Build GRCh38
3399960 c.256T>G p.S86A Chr.1: 167765747-167765747 on Build GRCh38
4629824 c.170C>T p.T57I Chr.1: 167765661-167765661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987584516 Chr.1: 167765941-167765941 on Build GRCh38
rs759638083 Chr.1: 167765928-167765928 on Build GRCh38
rs748892467 Chr.1: 167765531-167765531 on Build GRCh38
rs756537406 Chr.1: 167765736-167765736 on Build GRCh38
rs766483516 Chr.1: 167765593-167765593 on Build GRCh38
rs553050336 Chr.1: 167765560-167765560 on Build GRCh38
rs778504573 Chr.1: 167765638-167765638 on Build GRCh38
rs987655089 Chr.1: 167765860-167765860 on Build GRCh38
rs761576390 Chr.1: 167765783-167765783 on Build GRCh38
rs958990127 Chr.1: 167765496-167765496 on Build GRCh38
rs757960465 Chr.1: 167765733-167765733 on Build GRCh38
rs769743210 Chr.1: 167765631-167765631 on Build GRCh38
rs777526297 Chr.1: 167765529-167765529 on Build GRCh38
rs760728324 Chr.1: 167765762-167765762 on Build GRCh38
rs747579438 Chr.1: 167765539-167765539 on Build GRCh38
rs761725756 Chr.1: 167765603-167765603 on Build GRCh38
rs947415302 Chr.1: 167765514-167765514 on Build GRCh38
rs752650513 Chr.1: 167765616-167765616 on Build GRCh38
rs546058324 Chr.1: 167765819-167765819 on Build GRCh38
rs746616122 Chr.1: 167765525-167765525 on Build GRCh38
rs764835005 Chr.1: 167765723-167765723 on Build GRCh38
rs775647178 Chr.1: 167765759-167765759 on Build GRCh38
rs763283 Chr.1: 167765556-167765556 on Build GRCh38
rs769099868 Chr.1: 167765547-167765547 on Build GRCh38
rs770207236 Chr.1: 167765672-167765672 on Build GRCh38
rs996176179 Chr.1: 167765908-167765908 on Build GRCh38
rs143448673 Chr.1: 167765704-167765704 on Build GRCh38
rs746079380 Chr.1: 167765749-167765749 on Build GRCh38
rs369822857 Chr.1: 167765627-167765627 on Build GRCh38
rs777096702 Chr.1: 167765549-167765549 on Build GRCh38
rs761643652 Chr.1: 167765715-167765715 on Build GRCh38
rs778230265 Chr.1: 167765534-167765534 on Build GRCh38
rs954877127 Chr.1: 167765647-167765647 on Build GRCh38
rs566886951 Chr.1: 167765511-167765511 on Build GRCh38
rs749557484 Chr.1: 167765678-167765678 on Build GRCh38
rs760634409 Chr.1: 167765696-167765696 on Build GRCh38
rs771296800 Chr.1: 167765740-167765740 on Build GRCh38
rs752492891 Chr.1: 167765524-167765524 on Build GRCh38
rs773540156 Chr.1: 167765557-167765557 on Build GRCh38
rs576857327 Chr.1: 167765869-167765869 on Build GRCh38
rs771012047 Chr.1: 167765681-167765681 on Build GRCh38
rs372927221 Chr.1: 167765756-167765756 on Build GRCh38
rs771191552 Chr.1: 167765632-167765632 on Build GRCh38
rs755951123 Chr.1: 167765526-167765526 on Build GRCh38
rs779345094 Chr.1: 167765748-167765748 on Build GRCh38
rs774611276 Chr.1: 167765685-167765685 on Build GRCh38
rs910796069 Chr.1: 167765927-167765927 on Build GRCh38
rs139013682 Chr.1: 167765779-167765779 on Build GRCh38
rs146968580 Chr.1: 167765693-167765693 on Build GRCh38
rs760617962 Chr.1: 167765617-167765617 on Build GRCh38
rs188483765 Chr.1: 167765821-167765821 on Build GRCh38
rs200435022 Chr.1: 167765608-167765608 on Build GRCh38
rs750067905 Chr.1: 167765731-167765731 on Build GRCh38
rs749981310 Chr.1: 167765649-167765649 on Build GRCh38
rs765067263 Chr.1: 167765789-167765789 on Build GRCh38
rs753205839 Chr.1: 167765735-167765735 on Build GRCh38
rs763026238 Chr.1: 167765587-167765587 on Build GRCh38
rs373159576 Chr.1: 167765533-167765533 on Build GRCh38
rs889366851 Chr.1: 167765548-167765548 on Build GRCh38
rs150711804 Chr.1: 167765686-167765686 on Build GRCh38
rs1003422423 Chr.1: 167765518-167765518 on Build GRCh38
rs779870075 Chr.1: 167765517-167765517 on Build GRCh38
rs1036052706 Chr.1: 167765692-167765692 on Build GRCh38
rs576042164 Chr.1: 167765814-167765814 on Build GRCh38
rs757868400 Chr.1: 167765657-167765657 on Build GRCh38
rs886963894 Chr.1: 167765651-167765651 on Build GRCh38
rs375713174 Chr.1: 167765661-167765661 on Build GRCh38
rs779480588 Chr.1: 167765734-167765734 on Build GRCh38
rs777931189 Chr.1: 167765677-167765677 on Build GRCh38
rs748640490 Chr.1: 167765671-167765671 on Build GRCh38
rs991500476 Chr.1: 167765501-167765501 on Build GRCh38
rs867392999 Chr.1: 167765817-167765817 on Build GRCh38
rs201705663 Chr.1: 167765707-167765707 on Build GRCh38
rs767613384 Chr.1: 167765615-167765615 on Build GRCh38
rs1015528661 Chr.1: 167765951-167765951 on Build GRCh38
rs369996563 Chr.1: 167765739-167765739 on Build GRCh38
rs778224369 Chr.1: 167765738-167765738 on Build GRCh38
rs768522360 Chr.1: 167765777-167765777 on Build GRCh38
rs3830183 Chr.1: 167765480-167765480 on Build GRCh38

View Full Product Details