Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32702919-32703517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD7
Entrez Gene ID
149951
Gene Name
COMM domain containing 7
Gene Aliases
C20orf92, dJ1085F17.3
UniGene
Hs.408427
Gene Chromosome Location
Chr.20: 32702691-32744008 on Build GRCh38
Transcripts
NM_001099339.1 NM_053041.2 XM_011528604.2 XM_017027685.1 XM_017027686.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4817744 c.541C>T p.Q181* Chr.20: 32703444-32703444 on Build GRCh38
723111 c.558G>T p.L186L Chr.20: 32703427-32703427 on Build GRCh38
2889440 c.600C>T p.C200C Chr.20: 32703385-32703385 on Build GRCh38
6898416 c.570G>A p.E190E Chr.20: 32703415-32703415 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765015528 Chr.20: 32703140-32703140 on Build GRCh38
rs113674525 Chr.20: 32703136-32703136 on Build GRCh38
rs562333995 Chr.20: 32703287-32703287 on Build GRCh38
rs778127454 Chr.20: 32702973-32702973 on Build GRCh38
rs930742342 Chr.20: 32703186-32703186 on Build GRCh38
rs888301615 Chr.20: 32703214-32703214 on Build GRCh38
rs561589624 Chr.20: 32703064-32703064 on Build GRCh38
rs753939296 Chr.20: 32703331-32703331 on Build GRCh38
rs376710761 Chr.20: 32703447-32703447 on Build GRCh38
rs774347495 Chr.20: 32703206-32703207 on Build GRCh38
rs116401459 Chr.20: 32703114-32703114 on Build GRCh38
rs1013872053 Chr.20: 32703255-32703255 on Build GRCh38
rs773690268 Chr.20: 32703430-32703430 on Build GRCh38
rs562334971 Chr.20: 32703280-32703280 on Build GRCh38
rs1040756760 Chr.20: 32703213-32703213 on Build GRCh38
rs902967381 Chr.20: 32703188-32703188 on Build GRCh38
rs909300735 Chr.20: 32703048-32703048 on Build GRCh38
rs771241924 Chr.20: 32703459-32703459 on Build GRCh38
rs558018851 Chr.20: 32703003-32703003 on Build GRCh38
rs867527103 Chr.20: 32703363-32703363 on Build GRCh38
rs928696024 Chr.20: 32703288-32703288 on Build GRCh38
rs150687049 Chr.20: 32703060-32703060 on Build GRCh38
rs895770277 Chr.20: 32703250-32703250 on Build GRCh38
rs748683170 Chr.20: 32703423-32703423 on Build GRCh38
rs529307753 Chr.20: 32703068-32703068 on Build GRCh38
rs562515802 Chr.20: 32703162-32703162 on Build GRCh38
rs141965065 Chr.20: 32703015-32703015 on Build GRCh38
rs577463814 Chr.20: 32703338-32703338 on Build GRCh38
rs868466873 Chr.20: 32703261-32703261 on Build GRCh38
rs777017964 Chr.20: 32703465-32703465 on Build GRCh38
rs551512854 Chr.20: 32703215-32703215 on Build GRCh38
rs777629607 Chr.20: 32703027-32703027 on Build GRCh38
rs976313505 Chr.20: 32702946-32702946 on Build GRCh38
rs1033211636 Chr.20: 32702976-32702976 on Build GRCh38
rs770306742 Chr.20: 32703099-32703099 on Build GRCh38
rs1033571421 Chr.20: 32703333-32703333 on Build GRCh38
rs1006428666 Chr.20: 32703260-32703260 on Build GRCh38
rs200667336 Chr.20: 32703424-32703424 on Build GRCh38
rs960041877 Chr.20: 32703305-32703305 on Build GRCh38
rs376839090 Chr.20: 32703356-32703356 on Build GRCh38
rs28536125 Chr.20: 32703229-32703229 on Build GRCh38
rs533378764 Chr.20: 32703178-32703178 on Build GRCh38
rs145381646 Chr.20: 32702982-32702982 on Build GRCh38
rs752698328 Chr.20: 32703352-32703352 on Build GRCh38
rs999961824 Chr.20: 32703282-32703282 on Build GRCh38
rs373091485 Chr.20: 32703419-32703419 on Build GRCh38
rs867234984 Chr.20: 32703318-32703318 on Build GRCh38
rs758065631 Chr.20: 32703353-32703353 on Build GRCh38
rs1055653563 Chr.20: 32703118-32703118 on Build GRCh38
rs953585452 Chr.20: 32703004-32703004 on Build GRCh38
rs928623944 Chr.20: 32703096-32703096 on Build GRCh38
rs28525659 Chr.20: 32703150-32703150 on Build GRCh38
rs73906017 Chr.20: 32703177-32703177 on Build GRCh38
rs9421 Chr.20: 32703359-32703359 on Build GRCh38
rs745332295 Chr.20: 32703396-32703396 on Build GRCh38
rs974727126 Chr.20: 32703417-32703417 on Build GRCh38
rs776226219 Chr.20: 32703191-32703191 on Build GRCh38
rs1022611173 Chr.20: 32703266-32703266 on Build GRCh38
rs984894746 Chr.20: 32703443-32703443 on Build GRCh38
rs866555603 Chr.20: 32703240-32703240 on Build GRCh38
rs900857745 Chr.20: 32703216-32703216 on Build GRCh38
rs535513070 Chr.20: 32703414-32703414 on Build GRCh38
rs759326261 Chr.20: 32703465-32703465 on Build GRCh38
rs915797944 Chr.20: 32703135-32703136 on Build GRCh38
rs28522650 Chr.20: 32703020-32703020 on Build GRCh38
rs199516403 Chr.20: 32703354-32703354 on Build GRCh38
rs747170475 Chr.20: 32703190-32703190 on Build GRCh38
rs759480070 Chr.20: 32703490-32703490 on Build GRCh38
rs1029224570 Chr.20: 32702971-32702971 on Build GRCh38
rs755120859 Chr.20: 32703336-32703336 on Build GRCh38
rs780851032 Chr.20: 32703394-32703394 on Build GRCh38

View Full Product Details