Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49246030-49246578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNFX1
Entrez Gene ID
57169
Gene Name
zinc finger NFX1-type containing 1
Gene Aliases
-
UniGene
Hs.371794
Gene Chromosome Location
Chr.20: 49245897-49278244 on Build GRCh38
Transcripts
NM_021035.2 XM_006723836.1 XM_011528924.1 XM_017027971.1

SNP Detail

SNP ID Chromosome Location
rs767768046 Chr.20: 49246527-49246527 on Build GRCh38
rs1002603119 Chr.20: 49246478-49246478 on Build GRCh38
rs1061510 Chr.20: 49246102-49246102 on Build GRCh38
rs926574471 Chr.20: 49246260-49246260 on Build GRCh38
rs1022730118 Chr.20: 49246128-49246128 on Build GRCh38
rs1025925663 Chr.20: 49246479-49246479 on Build GRCh38
rs568051266 Chr.20: 49246178-49246178 on Build GRCh38
rs879234150 Chr.20: 49246423-49246429 on Build GRCh38
rs941075882 Chr.20: 49246343-49246343 on Build GRCh38
rs374627941 Chr.20: 49246086-49246086 on Build GRCh38
rs985039062 Chr.20: 49246207-49246207 on Build GRCh38
rs1054391823 Chr.20: 49246396-49246396 on Build GRCh38
rs141361012 Chr.20: 49246312-49246312 on Build GRCh38
rs377350435 Chr.20: 49246171-49246171 on Build GRCh38
rs184323944 Chr.20: 49246512-49246512 on Build GRCh38
rs533906386 Chr.20: 49246379-49246379 on Build GRCh38
rs751935900 Chr.20: 49246320-49246320 on Build GRCh38
rs191774143 Chr.20: 49246496-49246496 on Build GRCh38
rs551361081 Chr.20: 49246307-49246307 on Build GRCh38
rs553626899 Chr.20: 49246398-49246398 on Build GRCh38
rs531404216 Chr.20: 49246151-49246151 on Build GRCh38
rs988362559 Chr.20: 49246181-49246182 on Build GRCh38
rs980852148 Chr.20: 49246220-49246220 on Build GRCh38
rs939361722 Chr.20: 49246273-49246273 on Build GRCh38
rs964926247 Chr.20: 49246276-49246276 on Build GRCh38
rs6019709 Chr.20: 49246250-49246250 on Build GRCh38
rs998811413 Chr.20: 49246056-49246056 on Build GRCh38
rs774140833 Chr.20: 49246452-49246452 on Build GRCh38
rs906952634 Chr.20: 49246475-49246475 on Build GRCh38
rs533581766 Chr.20: 49246326-49246326 on Build GRCh38
rs967879332 Chr.20: 49246440-49246440 on Build GRCh38
rs1026377544 Chr.20: 49246109-49246117 on Build GRCh38
rs11478905 Chr.20: 49246380-49246380 on Build GRCh38
rs559031223 Chr.20: 49246439-49246439 on Build GRCh38
rs971428825 Chr.20: 49246135-49246135 on Build GRCh38
rs951032824 Chr.20: 49246538-49246538 on Build GRCh38
rs923389192 Chr.20: 49246291-49246291 on Build GRCh38
rs1039436575 Chr.20: 49246350-49246350 on Build GRCh38
rs985012228 Chr.20: 49246546-49246546 on Build GRCh38
rs975193599 Chr.20: 49246287-49246287 on Build GRCh38
rs1002258483 Chr.20: 49246444-49246444 on Build GRCh38
rs535880557 Chr.20: 49246377-49246377 on Build GRCh38
rs566841240 Chr.20: 49246328-49246328 on Build GRCh38
rs142547545 Chr.20: 49246510-49246510 on Build GRCh38
rs892998756 Chr.20: 49246118-49246118 on Build GRCh38
rs933427483 Chr.20: 49246302-49246302 on Build GRCh38
rs1033240138 Chr.20: 49246084-49246084 on Build GRCh38
rs759828802 Chr.20: 49246366-49246366 on Build GRCh38
rs748963721 Chr.20: 49246410-49246410 on Build GRCh38
rs879590212 Chr.20: 49246224-49246224 on Build GRCh38
rs1006860769 Chr.20: 49246505-49246505 on Build GRCh38
rs772896605 Chr.20: 49246083-49246083 on Build GRCh38
rs566448892 Chr.20: 49246321-49246321 on Build GRCh38
rs892934833 Chr.20: 49246399-49246399 on Build GRCh38
rs903092647 Chr.20: 49246053-49246053 on Build GRCh38
rs1013050466 Chr.20: 49246122-49246122 on Build GRCh38
rs1012753835 Chr.20: 49246436-49246436 on Build GRCh38
rs139708662 Chr.20: 49246198-49246198 on Build GRCh38
rs548319485 Chr.20: 49246212-49246212 on Build GRCh38
rs770676311 Chr.20: 49246415-49246415 on Build GRCh38
rs529145425 Chr.20: 49246188-49246188 on Build GRCh38
rs539559462 Chr.20: 49246181-49246181 on Build GRCh38
rs575239674 Chr.20: 49246555-49246555 on Build GRCh38
rs892366031 Chr.20: 49246409-49246409 on Build GRCh38
rs934550518 Chr.20: 49246387-49246387 on Build GRCh38
rs899472256 Chr.20: 49246354-49246354 on Build GRCh38

View Full Product Details