Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36744868-36745466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIM2
Entrez Gene ID
6493
Gene Name
single-minded family bHLH transcription factor 2
Gene Aliases
HMC13F06, HMC29C01, SIM, bHLHe15
UniGene
Hs.146186
Gene Chromosome Location
Chr.21: 36699117-36750219 on Build GRCh38
Transcripts
NM_005069.4 NM_009586.3 XM_011529694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1713974 c.1348G>A p.D450N Chr.21: 36744908-36744908 on Build GRCh38
7389932 c.1315C>T p.P439S Chr.21: 36744875-36744875 on Build GRCh38
315191 c.1406A>C p.Q469P Chr.21: 36744966-36744966 on Build GRCh38
4134921 c.1447C>A p.L483M Chr.21: 36745007-36745007 on Build GRCh38
4134922 c.1447C>A p.L483M Chr.21: 36745007-36745007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990981292 Chr.21: 36745344-36745344 on Build GRCh38
rs764720358 Chr.21: 36745199-36745199 on Build GRCh38
rs754671706 Chr.21: 36745353-36745353 on Build GRCh38
rs562948609 Chr.21: 36745126-36745126 on Build GRCh38
rs769858094 Chr.21: 36744948-36744948 on Build GRCh38
rs867741391 Chr.21: 36745160-36745160 on Build GRCh38
rs759483083 Chr.21: 36744990-36744990 on Build GRCh38
rs1048360864 Chr.21: 36744899-36744899 on Build GRCh38
rs777505968 Chr.21: 36744918-36744918 on Build GRCh38
rs148338000 Chr.21: 36745096-36745096 on Build GRCh38
rs554903042 Chr.21: 36745092-36745092 on Build GRCh38
rs747573399 Chr.21: 36744940-36744940 on Build GRCh38
rs888376496 Chr.21: 36745375-36745375 on Build GRCh38
rs144394618 Chr.21: 36745095-36745095 on Build GRCh38
rs779257260 Chr.21: 36745205-36745205 on Build GRCh38
rs879426414 Chr.21: 36744966-36744966 on Build GRCh38
rs141533294 Chr.21: 36745133-36745133 on Build GRCh38
rs754216148 Chr.21: 36745201-36745201 on Build GRCh38
rs764825979 Chr.21: 36745183-36745183 on Build GRCh38
rs756590082 Chr.21: 36745193-36745193 on Build GRCh38
rs771187646 Chr.21: 36744985-36744989 on Build GRCh38
rs753307817 Chr.21: 36744927-36744927 on Build GRCh38
rs192771022 Chr.21: 36745191-36745191 on Build GRCh38
rs1026818032 Chr.21: 36745314-36745314 on Build GRCh38
rs571774763 Chr.21: 36745062-36745062 on Build GRCh38
rs778635337 Chr.21: 36745128-36745128 on Build GRCh38
rs535112068 Chr.21: 36745132-36745132 on Build GRCh38
rs751598467 Chr.21: 36744893-36744893 on Build GRCh38
rs906896297 Chr.21: 36745171-36745171 on Build GRCh38
rs747223348 Chr.21: 36744981-36744981 on Build GRCh38
rs200755165 Chr.21: 36745173-36745173 on Build GRCh38
rs753537362 Chr.21: 36745010-36745010 on Build GRCh38
rs201708628 Chr.21: 36745019-36745019 on Build GRCh38
rs201951528 Chr.21: 36745018-36745018 on Build GRCh38
rs915653688 Chr.21: 36745127-36745127 on Build GRCh38
rs201126287 Chr.21: 36745136-36745136 on Build GRCh38
rs773103692 Chr.21: 36744890-36744890 on Build GRCh38
rs749688040 Chr.21: 36745150-36745150 on Build GRCh38
rs371125576 Chr.21: 36745190-36745190 on Build GRCh38
rs776261523 Chr.21: 36745097-36745097 on Build GRCh38
rs781731790 Chr.21: 36745030-36745030 on Build GRCh38
rs756553530 Chr.21: 36745045-36745045 on Build GRCh38
rs61750768 Chr.21: 36744946-36744946 on Build GRCh38
rs768585007 Chr.21: 36745176-36745176 on Build GRCh38
rs773596315 Chr.21: 36744955-36744955 on Build GRCh38
rs893338712 Chr.21: 36745000-36745000 on Build GRCh38
rs201454422 Chr.21: 36745184-36745184 on Build GRCh38
rs780515558 Chr.21: 36744939-36744939 on Build GRCh38
rs1045455758 Chr.21: 36744989-36744989 on Build GRCh38
rs200359415 Chr.21: 36744956-36744956 on Build GRCh38
rs768140573 Chr.21: 36745099-36745099 on Build GRCh38
rs752542906 Chr.21: 36744913-36744913 on Build GRCh38
rs769059784 Chr.21: 36744942-36744942 on Build GRCh38
rs374048214 Chr.21: 36745083-36745083 on Build GRCh38
rs758403084 Chr.21: 36745210-36745210 on Build GRCh38
rs368057275 Chr.21: 36744992-36744992 on Build GRCh38
rs748418447 Chr.21: 36745035-36745035 on Build GRCh38
rs373865332 Chr.21: 36745271-36745271 on Build GRCh38
rs750713368 Chr.21: 36745208-36745208 on Build GRCh38
rs769624132 Chr.21: 36745248-36745248 on Build GRCh38
rs768527730 Chr.21: 36745229-36745229 on Build GRCh38
rs1023486131 Chr.21: 36745438-36745438 on Build GRCh38
rs747032214 Chr.21: 36745175-36745175 on Build GRCh38
rs1030737267 Chr.21: 36745251-36745251 on Build GRCh38
rs957197618 Chr.21: 36745025-36745025 on Build GRCh38
rs11910583 Chr.21: 36745281-36745281 on Build GRCh38
rs563000221 Chr.21: 36744976-36744976 on Build GRCh38
rs201246885 Chr.21: 36745113-36745113 on Build GRCh38
rs367607207 Chr.21: 36745091-36745091 on Build GRCh38
rs149650483 Chr.21: 36745066-36745066 on Build GRCh38
rs565363510 Chr.21: 36745367-36745367 on Build GRCh38
rs76502794 Chr.21: 36745263-36745263 on Build GRCh38
rs757506420 Chr.21: 36745153-36745153 on Build GRCh38
rs140786374 Chr.21: 36744916-36744916 on Build GRCh38
rs527848385 Chr.21: 36745399-36745399 on Build GRCh38
rs377180868 Chr.21: 36745116-36745116 on Build GRCh38
rs376845812 Chr.21: 36745235-36745235 on Build GRCh38
rs948687041 Chr.21: 36744920-36744920 on Build GRCh38
rs776705793 Chr.21: 36745336-36745336 on Build GRCh38
rs747073193 Chr.21: 36745223-36745223 on Build GRCh38
rs761387486 Chr.21: 36745098-36745098 on Build GRCh38
rs772245049 Chr.21: 36745172-36745172 on Build GRCh38
rs916773476 Chr.21: 36745398-36745398 on Build GRCh38
rs1008276838 Chr.21: 36745067-36745067 on Build GRCh38
rs2073603 Chr.21: 36745441-36745441 on Build GRCh38
rs545439054 Chr.21: 36745307-36745307 on Build GRCh38
rs2073602 Chr.21: 36745204-36745204 on Build GRCh38
rs779310089 Chr.21: 36745156-36745156 on Build GRCh38
rs751039393 Chr.21: 36745192-36745192 on Build GRCh38
rs981471638 Chr.21: 36744922-36744922 on Build GRCh38
rs1017605435 Chr.21: 36745058-36745058 on Build GRCh38
rs756799246 Chr.21: 36744937-36744937 on Build GRCh38
rs772900536 Chr.21: 36745437-36745437 on Build GRCh38
rs781341610 Chr.21: 36744945-36744945 on Build GRCh38
rs976047465 Chr.21: 36745065-36745065 on Build GRCh38
rs759396897 Chr.21: 36744897-36744897 on Build GRCh38
rs776352057 Chr.21: 36745179-36745179 on Build GRCh38
rs1045405870 Chr.21: 36745161-36745161 on Build GRCh38
rs948437965 Chr.21: 36745410-36745410 on Build GRCh38
rs778130813 Chr.21: 36745060-36745060 on Build GRCh38
rs372565191 Chr.21: 36745180-36745180 on Build GRCh38
rs199788000 Chr.21: 36745118-36745118 on Build GRCh38
rs755379484 Chr.21: 36745022-36745022 on Build GRCh38
rs767876751 Chr.21: 36745121-36745121 on Build GRCh38
rs200609904 Chr.21: 36745181-36745181 on Build GRCh38
rs774516027 Chr.21: 36745080-36745080 on Build GRCh38
rs963998955 Chr.21: 36745313-36745313 on Build GRCh38
rs548039650 Chr.21: 36745104-36745104 on Build GRCh38
rs750413729 Chr.21: 36745185-36745185 on Build GRCh38
rs2073601 Chr.21: 36745007-36745007 on Build GRCh38
rs767285576 Chr.21: 36744898-36744898 on Build GRCh38
rs1012551396 Chr.21: 36744984-36744984 on Build GRCh38
rs756744287 Chr.21: 36745015-36745015 on Build GRCh38
rs746926522 Chr.21: 36745005-36745005 on Build GRCh38
rs774430209 Chr.21: 36744987-36744987 on Build GRCh38
rs575040244 Chr.21: 36745168-36745168 on Build GRCh38

View Full Product Details