Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37862971-37863509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3L
Entrez Gene ID
51386
Gene Name
eukaryotic translation initiation factor 3 subunit L
Gene Aliases
EIF3EIP, EIF3S11, EIF3S6IP, HSPC021, HSPC025, MSTP005
UniGene
Hs.446852
Gene Chromosome Location
Chr.22: 37848774-37888782 on Build GRCh38
Transcripts
NM_001242923.1 NM_016091.3 XM_005261625.1 XM_006724260.3 XM_017028814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680626 c.510C>T p.A170A Chr.22: 37863276-37863276 on Build GRCh38
6482801 c.581A>G p.E194G Chr.22: 37862985-37862985 on Build GRCh38
3842658 c.621C>T p.F207F Chr.22: 37863025-37863025 on Build GRCh38
6564671 c.592G>C p.E198Q Chr.22: 37862996-37862996 on Build GRCh38
6569380 c.658G>C p.E220Q Chr.22: 37863295-37863295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751540875 Chr.22: 37863308-37863308 on Build GRCh38
rs768219088 Chr.22: 37863286-37863286 on Build GRCh38
rs1007931138 Chr.22: 37863143-37863143 on Build GRCh38
rs927916088 Chr.22: 37863393-37863393 on Build GRCh38
rs919746060 Chr.22: 37863403-37863404 on Build GRCh38
rs779086975 Chr.22: 37863071-37863071 on Build GRCh38
rs76886280 Chr.22: 37863105-37863105 on Build GRCh38
rs781320528 Chr.22: 37863354-37863359 on Build GRCh38
rs765750566 Chr.22: 37863022-37863022 on Build GRCh38
rs763334822 Chr.22: 37863038-37863041 on Build GRCh38
rs935992358 Chr.22: 37863276-37863276 on Build GRCh38
rs1026026418 Chr.22: 37863150-37863150 on Build GRCh38
rs987631857 Chr.22: 37863209-37863209 on Build GRCh38
rs769429605 Chr.22: 37863490-37863490 on Build GRCh38
rs780827504 Chr.22: 37863244-37863244 on Build GRCh38
rs754708343 Chr.22: 37863312-37863312 on Build GRCh38
rs758385893 Chr.22: 37863224-37863224 on Build GRCh38
rs769328106 Chr.22: 37863255-37863255 on Build GRCh38
rs766805331 Chr.22: 37863034-37863034 on Build GRCh38
rs761129339 Chr.22: 37863030-37863030 on Build GRCh38
rs113036185 Chr.22: 37863346-37863346 on Build GRCh38
rs760158217 Chr.22: 37863017-37863017 on Build GRCh38
rs994480001 Chr.22: 37863468-37863468 on Build GRCh38
rs529613643 Chr.22: 37863104-37863104 on Build GRCh38
rs114098260 Chr.22: 37863474-37863474 on Build GRCh38
rs372316591 Chr.22: 37863354-37863354 on Build GRCh38
rs950957124 Chr.22: 37863198-37863198 on Build GRCh38
rs781109901 Chr.22: 37863390-37863390 on Build GRCh38
rs375153596 Chr.22: 37863309-37863309 on Build GRCh38
rs1011715384 Chr.22: 37863175-37863176 on Build GRCh38
rs750906417 Chr.22: 37863027-37863027 on Build GRCh38
rs199855201 Chr.22: 37863122-37863122 on Build GRCh38
rs746880813 Chr.22: 37863235-37863235 on Build GRCh38
rs769396962 Chr.22: 37863010-37863010 on Build GRCh38
rs956326935 Chr.22: 37863153-37863153 on Build GRCh38
rs898830400 Chr.22: 37863455-37863455 on Build GRCh38
rs964940448 Chr.22: 37863215-37863215 on Build GRCh38
rs752904926 Chr.22: 37863379-37863379 on Build GRCh38
rs762452441 Chr.22: 37863016-37863016 on Build GRCh38
rs767027126 Chr.22: 37863212-37863212 on Build GRCh38
rs764222492 Chr.22: 37863055-37863056 on Build GRCh38
rs777789278 Chr.22: 37863382-37863382 on Build GRCh38
rs376912297 Chr.22: 37863355-37863355 on Build GRCh38
rs745576237 Chr.22: 37863249-37863249 on Build GRCh38
rs977783274 Chr.22: 37863148-37863148 on Build GRCh38
rs751742556 Chr.22: 37863058-37863075 on Build GRCh38
rs774832603 Chr.22: 37863012-37863012 on Build GRCh38
rs1019247254 Chr.22: 37863214-37863214 on Build GRCh38
rs577341407 Chr.22: 37863064-37863064 on Build GRCh38
rs980171268 Chr.22: 37863256-37863256 on Build GRCh38
rs754119383 Chr.22: 37863352-37863352 on Build GRCh38
rs563224261 Chr.22: 37863454-37863454 on Build GRCh38
rs369186163 Chr.22: 37863384-37863384 on Build GRCh38
rs890767124 Chr.22: 37863127-37863129 on Build GRCh38
rs762385956 Chr.22: 37863282-37863282 on Build GRCh38
rs945235168 Chr.22: 37863339-37863339 on Build GRCh38
rs960311047 Chr.22: 37863318-37863318 on Build GRCh38
rs768127139 Chr.22: 37863019-37863019 on Build GRCh38
rs757460917 Chr.22: 37863076-37863078 on Build GRCh38
rs766895925 Chr.22: 37863336-37863336 on Build GRCh38
rs754108739 Chr.22: 37863059-37863059 on Build GRCh38
rs544821416 Chr.22: 37863152-37863152 on Build GRCh38
rs752714112 Chr.22: 37863085-37863085 on Build GRCh38
rs151317788 Chr.22: 37863327-37863327 on Build GRCh38
rs372075058 Chr.22: 37863236-37863236 on Build GRCh38
rs375241677 Chr.22: 37863483-37863483 on Build GRCh38
rs937945532 Chr.22: 37863405-37863405 on Build GRCh38
rs75149111 Chr.22: 37863121-37863121 on Build GRCh38
rs777701016 Chr.22: 37863230-37863230 on Build GRCh38
rs779963254 Chr.22: 37863277-37863277 on Build GRCh38
rs879115165 Chr.22: 37863104-37863105 on Build GRCh38
rs61195610 Chr.22: 37863121-37863121 on Build GRCh38
rs925955795 Chr.22: 37863268-37863268 on Build GRCh38
rs111677695 Chr.22: 37863271-37863271 on Build GRCh38
rs1046210904 Chr.22: 37863367-37863367 on Build GRCh38
rs11551381 Chr.22: 37863013-37863013 on Build GRCh38
rs989303246 Chr.22: 37863322-37863322 on Build GRCh38
rs753022886 Chr.22: 37863025-37863050 on Build GRCh38
rs62235090 Chr.22: 37863124-37863124 on Build GRCh38
rs538057738 Chr.22: 37863410-37863410 on Build GRCh38
rs765363056 Chr.22: 37863359-37863359 on Build GRCh38

View Full Product Details