Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48451501-48452051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRIP
Entrez Gene ID
84126
Gene Name
ATR interacting protein
Gene Aliases
-
UniGene
Hs.694840
Gene Chromosome Location
Chr.3: 48446710-48465655 on Build GRCh38
Transcripts
NM_001271022.1 NM_001271023.1 NM_032166.3 NM_130384.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
215912 c.437G>T p.R146L Chr.3: 48451784-48451784 on Build GRCh38
4980815 c.439G>C p.D147H Chr.3: 48451786-48451786 on Build GRCh38
1045817 c.406C>A p.L136I Chr.3: 48451753-48451753 on Build GRCh38
3824061 c.482G>A p.R161K Chr.3: 48451829-48451829 on Build GRCh38
4618342 c.436C>T p.R146* Chr.3: 48451783-48451783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774024081 Chr.3: 48451875-48451875 on Build GRCh38
rs763650403 Chr.3: 48451900-48451900 on Build GRCh38
rs754422416 Chr.3: 48451665-48451669 on Build GRCh38
rs758201067 Chr.3: 48451868-48451868 on Build GRCh38
rs760072859 Chr.3: 48451899-48451899 on Build GRCh38
rs748150512 Chr.3: 48451813-48451813 on Build GRCh38
rs532010755 Chr.3: 48451661-48451661 on Build GRCh38
rs367749611 Chr.3: 48451927-48451927 on Build GRCh38
rs773462495 Chr.3: 48452024-48452024 on Build GRCh38
rs368316197 Chr.3: 48451865-48451865 on Build GRCh38
rs541108999 Chr.3: 48451677-48451677 on Build GRCh38
rs755417423 Chr.3: 48451940-48451940 on Build GRCh38
rs1015222652 Chr.3: 48451524-48451524 on Build GRCh38
rs370971747 Chr.3: 48451810-48451810 on Build GRCh38
rs141432992 Chr.3: 48451869-48451869 on Build GRCh38
rs929802073 Chr.3: 48451870-48451870 on Build GRCh38
rs994740894 Chr.3: 48451814-48451814 on Build GRCh38
rs769860249 Chr.3: 48451957-48451957 on Build GRCh38
rs999735327 Chr.3: 48451557-48451557 on Build GRCh38
rs964019742 Chr.3: 48451787-48451787 on Build GRCh38
rs371062542 Chr.3: 48451691-48451691 on Build GRCh38
rs34958611 Chr.3: 48451891-48451892 on Build GRCh38
rs373841067 Chr.3: 48451922-48451922 on Build GRCh38
rs756299316 Chr.3: 48451838-48451838 on Build GRCh38
rs938421044 Chr.3: 48451997-48451997 on Build GRCh38
rs763150971 Chr.3: 48452030-48452030 on Build GRCh38
rs542982776 Chr.3: 48451578-48451578 on Build GRCh38
rs150966664 Chr.3: 48451803-48451803 on Build GRCh38
rs781135932 Chr.3: 48451726-48451726 on Build GRCh38
rs894108063 Chr.3: 48452025-48452025 on Build GRCh38
rs749455816 Chr.3: 48451867-48451867 on Build GRCh38
rs903984374 Chr.3: 48451540-48451540 on Build GRCh38
rs970848154 Chr.3: 48451996-48451996 on Build GRCh38
rs766688944 Chr.3: 48451924-48451924 on Build GRCh38
rs949543114 Chr.3: 48451660-48451660 on Build GRCh38
rs374427312 Chr.3: 48451859-48451859 on Build GRCh38
rs370107308 Chr.3: 48451782-48451782 on Build GRCh38
rs375297834 Chr.3: 48451695-48451695 on Build GRCh38
rs745780294 Chr.3: 48451879-48451879 on Build GRCh38
rs753529542 Chr.3: 48451798-48451798 on Build GRCh38
rs762807851 Chr.3: 48451745-48451745 on Build GRCh38
rs928355289 Chr.3: 48451990-48451990 on Build GRCh38
rs982663940 Chr.3: 48451971-48451971 on Build GRCh38
rs750808938 Chr.3: 48451697-48451697 on Build GRCh38
rs150047731 Chr.3: 48451862-48451862 on Build GRCh38
rs752036149 Chr.3: 48451809-48451809 on Build GRCh38
rs371819088 Chr.3: 48451643-48451643 on Build GRCh38
rs773557915 Chr.3: 48451941-48451941 on Build GRCh38
rs371656741 Chr.3: 48451921-48451921 on Build GRCh38
rs564526533 Chr.3: 48451629-48451629 on Build GRCh38
rs893512708 Chr.3: 48451717-48451717 on Build GRCh38
rs752378897 Chr.3: 48451659-48451659 on Build GRCh38
rs759408256 Chr.3: 48451763-48451763 on Build GRCh38
rs758812183 Chr.3: 48451699-48451699 on Build GRCh38
rs569929723 Chr.3: 48451891-48451891 on Build GRCh38
rs377643484 Chr.3: 48451707-48451707 on Build GRCh38
rs758748581 Chr.3: 48451802-48451802 on Build GRCh38
rs760327399 Chr.3: 48451784-48451784 on Build GRCh38
rs771927804 Chr.3: 48451886-48451886 on Build GRCh38
rs530809060 Chr.3: 48452007-48452007 on Build GRCh38
rs576351838 Chr.3: 48451534-48451534 on Build GRCh38
rs112322775 Chr.3: 48451844-48451844 on Build GRCh38
rs773612911 Chr.3: 48451744-48451744 on Build GRCh38
rs771233287 Chr.3: 48451894-48451894 on Build GRCh38
rs770160806 Chr.3: 48451730-48451730 on Build GRCh38
rs990898468 Chr.3: 48451635-48451635 on Build GRCh38
rs780534119 Chr.3: 48451706-48451706 on Build GRCh38
rs373507607 Chr.3: 48451751-48451751 on Build GRCh38
rs962000560 Chr.3: 48451532-48451532 on Build GRCh38
rs766988675 Chr.3: 48451780-48451780 on Build GRCh38
rs369352614 Chr.3: 48451729-48451729 on Build GRCh38
rs770921027 Chr.3: 48451748-48451748 on Build GRCh38
rs763713809 Chr.3: 48451785-48451785 on Build GRCh38
rs781110153 Chr.3: 48451741-48451743 on Build GRCh38
rs767876526 Chr.3: 48451942-48451942 on Build GRCh38
rs376983909 Chr.3: 48451770-48451770 on Build GRCh38
rs919950726 Chr.3: 48451625-48451626 on Build GRCh38

View Full Product Details