Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44658683-44659207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF35
Entrez Gene ID
7584
Gene Name
zinc finger protein 35
Gene Aliases
HF.10, HF10, Zfp105
UniGene
Hs.288658
Gene Chromosome Location
Chr.3: 44648723-44660791 on Build GRCh38
Transcripts
NM_003420.3 XM_017007129.1 XM_017007130.1 XM_017007131.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7239007 c.724C>A p.R242R Chr.3: 44659087-44659087 on Build GRCh38
1537762 c.442C>T p.Q148* Chr.3: 44658805-44658805 on Build GRCh38
5680768 c.767G>T p.G256V Chr.3: 44659130-44659130 on Build GRCh38
730802 c.624C>T p.G208G Chr.3: 44658987-44658987 on Build GRCh38
5864556 c.745_746CC>TT p.P249F Chr.3: 44659108-44659109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753272048 Chr.3: 44658987-44658987 on Build GRCh38
rs753250758 Chr.3: 44658953-44658953 on Build GRCh38
rs755055432 Chr.3: 44658911-44658914 on Build GRCh38
rs756851996 Chr.3: 44658879-44658879 on Build GRCh38
rs763065217 Chr.3: 44658785-44658785 on Build GRCh38
rs763723480 Chr.3: 44658975-44658975 on Build GRCh38
rs775825402 Chr.3: 44659086-44659086 on Build GRCh38
rs201658931 Chr.3: 44659122-44659122 on Build GRCh38
rs754744154 Chr.3: 44659158-44659158 on Build GRCh38
rs764795631 Chr.3: 44659114-44659114 on Build GRCh38
rs773507592 Chr.3: 44658780-44658780 on Build GRCh38
rs370522018 Chr.3: 44658794-44658794 on Build GRCh38
rs759924090 Chr.3: 44658945-44658945 on Build GRCh38
rs771002534 Chr.3: 44659074-44659074 on Build GRCh38
rs778185468 Chr.3: 44659025-44659026 on Build GRCh38
rs780746773 Chr.3: 44658720-44658720 on Build GRCh38
rs567126364 Chr.3: 44659053-44659053 on Build GRCh38
rs778438749 Chr.3: 44658989-44658989 on Build GRCh38
rs761992185 Chr.3: 44658776-44658776 on Build GRCh38
rs530319721 Chr.3: 44658891-44658891 on Build GRCh38
rs752931213 Chr.3: 44659020-44659021 on Build GRCh38
rs750064286 Chr.3: 44659031-44659031 on Build GRCh38
rs774412028 Chr.3: 44659087-44659087 on Build GRCh38
rs773136348 Chr.3: 44658927-44658927 on Build GRCh38
rs879012440 Chr.3: 44659107-44659107 on Build GRCh38
rs749556029 Chr.3: 44659075-44659077 on Build GRCh38
rs765212781 Chr.3: 44658770-44658772 on Build GRCh38
rs757329009 Chr.3: 44659182-44659182 on Build GRCh38
rs781021867 Chr.3: 44659159-44659159 on Build GRCh38
rs761265622 Chr.3: 44659108-44659108 on Build GRCh38
rs140633689 Chr.3: 44659069-44659069 on Build GRCh38
rs370667591 Chr.3: 44658842-44658842 on Build GRCh38
rs746985754 Chr.3: 44659039-44659039 on Build GRCh38
rs767819466 Chr.3: 44658949-44658949 on Build GRCh38
rs745666779 Chr.3: 44658897-44658897 on Build GRCh38
rs777967101 Chr.3: 44659059-44659059 on Build GRCh38
rs374168391 Chr.3: 44658968-44658968 on Build GRCh38
rs760093722 Chr.3: 44658819-44658819 on Build GRCh38
rs1038848207 Chr.3: 44658868-44658868 on Build GRCh38
rs771930825 Chr.3: 44658899-44658899 on Build GRCh38
rs777511950 Chr.3: 44658750-44658750 on Build GRCh38
rs138010130 Chr.3: 44659032-44659032 on Build GRCh38
rs550173382 Chr.3: 44659038-44659038 on Build GRCh38
rs768016217 Chr.3: 44658831-44658831 on Build GRCh38
rs780175079 Chr.3: 44658906-44658906 on Build GRCh38
rs777123629 Chr.3: 44658867-44658867 on Build GRCh38
rs748069595 Chr.3: 44659169-44659169 on Build GRCh38
rs761299958 Chr.3: 44659088-44659088 on Build GRCh38
rs764518906 Chr.3: 44659091-44659091 on Build GRCh38
rs751917700 Chr.3: 44658818-44658818 on Build GRCh38
rs867135640 Chr.3: 44659001-44659001 on Build GRCh38
rs753013640 Chr.3: 44659030-44659030 on Build GRCh38
rs947355975 Chr.3: 44658749-44658749 on Build GRCh38
rs775514273 Chr.3: 44658775-44658775 on Build GRCh38
rs770806231 Chr.3: 44658765-44658765 on Build GRCh38
rs901672719 Chr.3: 44658871-44658871 on Build GRCh38
rs988933456 Chr.3: 44659144-44659144 on Build GRCh38
rs144064090 Chr.3: 44659142-44659142 on Build GRCh38
rs919210096 Chr.3: 44659130-44659130 on Build GRCh38
rs772464398 Chr.3: 44659079-44659079 on Build GRCh38
rs745864639 Chr.3: 44658772-44658772 on Build GRCh38
rs758095422 Chr.3: 44659118-44659118 on Build GRCh38
rs751299039 Chr.3: 44659149-44659149 on Build GRCh38
rs750152264 Chr.3: 44659116-44659116 on Build GRCh38
rs764596073 Chr.3: 44659121-44659121 on Build GRCh38
rs777124890 Chr.3: 44659098-44659098 on Build GRCh38
rs34183917 Chr.3: 44659158-44659159 on Build GRCh38
rs753564283 Chr.3: 44658876-44658876 on Build GRCh38
rs747671498 Chr.3: 44658743-44658743 on Build GRCh38
rs146760313 Chr.3: 44658928-44658928 on Build GRCh38
rs756500722 Chr.3: 44659023-44659024 on Build GRCh38
rs139593995 Chr.3: 44658778-44658778 on Build GRCh38
rs139090092 Chr.3: 44658855-44658855 on Build GRCh38
rs970300878 Chr.3: 44658972-44658972 on Build GRCh38
rs767898030 Chr.3: 44659017-44659018 on Build GRCh38
rs879195141 Chr.3: 44659134-44659134 on Build GRCh38
rs755554359 Chr.3: 44658748-44658748 on Build GRCh38
rs367753047 Chr.3: 44658988-44658988 on Build GRCh38
rs746896269 Chr.3: 44658910-44658910 on Build GRCh38
rs532535433 Chr.3: 44658766-44658766 on Build GRCh38
rs769968112 Chr.3: 44658925-44658925 on Build GRCh38
rs748841468 Chr.3: 44658760-44658760 on Build GRCh38
rs140378573 Chr.3: 44658941-44658941 on Build GRCh38
rs772117188 Chr.3: 44658774-44658774 on Build GRCh38
rs751716671 Chr.3: 44658784-44658785 on Build GRCh38
rs138185290 Chr.3: 44659075-44659075 on Build GRCh38
rs3732518 Chr.3: 44658889-44658889 on Build GRCh38
rs746066752 Chr.3: 44659076-44659076 on Build GRCh38

View Full Product Details