Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 71516895-71517186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYSF
Entrez Gene ID
8291
Gene Name
dysferlin
Gene Aliases
FER1L1, LGMD2B, MMD1
UniGene
Hs.252180
Gene Chromosome Location
Chr.2: 71453155-71686763 on Build GRCh38
Transcripts
NM_001130455.1 NM_001130976.1 NM_001130977.1 NM_001130978.1 NM_001130979.1 NM_001130980.1 NM_001130981.1 NM_001130982.1 NM_001130983.1 NM_001130984.1 NM_001130985.1 NM_001130986.1 NM_001130987.1 NM_003494.3 XM_005264584.4 XM_005264585.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1022662 c.878G>A p.R293K Chr.2: 71517011-71517011 on Build GRCh38
1752646 c.856-1G>A p.? Chr.2: 71516988-71516988 on Build GRCh38
1592397 c.974G>A p.R325K Chr.2: 71517011-71517011 on Build GRCh38
4515115 c.999C>T p.F333F Chr.2: 71517036-71517036 on Build GRCh38
443171 c.875T>C p.L292P Chr.2: 71517008-71517008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771325384 Chr.2: 71516997-71516997 on Build GRCh38
rs753987509 Chr.2: 71516991-71516991 on Build GRCh38
rs112286757 Chr.2: 71517057-71517057 on Build GRCh38
rs747009076 Chr.2: 71517084-71517084 on Build GRCh38
rs771357626 Chr.2: 71517083-71517083 on Build GRCh38
rs756335217 Chr.2: 71516967-71516967 on Build GRCh38
rs759748502 Chr.2: 71517003-71517003 on Build GRCh38
rs121908963 Chr.2: 71517028-71517028 on Build GRCh38
rs564949517 Chr.2: 71516935-71516935 on Build GRCh38
rs747513056 Chr.2: 71516995-71516995 on Build GRCh38
rs944678848 Chr.2: 71516946-71516946 on Build GRCh38
rs367878277 Chr.2: 71517036-71517036 on Build GRCh38
rs773742883 Chr.2: 71517103-71517103 on Build GRCh38
rs868254368 Chr.2: 71517124-71517124 on Build GRCh38
rs398123808 Chr.2: 71516998-71516998 on Build GRCh38
rs11688242 Chr.2: 71517128-71517128 on Build GRCh38
rs140809078 Chr.2: 71517027-71517027 on Build GRCh38
rs111279933 Chr.2: 71516919-71516919 on Build GRCh38
rs753577400 Chr.2: 71517047-71517047 on Build GRCh38
rs776921154 Chr.2: 71516999-71516999 on Build GRCh38
rs886044386 Chr.2: 71517032-71517032 on Build GRCh38
rs746263554 Chr.2: 71517086-71517086 on Build GRCh38
rs778496697 Chr.2: 71516972-71516972 on Build GRCh38
rs868080649 Chr.2: 71517073-71517073 on Build GRCh38
rs905322985 Chr.2: 71517043-71517043 on Build GRCh38
rs112007817 Chr.2: 71517015-71517015 on Build GRCh38
rs184771575 Chr.2: 71517037-71517037 on Build GRCh38
rs760585799 Chr.2: 71517038-71517038 on Build GRCh38
rs555599178 Chr.2: 71517165-71517165 on Build GRCh38
rs548634243 Chr.2: 71517116-71517116 on Build GRCh38
rs747688530 Chr.2: 71516975-71516975 on Build GRCh38
rs754688393 Chr.2: 71516949-71516949 on Build GRCh38
rs766137790 Chr.2: 71517039-71517039 on Build GRCh38
rs375755423 Chr.2: 71517077-71517077 on Build GRCh38
rs772653237 Chr.2: 71516983-71516983 on Build GRCh38
rs777889370 Chr.2: 71517089-71517089 on Build GRCh38
rs529992457 Chr.2: 71517054-71517054 on Build GRCh38
rs398123807 Chr.2: 71516990-71516990 on Build GRCh38
rs530278138 Chr.2: 71516944-71516945 on Build GRCh38
rs752450312 Chr.2: 71517061-71517061 on Build GRCh38
rs757954559 Chr.2: 71517067-71517067 on Build GRCh38
rs747540377 Chr.2: 71517080-71517080 on Build GRCh38

View Full Product Details