Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202036006-202036509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD7
Entrez Gene ID
8324
Gene Name
frizzled class receptor 7
Gene Aliases
FzE3
UniGene
Hs.173859
Gene Chromosome Location
Chr.2: 202034587-202038437 on Build GRCh38
Transcripts
NM_003507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668179 c.1452C>T p.I484I Chr.2: 202036099-202036099 on Build GRCh38
6668180 c.1537G>A p.V513M Chr.2: 202036184-202036184 on Build GRCh38
6668178 c.1409G>A p.R470H Chr.2: 202036056-202036056 on Build GRCh38
7143432 c.1605G>A p.L535L Chr.2: 202036252-202036252 on Build GRCh38
5037268 c.1510C>A p.L504I Chr.2: 202036157-202036157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000638888 Chr.2: 202036030-202036030 on Build GRCh38
rs182592123 Chr.2: 202036195-202036195 on Build GRCh38
rs946938708 Chr.2: 202036205-202036205 on Build GRCh38
rs891140187 Chr.2: 202036209-202036209 on Build GRCh38
rs1038885248 Chr.2: 202036090-202036090 on Build GRCh38
rs758853443 Chr.2: 202036276-202036276 on Build GRCh38
rs769424376 Chr.2: 202036288-202036288 on Build GRCh38
rs201452450 Chr.2: 202036118-202036118 on Build GRCh38
rs763782149 Chr.2: 202036214-202036214 on Build GRCh38
rs748151902 Chr.2: 202036403-202036403 on Build GRCh38
rs746743333 Chr.2: 202036087-202036087 on Build GRCh38
rs754937674 Chr.2: 202036416-202036418 on Build GRCh38
rs932729482 Chr.2: 202036478-202036478 on Build GRCh38
rs200370632 Chr.2: 202036186-202036186 on Build GRCh38
rs753302518 Chr.2: 202036366-202036366 on Build GRCh38
rs761575550 Chr.2: 202036322-202036322 on Build GRCh38
rs761598219 Chr.2: 202036231-202036231 on Build GRCh38
rs771841493 Chr.2: 202036404-202036404 on Build GRCh38
rs546449437 Chr.2: 202036377-202036377 on Build GRCh38
rs534588552 Chr.2: 202036407-202036407 on Build GRCh38
rs755768740 Chr.2: 202036380-202036380 on Build GRCh38
rs143528185 Chr.2: 202036183-202036183 on Build GRCh38
rs767737659 Chr.2: 202036162-202036162 on Build GRCh38
rs772709570 Chr.2: 202036070-202036070 on Build GRCh38
rs769473710 Chr.2: 202036203-202036203 on Build GRCh38
rs541873049 Chr.2: 202036182-202036182 on Build GRCh38
rs756155956 Chr.2: 202036255-202036255 on Build GRCh38
rs900336952 Chr.2: 202036468-202036468 on Build GRCh38
rs751432827 Chr.2: 202036178-202036179 on Build GRCh38
rs780572956 Chr.2: 202036029-202036029 on Build GRCh38
rs35600847 Chr.2: 202036107-202036107 on Build GRCh38
rs199589707 Chr.2: 202036389-202036389 on Build GRCh38
rs754291190 Chr.2: 202036265-202036265 on Build GRCh38
rs1033879510 Chr.2: 202036374-202036374 on Build GRCh38
rs773419766 Chr.2: 202036336-202036336 on Build GRCh38
rs371345546 Chr.2: 202036382-202036382 on Build GRCh38
rs372525407 Chr.2: 202036174-202036174 on Build GRCh38
rs774179533 Chr.2: 202036303-202036303 on Build GRCh38
rs745320558 Chr.2: 202036198-202036198 on Build GRCh38
rs755458386 Chr.2: 202036267-202036267 on Build GRCh38
rs140867221 Chr.2: 202036238-202036238 on Build GRCh38
rs772230105 Chr.2: 202036052-202036052 on Build GRCh38
rs766517331 Chr.2: 202036260-202036260 on Build GRCh38
rs765225022 Chr.2: 202036117-202036117 on Build GRCh38
rs750694306 Chr.2: 202036246-202036246 on Build GRCh38
rs562818320 Chr.2: 202036150-202036150 on Build GRCh38
rs759120037 Chr.2: 202036103-202036103 on Build GRCh38
rs763013166 Chr.2: 202036207-202036207 on Build GRCh38
rs111263028 Chr.2: 202036273-202036273 on Build GRCh38
rs768569299 Chr.2: 202036296-202036296 on Build GRCh38
rs201191053 Chr.2: 202036064-202036064 on Build GRCh38
rs977753392 Chr.2: 202036419-202036419 on Build GRCh38
rs762632983 Chr.2: 202036128-202036128 on Build GRCh38
rs955763827 Chr.2: 202036456-202036456 on Build GRCh38
rs765723806 Chr.2: 202036360-202036360 on Build GRCh38
rs974234940 Chr.2: 202036367-202036367 on Build GRCh38
rs547551444 Chr.2: 202036428-202036428 on Build GRCh38
rs1038809392 Chr.2: 202036452-202036452 on Build GRCh38
rs759636568 Chr.2: 202036416-202036416 on Build GRCh38
rs536312561 Chr.2: 202036483-202036483 on Build GRCh38
rs775589753 Chr.2: 202036126-202036126 on Build GRCh38
rs763985883 Chr.2: 202036129-202036129 on Build GRCh38
rs528874438 Chr.2: 202036281-202036281 on Build GRCh38
rs759567940 Chr.2: 202036354-202036354 on Build GRCh38
rs925202604 Chr.2: 202036424-202036424 on Build GRCh38
rs772937162 Chr.2: 202036406-202036406 on Build GRCh38
rs774136031 Chr.2: 202036223-202036223 on Build GRCh38
rs745552067 Chr.2: 202036282-202036282 on Build GRCh38
rs989868994 Chr.2: 202036479-202036479 on Build GRCh38
rs748503156 Chr.2: 202036275-202036275 on Build GRCh38
rs748759824 Chr.2: 202036290-202036290 on Build GRCh38
rs749570044 Chr.2: 202036031-202036031 on Build GRCh38
rs368139563 Chr.2: 202036263-202036263 on Build GRCh38
rs750086736 Chr.2: 202036378-202036378 on Build GRCh38
rs770553279 Chr.2: 202036097-202036097 on Build GRCh38
rs372045596 Chr.2: 202036289-202036289 on Build GRCh38
rs997348548 Chr.2: 202036141-202036141 on Build GRCh38
rs1015839962 Chr.2: 202036337-202036337 on Build GRCh38
rs1029625624 Chr.2: 202036172-202036172 on Build GRCh38
rs771894898 Chr.2: 202036332-202036332 on Build GRCh38
rs565714072 Chr.2: 202036340-202036340 on Build GRCh38
rs775255613 Chr.2: 202036204-202036204 on Build GRCh38
rs368422200 Chr.2: 202036159-202036159 on Build GRCh38
rs766526311 Chr.2: 202036342-202036342 on Build GRCh38
rs112052844 Chr.2: 202036422-202036422 on Build GRCh38
rs779576113 Chr.2: 202036384-202036384 on Build GRCh38
rs770643171 Chr.2: 202036192-202036192 on Build GRCh38
rs367875554 Chr.2: 202036410-202036410 on Build GRCh38
rs767678664 Chr.2: 202036237-202036237 on Build GRCh38
rs776225705 Chr.2: 202036099-202036099 on Build GRCh38
rs750464553 Chr.2: 202036168-202036168 on Build GRCh38
rs373957083 Chr.2: 202036386-202036386 on Build GRCh38
rs771147451 Chr.2: 202036408-202036408 on Build GRCh38
rs758800464 Chr.2: 202036375-202036375 on Build GRCh38
rs1004437982 Chr.2: 202036222-202036222 on Build GRCh38

View Full Product Details