Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60435886-60436469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf39
Entrez Gene ID
317761
Gene Name
chromosome 14 open reading frame 39
Gene Aliases
Six6os1
UniGene
Hs.335754
Gene Chromosome Location
Chr.14: 60435954-60499773 on Build GRCh38
Transcripts
NM_174978.2 XM_011536690.2 XM_017021247.1 XM_017021248.1 XM_017021249.1 XM_017021250.1 XM_017021251.1

SNP Detail

SNP ID Chromosome Location
rs186370361 Chr.14: 60436045-60436045 on Build GRCh38
rs567430809 Chr.14: 60436216-60436216 on Build GRCh38
rs765269446 Chr.14: 60436306-60436306 on Build GRCh38
rs777955685 Chr.14: 60436400-60436400 on Build GRCh38
rs547297963 Chr.14: 60436411-60436411 on Build GRCh38
rs1012557700 Chr.14: 60436313-60436313 on Build GRCh38
rs571302950 Chr.14: 60436007-60436007 on Build GRCh38
rs1005870284 Chr.14: 60436370-60436370 on Build GRCh38
rs991974526 Chr.14: 60435968-60435968 on Build GRCh38
rs925089408 Chr.14: 60436338-60436340 on Build GRCh38
rs902963496 Chr.14: 60435956-60435956 on Build GRCh38
rs868712112 Chr.14: 60435962-60435962 on Build GRCh38
rs1013251915 Chr.14: 60436036-60436036 on Build GRCh38
rs566897766 Chr.14: 60436425-60436425 on Build GRCh38
rs903354555 Chr.14: 60436253-60436253 on Build GRCh38
rs372745544 Chr.14: 60436316-60436316 on Build GRCh38
rs1053356451 Chr.14: 60436128-60436128 on Build GRCh38
rs865998575 Chr.14: 60436115-60436115 on Build GRCh38
rs991923467 Chr.14: 60436301-60436301 on Build GRCh38
rs551116429 Chr.14: 60436006-60436006 on Build GRCh38
rs888394728 Chr.14: 60436330-60436330 on Build GRCh38
rs140005817 Chr.14: 60436296-60436296 on Build GRCh38
rs923268437 Chr.14: 60436196-60436196 on Build GRCh38
rs1015419494 Chr.14: 60436100-60436100 on Build GRCh38
rs910780661 Chr.14: 60436224-60436224 on Build GRCh38
rs750883119 Chr.14: 60436081-60436084 on Build GRCh38
rs972629528 Chr.14: 60435916-60435916 on Build GRCh38
rs550848505 Chr.14: 60436276-60436276 on Build GRCh38
rs916406064 Chr.14: 60436310-60436310 on Build GRCh38
rs998547536 Chr.14: 60435958-60435958 on Build GRCh38
rs146552661 Chr.14: 60435923-60435923 on Build GRCh38
rs558300978 Chr.14: 60436129-60436129 on Build GRCh38
rs1011716494 Chr.14: 60435915-60435915 on Build GRCh38
rs994164194 Chr.14: 60436267-60436267 on Build GRCh38
rs191129808 Chr.14: 60436415-60436415 on Build GRCh38
rs889289399 Chr.14: 60435914-60435914 on Build GRCh38
rs781461225 Chr.14: 60435993-60435993 on Build GRCh38
rs547520496 Chr.14: 60436096-60436096 on Build GRCh38
rs750213302 Chr.14: 60436373-60436373 on Build GRCh38
rs998337154 Chr.14: 60436163-60436163 on Build GRCh38
rs181268399 Chr.14: 60435967-60435967 on Build GRCh38
rs753985895 Chr.14: 60436043-60436043 on Build GRCh38
rs368695767 Chr.14: 60436024-60436024 on Build GRCh38
rs1043361683 Chr.14: 60436025-60436025 on Build GRCh38
rs962150966 Chr.14: 60436104-60436104 on Build GRCh38
rs1022848011 Chr.14: 60436041-60436041 on Build GRCh38
rs933272152 Chr.14: 60436204-60436204 on Build GRCh38
rs1043258086 Chr.14: 60436240-60436240 on Build GRCh38
rs1016898822 Chr.14: 60436218-60436218 on Build GRCh38
rs947607247 Chr.14: 60436225-60436225 on Build GRCh38
rs1007394196 Chr.14: 60436217-60436217 on Build GRCh38
rs916307322 Chr.14: 60435999-60435999 on Build GRCh38
rs578182191 Chr.14: 60436132-60436132 on Build GRCh38
rs571053124 Chr.14: 60436279-60436279 on Build GRCh38
rs924942613 Chr.14: 60436069-60436069 on Build GRCh38
rs972147434 Chr.14: 60436417-60436417 on Build GRCh38
rs895484684 Chr.14: 60436021-60436021 on Build GRCh38

View Full Product Details