Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 65616117-65616640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT8
Entrez Gene ID
2530
Gene Name
fucosyltransferase 8
Gene Aliases
-
UniGene
Hs.597649
Gene Chromosome Location
Chr.14: 65410592-65744122 on Build GRCh38
Transcripts
NM_004480.4 NM_178155.2 NM_178156.2 NR_038167.1 NR_038170.1 XM_017021136.1 XM_017021137.1 XM_017021138.1 XM_017021139.1 XM_017021140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5954611 c.320G>T p.G107V Chr.14: 65616211-65616211 on Build GRCh38
5954612 c.320G>T p.G107V Chr.14: 65616211-65616211 on Build GRCh38
3497210 c.387C>T p.F129F Chr.14: 65616278-65616278 on Build GRCh38
6076241 c.360A>C p.E120D Chr.14: 65616251-65616251 on Build GRCh38
6076242 c.360A>C p.E120D Chr.14: 65616251-65616251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368517345 Chr.14: 65616287-65616287 on Build GRCh38
rs772490571 Chr.14: 65616319-65616319 on Build GRCh38
rs942800974 Chr.14: 65616186-65616186 on Build GRCh38
rs1025758897 Chr.14: 65616510-65616510 on Build GRCh38
rs776715455 Chr.14: 65616207-65616207 on Build GRCh38
rs534145588 Chr.14: 65616451-65616451 on Build GRCh38
rs151271028 Chr.14: 65616223-65616223 on Build GRCh38
rs573689097 Chr.14: 65616483-65616483 on Build GRCh38
rs990432949 Chr.14: 65616391-65616391 on Build GRCh38
rs771398830 Chr.14: 65616144-65616144 on Build GRCh38
rs1001581014 Chr.14: 65616491-65616491 on Build GRCh38
rs779443464 Chr.14: 65616309-65616309 on Build GRCh38
rs267604027 Chr.14: 65616221-65616221 on Build GRCh38
rs76077399 Chr.14: 65616150-65616150 on Build GRCh38
rs764202386 Chr.14: 65616540-65616540 on Build GRCh38
rs752266680 Chr.14: 65616418-65616418 on Build GRCh38
rs911210128 Chr.14: 65616143-65616143 on Build GRCh38
rs768675915 Chr.14: 65616196-65616196 on Build GRCh38
rs10483783 Chr.14: 65616485-65616485 on Build GRCh38
rs761905226 Chr.14: 65616367-65616367 on Build GRCh38
rs374236653 Chr.14: 65616211-65616211 on Build GRCh38
rs780567844 Chr.14: 65616281-65616281 on Build GRCh38
rs750268401 Chr.14: 65616231-65616231 on Build GRCh38
rs780382949 Chr.14: 65616336-65616336 on Build GRCh38
rs1008648557 Chr.14: 65616599-65616599 on Build GRCh38
rs952991108 Chr.14: 65616523-65616523 on Build GRCh38
rs776426178 Chr.14: 65616284-65616284 on Build GRCh38
rs1041152916 Chr.14: 65616192-65616192 on Build GRCh38
rs571334030 Chr.14: 65616425-65616425 on Build GRCh38
rs78083604 Chr.14: 65616348-65616348 on Build GRCh38
rs762982895 Chr.14: 65616382-65616382 on Build GRCh38
rs369578832 Chr.14: 65616392-65616392 on Build GRCh38
rs112292748 Chr.14: 65616206-65616206 on Build GRCh38
rs754649929 Chr.14: 65616278-65616278 on Build GRCh38
rs923692528 Chr.14: 65616480-65616480 on Build GRCh38
rs775727296 Chr.14: 65616193-65616193 on Build GRCh38
rs370523624 Chr.14: 65616170-65616170 on Build GRCh38
rs911440163 Chr.14: 65616593-65616593 on Build GRCh38
rs762893251 Chr.14: 65616241-65616241 on Build GRCh38
rs544763878 Chr.14: 65616601-65616601 on Build GRCh38
rs900456780 Chr.14: 65616169-65616169 on Build GRCh38
rs746341099 Chr.14: 65616317-65616317 on Build GRCh38
rs181322184 Chr.14: 65616359-65616359 on Build GRCh38
rs577389232 Chr.14: 65616437-65616437 on Build GRCh38
rs10483782 Chr.14: 65616452-65616452 on Build GRCh38
rs977828176 Chr.14: 65616456-65616456 on Build GRCh38
rs773324695 Chr.14: 65616377-65616377 on Build GRCh38
rs772401093 Chr.14: 65616181-65616181 on Build GRCh38
rs868263242 Chr.14: 65616542-65616542 on Build GRCh38
rs764789704 Chr.14: 65616410-65616410 on Build GRCh38
rs974219574 Chr.14: 65616302-65616302 on Build GRCh38
rs996849814 Chr.14: 65616175-65616175 on Build GRCh38
rs759138158 Chr.14: 65616408-65616408 on Build GRCh38
rs186342230 Chr.14: 65616499-65616499 on Build GRCh38
rs760763835 Chr.14: 65616498-65616498 on Build GRCh38
rs751327808 Chr.14: 65616269-65616269 on Build GRCh38
rs757724110 Chr.14: 65616308-65616308 on Build GRCh38
rs760882431 Chr.14: 65616194-65616194 on Build GRCh38
rs9323462 Chr.14: 65616496-65616496 on Build GRCh38
rs201242327 Chr.14: 65616337-65616337 on Build GRCh38
rs746179555 Chr.14: 65616178-65616178 on Build GRCh38
rs769741984 Chr.14: 65616370-65616370 on Build GRCh38
rs200973027 Chr.14: 65616261-65616261 on Build GRCh38
rs917562068 Chr.14: 65616444-65616444 on Build GRCh38

View Full Product Details