Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124811745-124812106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCARB1
Entrez Gene ID
949
Gene Name
scavenger receptor class B member 1
Gene Aliases
CD36L1, CLA-1, CLA1, HDLQTL6, SR-BI, SRB1
UniGene
Hs.731377
Gene Chromosome Location
Chr.12: 124777628-124863973 on Build GRCh38
Transcripts
NM_001082959.1 NM_005505.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4813580 c.652C>T p.L218F Chr.12: 124811944-124811944 on Build GRCh38
6053050 c.631-4G>C p.? Chr.12: 124811969-124811969 on Build GRCh38
4204635 c.604G>A p.G202R Chr.12: 124811881-124811881 on Build GRCh38
4816415 c.690G>A p.R230R Chr.12: 124811906-124811906 on Build GRCh38
4504042 c.654C>T p.L218L Chr.12: 124811942-124811942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369936442 Chr.12: 124811854-124811854 on Build GRCh38
rs113910315 Chr.12: 124811979-124811979 on Build GRCh38
rs750274986 Chr.12: 124812010-124812010 on Build GRCh38
rs749365287 Chr.12: 124811928-124811928 on Build GRCh38
rs372212527 Chr.12: 124811846-124811846 on Build GRCh38
rs755600850 Chr.12: 124812076-124812076 on Build GRCh38
rs144347463 Chr.12: 124811840-124811840 on Build GRCh38
rs975535442 Chr.12: 124812033-124812033 on Build GRCh38
rs754627228 Chr.12: 124811844-124811845 on Build GRCh38
rs934213440 Chr.12: 124811912-124811912 on Build GRCh38
rs767372321 Chr.12: 124812003-124812003 on Build GRCh38
rs764073908 Chr.12: 124811900-124811900 on Build GRCh38
rs1022003362 Chr.12: 124811801-124811801 on Build GRCh38
rs764600414 Chr.12: 124811888-124811888 on Build GRCh38
rs757116603 Chr.12: 124811909-124811909 on Build GRCh38
rs997641243 Chr.12: 124812068-124812068 on Build GRCh38
rs755909827 Chr.12: 124812017-124812017 on Build GRCh38
rs922736657 Chr.12: 124812039-124812039 on Build GRCh38
rs780989061 Chr.12: 124811953-124811953 on Build GRCh38
rs760824799 Chr.12: 124811848-124811848 on Build GRCh38
rs964504079 Chr.12: 124812032-124812032 on Build GRCh38
rs370285171 Chr.12: 124811948-124811948 on Build GRCh38
rs867946310 Chr.12: 124812004-124812004 on Build GRCh38
rs757891509 Chr.12: 124811897-124811897 on Build GRCh38
rs753545464 Chr.12: 124811824-124811824 on Build GRCh38
rs762513823 Chr.12: 124811987-124811987 on Build GRCh38
rs201556935 Chr.12: 124811853-124811853 on Build GRCh38
rs773585501 Chr.12: 124811958-124811958 on Build GRCh38
rs778486309 Chr.12: 124811839-124811839 on Build GRCh38
rs775653998 Chr.12: 124811857-124811857 on Build GRCh38
rs769174348 Chr.12: 124811861-124811861 on Build GRCh38
rs1034942905 Chr.12: 124811834-124811834 on Build GRCh38
rs754478086 Chr.12: 124811837-124811837 on Build GRCh38
rs10396213 Chr.12: 124811911-124811911 on Build GRCh38
rs372806890 Chr.12: 124811954-124811954 on Build GRCh38
rs768674012 Chr.12: 124811936-124811936 on Build GRCh38
rs1043474498 Chr.12: 124811939-124811939 on Build GRCh38
rs144358906 Chr.12: 124811896-124811896 on Build GRCh38
rs960091019 Chr.12: 124811965-124811965 on Build GRCh38
rs761913857 Chr.12: 124811780-124811780 on Build GRCh38
rs77740046 Chr.12: 124812018-124812018 on Build GRCh38
rs375927939 Chr.12: 124811985-124811985 on Build GRCh38
rs372960506 Chr.12: 124811860-124811860 on Build GRCh38
rs1010535276 Chr.12: 124811937-124811937 on Build GRCh38
rs889129693 Chr.12: 124812086-124812086 on Build GRCh38
rs759068321 Chr.12: 124811975-124811975 on Build GRCh38
rs35857542 Chr.12: 124811979-124811980 on Build GRCh38
rs746030637 Chr.12: 124811914-124811914 on Build GRCh38
rs948344876 Chr.12: 124811802-124811802 on Build GRCh38
rs770902587 Chr.12: 124811852-124811852 on Build GRCh38
rs59809936 Chr.12: 124811815-124811815 on Build GRCh38
rs181516117 Chr.12: 124811841-124811841 on Build GRCh38
rs762150484 Chr.12: 124811873-124811873 on Build GRCh38
rs755341504 Chr.12: 124811949-124811949 on Build GRCh38
rs751606686 Chr.12: 124811901-124811901 on Build GRCh38
rs767934483 Chr.12: 124811880-124811880 on Build GRCh38
rs371058347 Chr.12: 124811991-124811991 on Build GRCh38
rs532003403 Chr.12: 124812055-124812055 on Build GRCh38
rs1010585925 Chr.12: 124811789-124811789 on Build GRCh38
rs186213925 Chr.12: 124811882-124811882 on Build GRCh38
rs757276005 Chr.12: 124811998-124811998 on Build GRCh38
rs61932577 Chr.12: 124811816-124811816 on Build GRCh38
rs780178642 Chr.12: 124811927-124811927 on Build GRCh38
rs756381665 Chr.12: 124811919-124811919 on Build GRCh38
rs774768995 Chr.12: 124811862-124811862 on Build GRCh38
rs374322109 Chr.12: 124811992-124811992 on Build GRCh38
rs199957324 Chr.12: 124811823-124811823 on Build GRCh38
rs201977189 Chr.12: 124811881-124811881 on Build GRCh38

View Full Product Details