Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66810462-66810978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAE1
Entrez Gene ID
8883
Gene Name
NEDD8 activating enzyme E1 subunit 1
Gene Aliases
A-116A10.1, APPBP1, HPP1, ula-1
UniGene
Hs.460978
Gene Chromosome Location
Chr.16: 66802878-66830977 on Build GRCh38
Transcripts
NM_001018159.1 NM_001018160.1 NM_001286500.1 NM_003905.3 XM_005256215.1 XM_011523422.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1493713 c.1035-2A>T p.? Chr.16: 66810774-66810774 on Build GRCh38
6721690 c.1039C>T p.R347C Chr.16: 66810768-66810768 on Build GRCh38
7125954 c.1100C>T p.S367F Chr.16: 66810707-66810707 on Build GRCh38
3510826 c.1042G>A p.E348K Chr.16: 66810765-66810765 on Build GRCh38
2993775 c.1102A>G p.I368V Chr.16: 66810705-66810705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768532088 Chr.16: 66810543-66810544 on Build GRCh38
rs1044358720 Chr.16: 66810525-66810525 on Build GRCh38
rs996409569 Chr.16: 66810527-66810527 on Build GRCh38
rs765879962 Chr.16: 66810784-66810784 on Build GRCh38
rs964431279 Chr.16: 66810656-66810656 on Build GRCh38
rs530808605 Chr.16: 66810688-66810688 on Build GRCh38
rs754279171 Chr.16: 66810774-66810774 on Build GRCh38
rs74749616 Chr.16: 66810604-66810604 on Build GRCh38
rs539170856 Chr.16: 66810742-66810742 on Build GRCh38
rs986934942 Chr.16: 66810767-66810767 on Build GRCh38
rs779185551 Chr.16: 66810695-66810695 on Build GRCh38
rs754435976 Chr.16: 66810689-66810689 on Build GRCh38
rs566143134 Chr.16: 66810864-66810864 on Build GRCh38
rs372939687 Chr.16: 66810736-66810736 on Build GRCh38
rs767177659 Chr.16: 66810670-66810670 on Build GRCh38
rs951284811 Chr.16: 66810895-66810895 on Build GRCh38
rs955590375 Chr.16: 66810687-66810687 on Build GRCh38
rs982984723 Chr.16: 66810907-66810907 on Build GRCh38
rs778149341 Chr.16: 66810697-66810697 on Build GRCh38
rs766808551 Chr.16: 66810768-66810768 on Build GRCh38
rs750713493 Chr.16: 66810676-66810676 on Build GRCh38
rs762353169 Chr.16: 66810651-66810651 on Build GRCh38
rs1002714968 Chr.16: 66810597-66810599 on Build GRCh38
rs552605906 Chr.16: 66810831-66810831 on Build GRCh38
rs752760955 Chr.16: 66810801-66810801 on Build GRCh38
rs948733195 Chr.16: 66810889-66810889 on Build GRCh38
rs199760584 Chr.16: 66810668-66810668 on Build GRCh38
rs768130596 Chr.16: 66810738-66810738 on Build GRCh38
rs1021777454 Chr.16: 66810640-66810640 on Build GRCh38
rs760944372 Chr.16: 66810753-66810753 on Build GRCh38
rs1013551463 Chr.16: 66810752-66810752 on Build GRCh38
rs919987588 Chr.16: 66810852-66810857 on Build GRCh38
rs772975515 Chr.16: 66810749-66810749 on Build GRCh38
rs376653476 Chr.16: 66810655-66810655 on Build GRCh38
rs778538975 Chr.16: 66810675-66810676 on Build GRCh38
rs111527622 Chr.16: 66810500-66810500 on Build GRCh38
rs535123079 Chr.16: 66810931-66810931 on Build GRCh38
rs138689562 Chr.16: 66810834-66810834 on Build GRCh38
rs775506941 Chr.16: 66810646-66810646 on Build GRCh38
rs969342239 Chr.16: 66810789-66810789 on Build GRCh38
rs891557446 Chr.16: 66810610-66810610 on Build GRCh38
rs1039353355 Chr.16: 66810506-66810506 on Build GRCh38
rs761832532 Chr.16: 66810512-66810512 on Build GRCh38
rs373394037 Chr.16: 66810835-66810835 on Build GRCh38
rs919828960 Chr.16: 66810844-66810844 on Build GRCh38
rs752293590 Chr.16: 66810780-66810780 on Build GRCh38
rs769684888 Chr.16: 66810714-66810714 on Build GRCh38
rs755079628 Chr.16: 66810693-66810693 on Build GRCh38
rs756504617 Chr.16: 66810678-66810678 on Build GRCh38
rs550534934 Chr.16: 66810735-66810735 on Build GRCh38
rs948716601 Chr.16: 66810516-66810516 on Build GRCh38
rs973989397 Chr.16: 66810837-66810837 on Build GRCh38
rs186580935 Chr.16: 66810741-66810741 on Build GRCh38
rs528454553 Chr.16: 66810530-66810530 on Build GRCh38
rs1022537453 Chr.16: 66810574-66810574 on Build GRCh38
rs1011293203 Chr.16: 66810619-66810619 on Build GRCh38
rs368762132 Chr.16: 66810681-66810681 on Build GRCh38
rs748263443 Chr.16: 66810696-66810696 on Build GRCh38
rs774131902 Chr.16: 66810728-66810728 on Build GRCh38
rs749891818 Chr.16: 66810785-66810785 on Build GRCh38
rs773959510 Chr.16: 66810661-66810661 on Build GRCh38

View Full Product Details