Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3716545-3717114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGAE
Entrez Gene ID
3682
Gene Name
integrin subunit alpha E
Gene Aliases
CD103, HUMINAE
UniGene
Hs.513867
Gene Chromosome Location
Chr.17: 3714625-3803766 on Build GRCh38
Transcripts
NM_002208.4 XM_011523823.2 XM_011523825.2 XM_011523827.2 XM_011523828.2 XM_017024586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1135924 c.3436C>A p.L1146M Chr.17: 3716696-3716696 on Build GRCh38
436448 c.3423G>A p.V1141V Chr.17: 3716709-3716709 on Build GRCh38
2836597 c.3343G>A p.V1115I Chr.17: 3716789-3716789 on Build GRCh38
7186720 c.3444+3C>G p.? Chr.17: 3716685-3716685 on Build GRCh38
6343445 c.3420C>A p.I1140I Chr.17: 3716712-3716712 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs532606106 Chr.17: 3717091-3717091 on Build GRCh38
rs745958802 Chr.17: 3716714-3716714 on Build GRCh38
rs560362735 Chr.17: 3717073-3717073 on Build GRCh38
rs185094011 Chr.17: 3717026-3717026 on Build GRCh38
rs749111583 Chr.17: 3716827-3716827 on Build GRCh38
rs189706106 Chr.17: 3717044-3717044 on Build GRCh38
rs914354795 Chr.17: 3716868-3716868 on Build GRCh38
rs186636209 Chr.17: 3716789-3716789 on Build GRCh38
rs933395141 Chr.17: 3716573-3716573 on Build GRCh38
rs913071471 Chr.17: 3716883-3716883 on Build GRCh38
rs552633532 Chr.17: 3716790-3716790 on Build GRCh38
rs954837252 Chr.17: 3717037-3717037 on Build GRCh38
rs745872275 Chr.17: 3716832-3716832 on Build GRCh38
rs767100731 Chr.17: 3716764-3716764 on Build GRCh38
rs567370514 Chr.17: 3716843-3716843 on Build GRCh38
rs555119696 Chr.17: 3717020-3717020 on Build GRCh38
rs775264592 Chr.17: 3716777-3716777 on Build GRCh38
rs1055487140 Chr.17: 3716919-3716919 on Build GRCh38
rs139778490 Chr.17: 3716712-3716712 on Build GRCh38
rs924301873 Chr.17: 3716824-3716824 on Build GRCh38
rs756522123 Chr.17: 3716685-3716685 on Build GRCh38
rs367915381 Chr.17: 3716646-3716646 on Build GRCh38
rs993620240 Chr.17: 3716720-3716720 on Build GRCh38
rs770793667 Chr.17: 3716753-3716753 on Build GRCh38
rs757846390 Chr.17: 3716711-3716711 on Build GRCh38
rs530165366 Chr.17: 3716639-3716639 on Build GRCh38
rs758608457 Chr.17: 3716721-3716721 on Build GRCh38
rs757736774 Chr.17: 3716799-3716799 on Build GRCh38
rs897868413 Chr.17: 3716927-3716927 on Build GRCh38
rs1049154441 Chr.17: 3716947-3716947 on Build GRCh38
rs1014400760 Chr.17: 3716746-3716746 on Build GRCh38
rs1025784528 Chr.17: 3716958-3716958 on Build GRCh38
rs759593542 Chr.17: 3716793-3716793 on Build GRCh38
rs747280807 Chr.17: 3716735-3716735 on Build GRCh38
rs571555154 Chr.17: 3716803-3716803 on Build GRCh38
rs904199344 Chr.17: 3716682-3716682 on Build GRCh38
rs921915599 Chr.17: 3716838-3716838 on Build GRCh38
rs764615796 Chr.17: 3716690-3716690 on Build GRCh38
rs372145193 Chr.17: 3716792-3716792 on Build GRCh38
rs750254984 Chr.17: 3716581-3716581 on Build GRCh38
rs2293882 Chr.17: 3717075-3717075 on Build GRCh38
rs151074270 Chr.17: 3716699-3716699 on Build GRCh38
rs769120413 Chr.17: 3716810-3716811 on Build GRCh38
rs777492867 Chr.17: 3716823-3716823 on Build GRCh38
rs145699046 Chr.17: 3716970-3716970 on Build GRCh38
rs561157916 Chr.17: 3716580-3716580 on Build GRCh38
rs1005094160 Chr.17: 3716988-3716988 on Build GRCh38
rs1017756476 Chr.17: 3717001-3717001 on Build GRCh38
rs758935543 Chr.17: 3716760-3716760 on Build GRCh38
rs138268886 Chr.17: 3717015-3717015 on Build GRCh38
rs937245160 Chr.17: 3716903-3716903 on Build GRCh38
rs945843968 Chr.17: 3716887-3716887 on Build GRCh38
rs778065461 Chr.17: 3716902-3716902 on Build GRCh38
rs780108948 Chr.17: 3716724-3716724 on Build GRCh38
rs780561785 Chr.17: 3716648-3716648 on Build GRCh38
rs558173024 Chr.17: 3716969-3716969 on Build GRCh38
rs773869101 Chr.17: 3716755-3716755 on Build GRCh38
rs770796238 Chr.17: 3716828-3716828 on Build GRCh38
rs780486501 Chr.17: 3716719-3716723 on Build GRCh38
rs749164841 Chr.17: 3716751-3716751 on Build GRCh38
rs750980598 Chr.17: 3716810-3716810 on Build GRCh38
rs2293881 Chr.17: 3716844-3716844 on Build GRCh38
rs769573033 Chr.17: 3716736-3716736 on Build GRCh38
rs538242753 Chr.17: 3716929-3716929 on Build GRCh38
rs530833990 Chr.17: 3716809-3716809 on Build GRCh38
rs201136101 Chr.17: 3716741-3716741 on Build GRCh38
rs541829334 Chr.17: 3716610-3716610 on Build GRCh38
rs371158270 Chr.17: 3716889-3716889 on Build GRCh38
rs146193411 Chr.17: 3716847-3716848 on Build GRCh38
rs945979322 Chr.17: 3716633-3716633 on Build GRCh38
rs752435583 Chr.17: 3716655-3716655 on Build GRCh38
rs758386918 Chr.17: 3716815-3716815 on Build GRCh38
rs770836838 Chr.17: 3716594-3716594 on Build GRCh38
rs1029007533 Chr.17: 3717036-3717036 on Build GRCh38
rs372884911 Chr.17: 3716829-3716829 on Build GRCh38
rs190697833 Chr.17: 3716818-3716818 on Build GRCh38
rs755205566 Chr.17: 3716819-3716819 on Build GRCh38
rs566588479 Chr.17: 3716900-3716900 on Build GRCh38
rs1014019324 Chr.17: 3716955-3716955 on Build GRCh38
rs747433087 Chr.17: 3716806-3716809 on Build GRCh38
rs546178190 Chr.17: 3717085-3717085 on Build GRCh38
rs759178212 Chr.17: 3716645-3716645 on Build GRCh38

View Full Product Details