Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18347871-18348467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHMT1
Entrez Gene ID
6470
Gene Name
serine hydroxymethyltransferase 1
Gene Aliases
CSHMT, SHMT
UniGene
Hs.513987
Gene Chromosome Location
Chr.17: 18327860-18363563 on Build GRCh38
Transcripts
NM_001281786.1 NM_004169.4 NM_148918.2 XM_005256767.3 XM_011523992.2 XM_017024957.1 XM_017024958.1 XM_017024959.1 XM_017024960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
336499 c.343G>T p.V115F Chr.17: 18348340-18348340 on Build GRCh38
1644249 c.256A>G p.T86A Chr.17: 18348427-18348427 on Build GRCh38
5508886 c.248A>G p.Y83C Chr.17: 18348435-18348435 on Build GRCh38
5516761 c.252C>T p.G84G Chr.17: 18348431-18348431 on Build GRCh38
2741115 c.249T>C p.Y83Y Chr.17: 18348434-18348434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777674610 Chr.17: 18348221-18348230 on Build GRCh38
rs201892779 Chr.17: 18347917-18347918 on Build GRCh38
rs753386260 Chr.17: 18348222-18348223 on Build GRCh38
rs781272443 Chr.17: 18348251-18348251 on Build GRCh38
rs771814434 Chr.17: 18348279-18348279 on Build GRCh38
rs539125909 Chr.17: 18348125-18348125 on Build GRCh38
rs887814529 Chr.17: 18348151-18348151 on Build GRCh38
rs572272163 Chr.17: 18348215-18348215 on Build GRCh38
rs756047338 Chr.17: 18348260-18348260 on Build GRCh38
rs998532296 Chr.17: 18348376-18348376 on Build GRCh38
rs192601872 Chr.17: 18348067-18348067 on Build GRCh38
rs886534933 Chr.17: 18347967-18347967 on Build GRCh38
rs768178817 Chr.17: 18348417-18348417 on Build GRCh38
rs1012256712 Chr.17: 18348305-18348305 on Build GRCh38
rs1007089760 Chr.17: 18348252-18348252 on Build GRCh38
rs928425360 Chr.17: 18348164-18348164 on Build GRCh38
rs913448136 Chr.17: 18348085-18348085 on Build GRCh38
rs748182608 Chr.17: 18348256-18348256 on Build GRCh38
rs201416040 Chr.17: 18347921-18347922 on Build GRCh38
rs1051017594 Chr.17: 18348387-18348387 on Build GRCh38
rs369859224 Chr.17: 18348294-18348294 on Build GRCh38
rs370302733 Chr.17: 18348291-18348291 on Build GRCh38
rs753794929 Chr.17: 18348350-18348350 on Build GRCh38
rs149044124 Chr.17: 18348297-18348297 on Build GRCh38
rs570401117 Chr.17: 18348082-18348082 on Build GRCh38
rs556630139 Chr.17: 18347980-18347980 on Build GRCh38
rs950266925 Chr.17: 18348102-18348102 on Build GRCh38
rs182979330 Chr.17: 18348148-18348148 on Build GRCh38
rs565857335 Chr.17: 18347920-18347920 on Build GRCh38
rs745592767 Chr.17: 18348368-18348368 on Build GRCh38
rs1032781619 Chr.17: 18348406-18348406 on Build GRCh38
rs536603170 Chr.17: 18348041-18348041 on Build GRCh38
rs935648562 Chr.17: 18348006-18348006 on Build GRCh38
rs188938011 Chr.17: 18347995-18347995 on Build GRCh38
rs201620237 Chr.17: 18348275-18348275 on Build GRCh38
rs756737767 Chr.17: 18348345-18348346 on Build GRCh38
rs906019535 Chr.17: 18348147-18348147 on Build GRCh38
rs766575172 Chr.17: 18348238-18348238 on Build GRCh38
rs11649868 Chr.17: 18347999-18347999 on Build GRCh38
rs548470879 Chr.17: 18347948-18347948 on Build GRCh38
rs763264987 Chr.17: 18348301-18348301 on Build GRCh38
rs1032752134 Chr.17: 18348262-18348262 on Build GRCh38
rs1006690995 Chr.17: 18348394-18348394 on Build GRCh38
rs753251331 Chr.17: 18347914-18347915 on Build GRCh38
rs751773021 Chr.17: 18348239-18348239 on Build GRCh38
rs758183699 Chr.17: 18348292-18348292 on Build GRCh38
rs1029936829 Chr.17: 18348214-18348214 on Build GRCh38
rs758778542 Chr.17: 18348229-18348229 on Build GRCh38
rs1049133022 Chr.17: 18348149-18348149 on Build GRCh38
rs541251383 Chr.17: 18348268-18348268 on Build GRCh38
rs879494793 Chr.17: 18348172-18348172 on Build GRCh38
rs746171771 Chr.17: 18348431-18348431 on Build GRCh38
rs575084452 Chr.17: 18348328-18348328 on Build GRCh38
rs1036910543 Chr.17: 18348179-18348179 on Build GRCh38
rs756170223 Chr.17: 18348347-18348347 on Build GRCh38
rs889650535 Chr.17: 18348388-18348388 on Build GRCh38
rs528327583 Chr.17: 18347941-18347941 on Build GRCh38
rs997640279 Chr.17: 18347943-18347944 on Build GRCh38
rs759784953 Chr.17: 18348336-18348336 on Build GRCh38
rs576584211 Chr.17: 18348145-18348145 on Build GRCh38
rs143078306 Chr.17: 18348437-18348437 on Build GRCh38
rs779841160 Chr.17: 18348401-18348401 on Build GRCh38
rs757097029 Chr.17: 18348353-18348353 on Build GRCh38
rs762441672 Chr.17: 18348199-18348199 on Build GRCh38
rs369513722 Chr.17: 18348365-18348365 on Build GRCh38
rs991222301 Chr.17: 18348138-18348138 on Build GRCh38
rs12451807 Chr.17: 18347922-18347922 on Build GRCh38
rs953896603 Chr.17: 18347890-18347890 on Build GRCh38
rs868694307 Chr.17: 18347969-18347969 on Build GRCh38
rs867625667 Chr.17: 18348325-18348325 on Build GRCh38
rs892825556 Chr.17: 18347921-18347921 on Build GRCh38
rs1004998884 Chr.17: 18347981-18347981 on Build GRCh38
rs1005461108 Chr.17: 18348273-18348273 on Build GRCh38
rs775145425 Chr.17: 18348284-18348284 on Build GRCh38
rs867624411 Chr.17: 18347915-18347915 on Build GRCh38
rs909260915 Chr.17: 18348321-18348321 on Build GRCh38
rs760205244 Chr.17: 18348286-18348286 on Build GRCh38
rs752391707 Chr.17: 18348267-18348267 on Build GRCh38
rs765774231 Chr.17: 18348200-18348200 on Build GRCh38
rs372703278 Chr.17: 18348180-18348180 on Build GRCh38
rs754489023 Chr.17: 18348170-18348170 on Build GRCh38
rs373431106 Chr.17: 18348430-18348430 on Build GRCh38
rs370293626 Chr.17: 18348044-18348044 on Build GRCh38
rs752784615 Chr.17: 18348346-18348346 on Build GRCh38
rs376257532 Chr.17: 18348341-18348341 on Build GRCh38
rs1004805101 Chr.17: 18348173-18348173 on Build GRCh38
rs746585562 Chr.17: 18348412-18348412 on Build GRCh38
rs745559978 Chr.17: 18348278-18348278 on Build GRCh38
rs777584160 Chr.17: 18348318-18348318 on Build GRCh38
rs773639730 Chr.17: 18348418-18348418 on Build GRCh38
rs4924850 Chr.17: 18347918-18347918 on Build GRCh38
rs763537356 Chr.17: 18348427-18348427 on Build GRCh38
rs755099064 Chr.17: 18348247-18348247 on Build GRCh38

View Full Product Details