Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 35772707-35773246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK1
Entrez Gene ID
9201
Gene Name
doublecortin like kinase 1
Gene Aliases
CL1, CLICK1, DCAMKL1, DCDC3A, DCLK
UniGene
Hs.507755
Gene Chromosome Location
Chr.13: 35768652-36132020 on Build GRCh38
Transcripts
NM_001195415.1 NM_001195416.1 NM_004734.4 XM_005266592.2 XM_006719893.2 XM_017020847.1

SNP Detail

SNP ID Chromosome Location
rs554678396 Chr.13: 35773171-35773171 on Build GRCh38
rs143078391 Chr.13: 35772899-35772899 on Build GRCh38
rs916915148 Chr.13: 35773105-35773105 on Build GRCh38
rs563578508 Chr.13: 35772864-35772864 on Build GRCh38
rs898072324 Chr.13: 35773048-35773048 on Build GRCh38
rs994503986 Chr.13: 35773091-35773091 on Build GRCh38
rs895988776 Chr.13: 35773034-35773034 on Build GRCh38
rs1027261138 Chr.13: 35773103-35773103 on Build GRCh38
rs9574551 Chr.13: 35772869-35772869 on Build GRCh38
rs1014383206 Chr.13: 35773043-35773043 on Build GRCh38
rs571509202 Chr.13: 35773006-35773006 on Build GRCh38
rs2182060 Chr.13: 35773061-35773061 on Build GRCh38
rs987958467 Chr.13: 35773219-35773219 on Build GRCh38
rs953133037 Chr.13: 35772830-35772830 on Build GRCh38
rs557227665 Chr.13: 35773042-35773042 on Build GRCh38
rs1043074944 Chr.13: 35772956-35772956 on Build GRCh38
rs868760673 Chr.13: 35773162-35773162 on Build GRCh38
rs1001743164 Chr.13: 35772744-35772744 on Build GRCh38
rs1050635098 Chr.13: 35773201-35773201 on Build GRCh38
rs17052784 Chr.13: 35772825-35772825 on Build GRCh38
rs780320640 Chr.13: 35772958-35772958 on Build GRCh38
rs534489880 Chr.13: 35772967-35772967 on Build GRCh38
rs181927725 Chr.13: 35772773-35772773 on Build GRCh38
rs191926527 Chr.13: 35773135-35773135 on Build GRCh38
rs962016144 Chr.13: 35772817-35772817 on Build GRCh38
rs750690546 Chr.13: 35773128-35773128 on Build GRCh38
rs973007057 Chr.13: 35772820-35772820 on Build GRCh38
rs1015597333 Chr.13: 35773047-35773047 on Build GRCh38
rs1034488996 Chr.13: 35773053-35773053 on Build GRCh38
rs917469510 Chr.13: 35772762-35772762 on Build GRCh38
rs958460487 Chr.13: 35773096-35773096 on Build GRCh38
rs946039349 Chr.13: 35772939-35772939 on Build GRCh38
rs780806612 Chr.13: 35772848-35772848 on Build GRCh38
rs144219290 Chr.13: 35772900-35772900 on Build GRCh38
rs558935033 Chr.13: 35772875-35772875 on Build GRCh38
rs911703489 Chr.13: 35772858-35772858 on Build GRCh38
rs386769942 Chr.13: 35772869-35772870 on Build GRCh38
rs913842873 Chr.13: 35773221-35773221 on Build GRCh38
rs889103273 Chr.13: 35772860-35772860 on Build GRCh38
rs533487691 Chr.13: 35772810-35772810 on Build GRCh38
rs910611697 Chr.13: 35773209-35773209 on Build GRCh38
rs944782246 Chr.13: 35773217-35773217 on Build GRCh38
rs904727448 Chr.13: 35772780-35772780 on Build GRCh38
rs1055852969 Chr.13: 35773031-35773031 on Build GRCh38
rs1014933022 Chr.13: 35772776-35772776 on Build GRCh38
rs904573969 Chr.13: 35772730-35772730 on Build GRCh38
rs755767657 Chr.13: 35772893-35772893 on Build GRCh38
rs538603179 Chr.13: 35773013-35773013 on Build GRCh38
rs1015151914 Chr.13: 35772793-35772793 on Build GRCh38
rs992470429 Chr.13: 35772756-35772756 on Build GRCh38
rs541664939 Chr.13: 35772762-35772762 on Build GRCh38
rs186576496 Chr.13: 35773119-35773119 on Build GRCh38
rs546512142 Chr.13: 35772983-35772983 on Build GRCh38
rs978111480 Chr.13: 35773129-35773129 on Build GRCh38

View Full Product Details