Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23992445-23992982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC39C
Entrez Gene ID
125488
Gene Name
tetratricopeptide repeat domain 39C
Gene Aliases
C18orf17, HsT2697
UniGene
Hs.128576
Gene Chromosome Location
Chr.18: 23992773-24135952 on Build GRCh38
Transcripts
NM_153211.3 XM_017025550.1

SNP Detail

SNP ID Chromosome Location
rs992381962 Chr.18: 23992656-23992656 on Build GRCh38
rs867969551 Chr.18: 23992595-23992595 on Build GRCh38
rs535623751 Chr.18: 23992582-23992582 on Build GRCh38
rs966211904 Chr.18: 23992543-23992543 on Build GRCh38
rs576816971 Chr.18: 23992765-23992765 on Build GRCh38
rs984714009 Chr.18: 23992583-23992583 on Build GRCh38
rs150596671 Chr.18: 23992833-23992833 on Build GRCh38
rs553297917 Chr.18: 23992513-23992513 on Build GRCh38
rs928642421 Chr.18: 23992903-23992903 on Build GRCh38
rs188395939 Chr.18: 23992584-23992584 on Build GRCh38
rs557282507 Chr.18: 23992732-23992732 on Build GRCh38
rs890077914 Chr.18: 23992761-23992761 on Build GRCh38
rs138097323 Chr.18: 23992578-23992578 on Build GRCh38
rs948697853 Chr.18: 23992586-23992586 on Build GRCh38
rs978579178 Chr.18: 23992604-23992604 on Build GRCh38
rs772104181 Chr.18: 23992687-23992687 on Build GRCh38
rs1007216359 Chr.18: 23992510-23992510 on Build GRCh38
rs960845517 Chr.18: 23992630-23992630 on Build GRCh38
rs901690373 Chr.18: 23992819-23992819 on Build GRCh38
rs773001523 Chr.18: 23992725-23992725 on Build GRCh38
rs139567949 Chr.18: 23992949-23992949 on Build GRCh38
rs999641987 Chr.18: 23992934-23992934 on Build GRCh38
rs747653132 Chr.18: 23992488-23992488 on Build GRCh38
rs559595901 Chr.18: 23992802-23992802 on Build GRCh38
rs113565185 Chr.18: 23992947-23992947 on Build GRCh38
rs934726058 Chr.18: 23992915-23992915 on Build GRCh38
rs924154017 Chr.18: 23992628-23992628 on Build GRCh38
rs865785131 Chr.18: 23992535-23992535 on Build GRCh38
rs562163206 Chr.18: 23992874-23992874 on Build GRCh38
rs1022868579 Chr.18: 23992867-23992867 on Build GRCh38
rs533017809 Chr.18: 23992839-23992839 on Build GRCh38
rs74704974 Chr.18: 23992511-23992511 on Build GRCh38
rs993671768 Chr.18: 23992807-23992807 on Build GRCh38
rs953059483 Chr.18: 23992481-23992481 on Build GRCh38
rs940477545 Chr.18: 23992779-23992779 on Build GRCh38
rs984660211 Chr.18: 23992846-23992846 on Build GRCh38
rs568035439 Chr.18: 23992961-23992961 on Build GRCh38
rs142649290 Chr.18: 23992780-23992780 on Build GRCh38
rs1045783793 Chr.18: 23992610-23992610 on Build GRCh38
rs971910681 Chr.18: 23992561-23992561 on Build GRCh38
rs543574330 Chr.18: 23992692-23992692 on Build GRCh38
rs919097908 Chr.18: 23992563-23992563 on Build GRCh38
rs192835040 Chr.18: 23992671-23992671 on Build GRCh38
rs893299169 Chr.18: 23992717-23992717 on Build GRCh38
rs770664728 Chr.18: 23992771-23992771 on Build GRCh38
rs12327412 Chr.18: 23992873-23992873 on Build GRCh38
rs565824952 Chr.18: 23992670-23992670 on Build GRCh38
rs760558263 Chr.18: 23992756-23992756 on Build GRCh38

View Full Product Details