Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35813019-35813618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRODH2
Entrez Gene ID
58510
Gene Name
proline dehydrogenase 2
Gene Aliases
HSPOX1, HYPDH
UniGene
Hs.515366
Gene Chromosome Location
Chr.19: 35799987-35813299 on Build GRCh38
Transcripts
NM_021232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7385628 c.135C>A p.H45Q Chr.19: 35813147-35813147 on Build GRCh38
6392885 c.43C>G p.Q15E Chr.19: 35813239-35813239 on Build GRCh38
6006190 c.4T>A p.S2T Chr.19: 35813278-35813278 on Build GRCh38
246789 c.6G>A p.S2S Chr.19: 35813276-35813276 on Build GRCh38
6756412 c.115T>G p.L39V Chr.19: 35813167-35813167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537085071 Chr.19: 35813374-35813374 on Build GRCh38
rs963359346 Chr.19: 35813192-35813192 on Build GRCh38
rs759394636 Chr.19: 35813108-35813108 on Build GRCh38
rs975227681 Chr.19: 35813350-35813350 on Build GRCh38
rs370045721 Chr.19: 35813279-35813279 on Build GRCh38
rs756980355 Chr.19: 35813177-35813177 on Build GRCh38
rs894767281 Chr.19: 35813056-35813056 on Build GRCh38
rs765106136 Chr.19: 35813234-35813234 on Build GRCh38
rs983522588 Chr.19: 35813156-35813156 on Build GRCh38
rs191814559 Chr.19: 35813502-35813502 on Build GRCh38
rs775255491 Chr.19: 35813238-35813238 on Build GRCh38
rs1025961572 Chr.19: 35813272-35813272 on Build GRCh38
rs893494522 Chr.19: 35813080-35813080 on Build GRCh38
rs142757847 Chr.19: 35813243-35813243 on Build GRCh38
rs138605929 Chr.19: 35813241-35813241 on Build GRCh38
rs774195173 Chr.19: 35813128-35813128 on Build GRCh38
rs1003867046 Chr.19: 35813125-35813125 on Build GRCh38
rs755345232 Chr.19: 35813266-35813266 on Build GRCh38
rs754269960 Chr.19: 35813530-35813530 on Build GRCh38
rs375758158 Chr.19: 35813253-35813253 on Build GRCh38
rs906183924 Chr.19: 35813258-35813258 on Build GRCh38
rs145525145 Chr.19: 35813453-35813453 on Build GRCh38
rs973917501 Chr.19: 35813440-35813440 on Build GRCh38
rs61995739 Chr.19: 35813178-35813178 on Build GRCh38
rs376497837 Chr.19: 35813107-35813107 on Build GRCh38
rs891099750 Chr.19: 35813393-35813393 on Build GRCh38
rs1005424070 Chr.19: 35813534-35813534 on Build GRCh38
rs547294948 Chr.19: 35813276-35813276 on Build GRCh38
rs749090323 Chr.19: 35813209-35813209 on Build GRCh38
rs1008150427 Chr.19: 35813406-35813406 on Build GRCh38
rs767272272 Chr.19: 35813527-35813527 on Build GRCh38
rs1011890674 Chr.19: 35813100-35813100 on Build GRCh38
rs961269580 Chr.19: 35813422-35813422 on Build GRCh38
rs1025509390 Chr.19: 35813150-35813150 on Build GRCh38
rs759304825 Chr.19: 35813232-35813232 on Build GRCh38
rs183424756 Chr.19: 35813588-35813588 on Build GRCh38
rs766738320 Chr.19: 35813461-35813461 on Build GRCh38
rs557520170 Chr.19: 35813247-35813247 on Build GRCh38
rs1045927146 Chr.19: 35813098-35813098 on Build GRCh38
rs141011767 Chr.19: 35813109-35813109 on Build GRCh38
rs779365971 Chr.19: 35813270-35813270 on Build GRCh38
rs575127601 Chr.19: 35813497-35813497 on Build GRCh38
rs899666098 Chr.19: 35813546-35813546 on Build GRCh38
rs938989287 Chr.19: 35813517-35813517 on Build GRCh38
rs559174881 Chr.19: 35813336-35813336 on Build GRCh38
rs373218136 Chr.19: 35813281-35813281 on Build GRCh38
rs757801652 Chr.19: 35813142-35813142 on Build GRCh38
rs576152305 Chr.19: 35813184-35813184 on Build GRCh38
rs547432930 Chr.19: 35813315-35813315 on Build GRCh38
rs748711402 Chr.19: 35813198-35813198 on Build GRCh38
rs569693602 Chr.19: 35813365-35813365 on Build GRCh38
rs138137261 Chr.19: 35813593-35813593 on Build GRCh38
rs376471343 Chr.19: 35813321-35813321 on Build GRCh38
rs553616186 Chr.19: 35813496-35813496 on Build GRCh38
rs915408400 Chr.19: 35813458-35813458 on Build GRCh38
rs779927154 Chr.19: 35813136-35813136 on Build GRCh38
rs758955765 Chr.19: 35813277-35813277 on Build GRCh38
rs535175924 Chr.19: 35813483-35813483 on Build GRCh38
rs750145940 Chr.19: 35813261-35813261 on Build GRCh38
rs144717897 Chr.19: 35813196-35813196 on Build GRCh38
rs1038202327 Chr.19: 35813544-35813544 on Build GRCh38
rs777377500 Chr.19: 35813146-35813146 on Build GRCh38
rs529946284 Chr.19: 35813338-35813338 on Build GRCh38
rs924161839 Chr.19: 35813381-35813381 on Build GRCh38
rs141771271 Chr.19: 35813421-35813421 on Build GRCh38
rs542606348 Chr.19: 35813154-35813154 on Build GRCh38
rs374395555 Chr.19: 35813216-35813216 on Build GRCh38
rs761989167 Chr.19: 35813250-35813250 on Build GRCh38
rs755429391 Chr.19: 35813188-35813188 on Build GRCh38
rs924130595 Chr.19: 35813219-35813219 on Build GRCh38
rs202071717 Chr.19: 35813130-35813130 on Build GRCh38
rs776879746 Chr.19: 35813218-35813218 on Build GRCh38
rs186871964 Chr.19: 35813351-35813351 on Build GRCh38
rs989607862 Chr.19: 35813446-35813446 on Build GRCh38
rs769056352 Chr.19: 35813210-35813210 on Build GRCh38
rs751098769 Chr.19: 35813252-35813252 on Build GRCh38
rs553676884 Chr.19: 35813079-35813079 on Build GRCh38
rs1045201916 Chr.19: 35813492-35813492 on Build GRCh38
rs145723152 Chr.19: 35813211-35813211 on Build GRCh38

View Full Product Details