Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55094567-55095026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R12C
Entrez Gene ID
54776
Gene Name
protein phosphatase 1 regulatory subunit 12C
Gene Aliases
LENG3, MBS85, p84, p85
UniGene
Hs.631579
Gene Chromosome Location
Chr.19: 55090913-55117600 on Build GRCh38
Transcripts
NM_001271618.1 NM_017607.3 XM_005259013.3 XM_011527045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5579376 c.1592+2T>G p.? Chr.19: 55094659-55094659 on Build GRCh38
1712851 c.1556_1557CC>TT p.S519F Chr.19: 55094696-55094697 on Build GRCh38
3539344 c.1513C>G p.P505A Chr.19: 55094740-55094740 on Build GRCh38
6038260 c.1497G>A p.S499S Chr.19: 55094756-55094756 on Build GRCh38
4528935 c.1573G>A p.D525N Chr.19: 55094680-55094680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767164579 Chr.19: 55094634-55094634 on Build GRCh38
rs946779482 Chr.19: 55094835-55094835 on Build GRCh38
rs141058127 Chr.19: 55094665-55094665 on Build GRCh38
rs754411879 Chr.19: 55094837-55094838 on Build GRCh38
rs911331940 Chr.19: 55094834-55094834 on Build GRCh38
rs893577812 Chr.19: 55094849-55094849 on Build GRCh38
rs1016261281 Chr.19: 55094968-55094968 on Build GRCh38
rs191653168 Chr.19: 55094783-55094783 on Build GRCh38
rs545737805 Chr.19: 55094969-55094969 on Build GRCh38
rs755655737 Chr.19: 55094704-55094704 on Build GRCh38
rs755982305 Chr.19: 55094798-55094799 on Build GRCh38
rs1028221760 Chr.19: 55094899-55094899 on Build GRCh38
rs776879095 Chr.19: 55094610-55094610 on Build GRCh38
rs1013854198 Chr.19: 55094648-55094648 on Build GRCh38
rs746830894 Chr.19: 55094738-55094738 on Build GRCh38
rs775732875 Chr.19: 55094757-55094757 on Build GRCh38
rs556028464 Chr.19: 55094970-55094970 on Build GRCh38
rs376586289 Chr.19: 55094609-55094609 on Build GRCh38
rs376368537 Chr.19: 55094792-55094792 on Build GRCh38
rs752254667 Chr.19: 55094848-55094848 on Build GRCh38
rs972059540 Chr.19: 55094990-55094990 on Build GRCh38
rs889607140 Chr.19: 55094953-55094953 on Build GRCh38
rs776790606 Chr.19: 55094679-55094679 on Build GRCh38
rs1005700310 Chr.19: 55094681-55094681 on Build GRCh38
rs200477573 Chr.19: 55094823-55094823 on Build GRCh38
rs78085086 Chr.19: 55094713-55094713 on Build GRCh38
rs1044022957 Chr.19: 55094986-55094986 on Build GRCh38
rs767168829 Chr.19: 55094621-55094622 on Build GRCh38
rs766302425 Chr.19: 55094800-55094801 on Build GRCh38
rs770543247 Chr.19: 55094828-55094828 on Build GRCh38
rs377413098 Chr.19: 55094735-55094735 on Build GRCh38
rs758756283 Chr.19: 55094633-55094633 on Build GRCh38
rs750089999 Chr.19: 55094635-55094635 on Build GRCh38
rs142994105 Chr.19: 55094689-55094689 on Build GRCh38
rs998773529 Chr.19: 55094623-55094623 on Build GRCh38
rs925814558 Chr.19: 55094885-55094885 on Build GRCh38
rs1005348436 Chr.19: 55094963-55094963 on Build GRCh38
rs373151753 Chr.19: 55094721-55094721 on Build GRCh38
rs751987617 Chr.19: 55094699-55094699 on Build GRCh38
rs753533440 Chr.19: 55094807-55094807 on Build GRCh38
rs1042892605 Chr.19: 55094589-55094589 on Build GRCh38
rs748801901 Chr.19: 55094652-55094652 on Build GRCh38
rs541313591 Chr.19: 55094590-55094590 on Build GRCh38
rs372592192 Chr.19: 55094775-55094775 on Build GRCh38
rs759747513 Chr.19: 55094682-55094682 on Build GRCh38
rs778609444 Chr.19: 55094858-55094858 on Build GRCh38
rs770041262 Chr.19: 55094756-55094756 on Build GRCh38
rs369559413 Chr.19: 55094739-55094739 on Build GRCh38
rs763353251 Chr.19: 55094691-55094691 on Build GRCh38
rs756759969 Chr.19: 55094942-55094942 on Build GRCh38
rs533800108 Chr.19: 55094877-55094877 on Build GRCh38
rs202073659 Chr.19: 55094620-55094620 on Build GRCh38
rs901087310 Chr.19: 55094608-55094608 on Build GRCh38
rs750235634 Chr.19: 55094774-55094776 on Build GRCh38
rs754554284 Chr.19: 55094720-55094720 on Build GRCh38
rs73932302 Chr.19: 55094732-55094732 on Build GRCh38
rs1053064060 Chr.19: 55094900-55094900 on Build GRCh38
rs568038065 Chr.19: 55094856-55094856 on Build GRCh38
rs764680624 Chr.19: 55094832-55094832 on Build GRCh38
rs778608499 Chr.19: 55094987-55094987 on Build GRCh38
rs759026882 Chr.19: 55094818-55094818 on Build GRCh38
rs748089061 Chr.19: 55094727-55094727 on Build GRCh38
rs764318597 Chr.19: 55094694-55094694 on Build GRCh38
rs377524915 Chr.19: 55094772-55094772 on Build GRCh38
rs753097909 Chr.19: 55094627-55094627 on Build GRCh38
rs771184538 Chr.19: 55094673-55094673 on Build GRCh38
rs753819183 Chr.19: 55094803-55094804 on Build GRCh38
rs765835542 Chr.19: 55094687-55094687 on Build GRCh38
rs779500948 Chr.19: 55094649-55094649 on Build GRCh38
rs753397894 Chr.19: 55094905-55094905 on Build GRCh38
rs373932501 Chr.19: 55094740-55094740 on Build GRCh38
rs1042455771 Chr.19: 55094867-55094867 on Build GRCh38
rs150000536 Chr.19: 55094780-55094780 on Build GRCh38
rs771805284 Chr.19: 55094667-55094667 on Build GRCh38
rs900871151 Chr.19: 55094872-55094872 on Build GRCh38
rs895696072 Chr.19: 55094915-55094915 on Build GRCh38
rs754852821 Chr.19: 55094657-55094657 on Build GRCh38
rs760049632 Chr.19: 55094616-55094616 on Build GRCh38
rs576730426 Chr.19: 55094916-55094916 on Build GRCh38
rs377033725 Chr.19: 55094999-55094999 on Build GRCh38
rs34972456 Chr.19: 55094733-55094733 on Build GRCh38
rs763331146 Chr.19: 55094761-55094761 on Build GRCh38
rs745911836 Chr.19: 55094748-55094748 on Build GRCh38
rs998509140 Chr.19: 55094891-55094891 on Build GRCh38
rs753337144 Chr.19: 55094718-55094718 on Build GRCh38
rs755814602 Chr.19: 55094642-55094642 on Build GRCh38
rs553311415 Chr.19: 55094887-55094887 on Build GRCh38

View Full Product Details