Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 75620302-75620838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMO7
Entrez Gene ID
4008
Gene Name
LIM domain 7
Gene Aliases
FBX20, FBXO20, LOMP
UniGene
Hs.207631
Gene Chromosome Location
Chr.13: 75620434-75859870 on Build GRCh38
Transcripts
NM_005358.5 NR_120410.1 NR_120411.1 NR_120412.1

Gene Symbol
LMO7-AS1
Entrez Gene ID
101927155
Gene Name
LMO7 antisense RNA 1
Gene Aliases
UCHL3-AS1
UniGene
Hs.564550
Gene Chromosome Location
Chr.13: 75604700-75635994 on Build GRCh38
Transcripts
NM_005358.5 NR_120410.1 NR_120411.1 NR_120412.1

SNP Detail

SNP ID Chromosome Location
rs891513570 Chr.13: 75620408-75620410 on Build GRCh38
rs571812746 Chr.13: 75620744-75620744 on Build GRCh38
rs1031445318 Chr.13: 75620398-75620398 on Build GRCh38
rs1022462332 Chr.13: 75620498-75620498 on Build GRCh38
rs755193252 Chr.13: 75620585-75620585 on Build GRCh38
rs548336942 Chr.13: 75620424-75620424 on Build GRCh38
rs901499049 Chr.13: 75620393-75620393 on Build GRCh38
rs543090023 Chr.13: 75620735-75620736 on Build GRCh38
rs559827895 Chr.13: 75620611-75620611 on Build GRCh38
rs545521629 Chr.13: 75620801-75620801 on Build GRCh38
rs537276999 Chr.13: 75620546-75620546 on Build GRCh38
rs967048245 Chr.13: 75620800-75620800 on Build GRCh38
rs879877208 Chr.13: 75620592-75620597 on Build GRCh38
rs111729806 Chr.13: 75620788-75620788 on Build GRCh38
rs1026744854 Chr.13: 75620627-75620627 on Build GRCh38
rs943580410 Chr.13: 75620466-75620466 on Build GRCh38
rs925033610 Chr.13: 75620364-75620364 on Build GRCh38
rs887811775 Chr.13: 75620633-75620633 on Build GRCh38
rs553887019 Chr.13: 75620762-75620762 on Build GRCh38
rs984628775 Chr.13: 75620718-75620718 on Build GRCh38
rs933006268 Chr.13: 75620763-75620763 on Build GRCh38
rs529955696 Chr.13: 75620358-75620358 on Build GRCh38
rs1049525084 Chr.13: 75620397-75620397 on Build GRCh38
rs992001166 Chr.13: 75620773-75620773 on Build GRCh38
rs1002439043 Chr.13: 75620578-75620578 on Build GRCh38
rs1040599770 Chr.13: 75620548-75620548 on Build GRCh38
rs1034125703 Chr.13: 75620580-75620580 on Build GRCh38
rs868185341 Chr.13: 75620805-75620805 on Build GRCh38
rs1033802852 Chr.13: 75620725-75620725 on Build GRCh38
rs12427750 Chr.13: 75620636-75620636 on Build GRCh38
rs375337932 Chr.13: 75620748-75620748 on Build GRCh38
rs182941279 Chr.13: 75620648-75620648 on Build GRCh38
rs968743093 Chr.13: 75620502-75620502 on Build GRCh38
rs944989355 Chr.13: 75620329-75620329 on Build GRCh38
rs936432848 Chr.13: 75620381-75620381 on Build GRCh38
rs978164115 Chr.13: 75620343-75620343 on Build GRCh38
rs774157451 Chr.13: 75620497-75620497 on Build GRCh38
rs745716163 Chr.13: 75620558-75620558 on Build GRCh38
rs889266307 Chr.13: 75620439-75620439 on Build GRCh38
rs993487611 Chr.13: 75620626-75620626 on Build GRCh38
rs924927951 Chr.13: 75620802-75620802 on Build GRCh38
rs950796442 Chr.13: 75620655-75620655 on Build GRCh38
rs569843361 Chr.13: 75620505-75620505 on Build GRCh38
rs535897083 Chr.13: 75620764-75620779 on Build GRCh38

View Full Product Details