Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 69160490-69161015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PJA1
Entrez Gene ID
64219
Gene Name
praja ring finger ubiquitin ligase 1
Gene Aliases
PRAJA1, RNF70
UniGene
Hs.522679
Gene Chromosome Location
Chr.X: 69160738-69165522 on Build GRCh38
Transcripts
NM_001032396.2 NM_022368.4 NM_145119.3 XM_005262292.1 XM_011531011.2

SNP Detail

SNP ID Chromosome Location
rs751635147 Chr.X: 69160720-69160723 on Build GRCh38
rs1047011687 Chr.X: 69160987-69160987 on Build GRCh38
rs34008712 Chr.X: 69160737-69160738 on Build GRCh38
rs3210690 Chr.X: 69160877-69160877 on Build GRCh38
rs949950432 Chr.X: 69160917-69160917 on Build GRCh38
rs781564583 Chr.X: 69160721-69160723 on Build GRCh38
rs909490842 Chr.X: 69160657-69160657 on Build GRCh38
rs968278829 Chr.X: 69160610-69160611 on Build GRCh38
rs12845039 Chr.X: 69160582-69160582 on Build GRCh38
rs965190339 Chr.X: 69160559-69160559 on Build GRCh38
rs925664931 Chr.X: 69160651-69160651 on Build GRCh38
rs910959526 Chr.X: 69160548-69160548 on Build GRCh38
rs141812548 Chr.X: 69160577-69160577 on Build GRCh38
rs139602630 Chr.X: 69160911-69160911 on Build GRCh38
rs1055339726 Chr.X: 69160907-69160907 on Build GRCh38
rs985334896 Chr.X: 69160517-69160517 on Build GRCh38
rs67825424 Chr.X: 69160736-69160738 on Build GRCh38
rs869110184 Chr.X: 69160738-69160738 on Build GRCh38
rs557666666 Chr.X: 69160722-69160723 on Build GRCh38
rs976914930 Chr.X: 69160566-69160566 on Build GRCh38
rs1055357222 Chr.X: 69160855-69160855 on Build GRCh38
rs886942146 Chr.X: 69160991-69160991 on Build GRCh38
rs774453394 Chr.X: 69160939-69160939 on Build GRCh38
rs942160490 Chr.X: 69160685-69160685 on Build GRCh38
rs763261021 Chr.X: 69160895-69160895 on Build GRCh38
rs375499183 Chr.X: 69160601-69160601 on Build GRCh38
rs949464539 Chr.X: 69160918-69160918 on Build GRCh38
rs144967927 Chr.X: 69160721-69160722 on Build GRCh38
rs746504694 Chr.X: 69160625-69160625 on Build GRCh38
rs753020260 Chr.X: 69160720-69160722 on Build GRCh38
rs76248501 Chr.X: 69160736-69160736 on Build GRCh38
rs770332916 Chr.X: 69160721-69160721 on Build GRCh38
rs17217584 Chr.X: 69160763-69160763 on Build GRCh38
rs914043189 Chr.X: 69160600-69160600 on Build GRCh38

View Full Product Details