Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111294560-111295091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCX
Entrez Gene ID
1641
Gene Name
doublecortin
Gene Aliases
DBCN, DC, LISX, SCLH, XLIS
UniGene
Hs.34780
Gene Chromosome Location
Chr.X: 111293779-111412232 on Build GRCh38
Transcripts
NM_000555.3 NM_001195553.1 NM_178151.2 NM_178152.2 NM_178153.2 XM_011530878.2 XM_011530879.2 XM_011530880.2 XM_017029312.1

SNP Detail

SNP ID Chromosome Location
rs761394618 Chr.X: 111294846-111294846 on Build GRCh38
rs772599099 Chr.X: 111294906-111294906 on Build GRCh38
rs746290222 Chr.X: 111294686-111294686 on Build GRCh38
rs878858509 Chr.X: 111294900-111294900 on Build GRCh38
rs1008141518 Chr.X: 111294839-111294839 on Build GRCh38
rs972851213 Chr.X: 111294589-111294589 on Build GRCh38
rs768265399 Chr.X: 111294704-111294704 on Build GRCh38
rs1032513885 Chr.X: 111294952-111294952 on Build GRCh38
rs956442995 Chr.X: 111294988-111294988 on Build GRCh38
rs922402509 Chr.X: 111294884-111294884 on Build GRCh38
rs767171752 Chr.X: 111295011-111295011 on Build GRCh38
rs750028081 Chr.X: 111295014-111295014 on Build GRCh38
rs868106050 Chr.X: 111294892-111294892 on Build GRCh38
rs1000675630 Chr.X: 111294898-111294898 on Build GRCh38
rs762996715 Chr.X: 111294648-111294648 on Build GRCh38
rs1041285858 Chr.X: 111294861-111294861 on Build GRCh38
rs186728581 Chr.X: 111294594-111294594 on Build GRCh38
rs760652354 Chr.X: 111294946-111294946 on Build GRCh38
rs999833820 Chr.X: 111295016-111295016 on Build GRCh38
rs756999424 Chr.X: 111294807-111294807 on Build GRCh38
rs773951275 Chr.X: 111294794-111294794 on Build GRCh38
rs927450217 Chr.X: 111294593-111294593 on Build GRCh38
rs1055435115 Chr.X: 111295052-111295052 on Build GRCh38
rs766994569 Chr.X: 111294883-111294883 on Build GRCh38
rs994584972 Chr.X: 111294745-111294745 on Build GRCh38
rs1041483790 Chr.X: 111294962-111294962 on Build GRCh38
rs1038878066 Chr.X: 111294649-111294649 on Build GRCh38
rs964202737 Chr.X: 111294880-111294880 on Build GRCh38
rs984119682 Chr.X: 111294655-111294655 on Build GRCh38
rs945883263 Chr.X: 111294930-111294930 on Build GRCh38
rs763989337 Chr.X: 111294776-111294776 on Build GRCh38
rs577339839 Chr.X: 111294953-111294953 on Build GRCh38
rs1018149829 Chr.X: 111294855-111294855 on Build GRCh38

View Full Product Details