Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 119590570-119591158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKRF
Entrez Gene ID
55922
Gene Name
NFKB repressing factor
Gene Aliases
ITBA4, NRF
UniGene
Hs.437084
Gene Chromosome Location
Chr.X: 119588337-119606464 on Build GRCh38
Transcripts
NM_001173487.1 NM_001173488.1 NM_017544.3 XM_011531365.2

SNP Detail

SNP ID Chromosome Location
rs772752791 Chr.X: 119590626-119590626 on Build GRCh38
rs763178022 Chr.X: 119591073-119591073 on Build GRCh38
rs371790847 Chr.X: 119590811-119590811 on Build GRCh38
rs369609047 Chr.X: 119591019-119591019 on Build GRCh38
rs763905869 Chr.X: 119590596-119590596 on Build GRCh38
rs147158383 Chr.X: 119590799-119590799 on Build GRCh38
rs754731180 Chr.X: 119590927-119590927 on Build GRCh38
rs755645742 Chr.X: 119590912-119590912 on Build GRCh38
rs201492954 Chr.X: 119590747-119590747 on Build GRCh38
rs779655222 Chr.X: 119590733-119590733 on Build GRCh38
rs766020454 Chr.X: 119590921-119590921 on Build GRCh38
rs752295461 Chr.X: 119590857-119590857 on Build GRCh38
rs760714444 Chr.X: 119591094-119591094 on Build GRCh38
rs753802390 Chr.X: 119591028-119591028 on Build GRCh38
rs1024696893 Chr.X: 119591134-119591134 on Build GRCh38
rs144614862 Chr.X: 119591018-119591018 on Build GRCh38
rs756050957 Chr.X: 119590920-119590920 on Build GRCh38
rs1007709547 Chr.X: 119590888-119590888 on Build GRCh38
rs759684907 Chr.X: 119590595-119590595 on Build GRCh38
rs144370117 Chr.X: 119590892-119590892 on Build GRCh38
rs765423675 Chr.X: 119590873-119590873 on Build GRCh38
rs767548753 Chr.X: 119590893-119590893 on Build GRCh38
rs775562576 Chr.X: 119591057-119591057 on Build GRCh38
rs201938262 Chr.X: 119590743-119590743 on Build GRCh38
rs757302342 Chr.X: 119591076-119591076 on Build GRCh38
rs763718010 Chr.X: 119590930-119590930 on Build GRCh38
rs143551890 Chr.X: 119590770-119590770 on Build GRCh38
rs370621437 Chr.X: 119590825-119590825 on Build GRCh38
rs751410646 Chr.X: 119590891-119590891 on Build GRCh38
rs137888647 Chr.X: 119590744-119590744 on Build GRCh38
rs1009241195 Chr.X: 119590752-119590752 on Build GRCh38
rs773574560 Chr.X: 119590774-119590774 on Build GRCh38
rs148494768 Chr.X: 119591083-119591083 on Build GRCh38
rs745916483 Chr.X: 119591051-119591051 on Build GRCh38
rs376820526 Chr.X: 119590992-119590992 on Build GRCh38
rs140519428 Chr.X: 119590846-119590846 on Build GRCh38
rs748442891 Chr.X: 119590754-119590754 on Build GRCh38
rs753352886 Chr.X: 119590926-119590926 on Build GRCh38
rs950928506 Chr.X: 119591059-119591059 on Build GRCh38
rs770119545 Chr.X: 119591052-119591052 on Build GRCh38
rs748601120 Chr.X: 119590935-119590935 on Build GRCh38
rs780960159 Chr.X: 119590666-119590666 on Build GRCh38
rs766399492 Chr.X: 119591135-119591135 on Build GRCh38
rs765871375 Chr.X: 119590823-119590823 on Build GRCh38
rs750017639 Chr.X: 119590690-119590690 on Build GRCh38
rs755672508 Chr.X: 119590723-119590723 on Build GRCh38
rs758699206 Chr.X: 119590978-119590978 on Build GRCh38
rs757268163 Chr.X: 119590642-119590642 on Build GRCh38
rs150729122 Chr.X: 119590715-119590715 on Build GRCh38
rs751261923 Chr.X: 119590614-119590614 on Build GRCh38

View Full Product Details