Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186797348-186797869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC4
Entrez Gene ID
5984
Gene Name
replication factor C subunit 4
Gene Aliases
A1, RFC37
UniGene
Hs.732098
Gene Chromosome Location
Chr.3: 186789892-186806695 on Build GRCh38
Transcripts
NM_002916.3 NM_181573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6219199 c.231C>T p.Y77Y Chr.3: 186797594-186797594 on Build GRCh38
277048 c.220C>A p.L74I Chr.3: 186797605-186797605 on Build GRCh38
6206065 c.231_232delCG p.Y77fs*1 Chr.3: 186797593-186797594 on Build GRCh38
6097379 c.270A>T p.A90A Chr.3: 186797555-186797555 on Build GRCh38
3591048 c.255A>T p.T85T Chr.3: 186797570-186797570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777723840 Chr.3: 186797558-186797558 on Build GRCh38
rs552067806 Chr.3: 186797444-186797444 on Build GRCh38
rs11544065 Chr.3: 186797564-186797564 on Build GRCh38
rs113938310 Chr.3: 186797554-186797554 on Build GRCh38
rs1047976555 Chr.3: 186797645-186797645 on Build GRCh38
rs559639253 Chr.3: 186797409-186797409 on Build GRCh38
rs966962994 Chr.3: 186797802-186797802 on Build GRCh38
rs905049567 Chr.3: 186797474-186797474 on Build GRCh38
rs1002341772 Chr.3: 186797726-186797726 on Build GRCh38
rs962711998 Chr.3: 186797372-186797372 on Build GRCh38
rs577345862 Chr.3: 186797371-186797371 on Build GRCh38
rs1006367084 Chr.3: 186797672-186797672 on Build GRCh38
rs183013523 Chr.3: 186797734-186797734 on Build GRCh38
rs899226019 Chr.3: 186797642-186797642 on Build GRCh38
rs370686394 Chr.3: 186797492-186797492 on Build GRCh38
rs755867523 Chr.3: 186797556-186797556 on Build GRCh38
rs1019155455 Chr.3: 186797711-186797711 on Build GRCh38
rs541394891 Chr.3: 186797402-186797402 on Build GRCh38
rs772790455 Chr.3: 186797449-186797449 on Build GRCh38
rs28903097 Chr.3: 186797567-186797567 on Build GRCh38
rs190308216 Chr.3: 186797829-186797829 on Build GRCh38
rs745531580 Chr.3: 186797502-186797503 on Build GRCh38
rs187349496 Chr.3: 186797772-186797772 on Build GRCh38
rs967771303 Chr.3: 186797808-186797808 on Build GRCh38
rs911541762 Chr.3: 186797433-186797433 on Build GRCh38
rs988128379 Chr.3: 186797701-186797701 on Build GRCh38
rs914930477 Chr.3: 186797807-186797807 on Build GRCh38
rs538407276 Chr.3: 186797443-186797443 on Build GRCh38
rs3917119 Chr.3: 186797681-186797681 on Build GRCh38
rs758462674 Chr.3: 186797587-186797587 on Build GRCh38
rs898271803 Chr.3: 186797708-186797708 on Build GRCh38
rs374776936 Chr.3: 186797590-186797590 on Build GRCh38
rs570395824 Chr.3: 186797448-186797448 on Build GRCh38
rs748553683 Chr.3: 186797618-186797618 on Build GRCh38
rs1001849892 Chr.3: 186797397-186797397 on Build GRCh38
rs537627242 Chr.3: 186797761-186797761 on Build GRCh38
rs767589112 Chr.3: 186797766-186797766 on Build GRCh38
rs201838412 Chr.3: 186797612-186797612 on Build GRCh38
rs1029105450 Chr.3: 186797661-186797661 on Build GRCh38
rs796867703 Chr.3: 186797476-186797476 on Build GRCh38
rs759194875 Chr.3: 186797530-186797530 on Build GRCh38
rs781515743 Chr.3: 186797613-186797613 on Build GRCh38
rs770261172 Chr.3: 186797637-186797637 on Build GRCh38
rs56150850 Chr.3: 186797436-186797436 on Build GRCh38
rs563500070 Chr.3: 186797442-186797442 on Build GRCh38
rs577609423 Chr.3: 186797824-186797824 on Build GRCh38
rs1033345750 Chr.3: 186797733-186797733 on Build GRCh38
rs767101951 Chr.3: 186797531-186797531 on Build GRCh38
rs966533011 Chr.3: 186797466-186797466 on Build GRCh38
rs979258392 Chr.3: 186797809-186797809 on Build GRCh38
rs528100284 Chr.3: 186797520-186797520 on Build GRCh38
rs530201315 Chr.3: 186797429-186797429 on Build GRCh38
rs1044896118 Chr.3: 186797464-186797464 on Build GRCh38
rs746993524 Chr.3: 186797594-186797594 on Build GRCh38
rs55892828 Chr.3: 186797680-186797680 on Build GRCh38
rs987797539 Chr.3: 186797805-186797805 on Build GRCh38
rs200551222 Chr.3: 186797652-186797652 on Build GRCh38
rs979925524 Chr.3: 186797469-186797469 on Build GRCh38
rs1013461615 Chr.3: 186797769-186797769 on Build GRCh38
rs370512988 Chr.3: 186797738-186797738 on Build GRCh38
rs762749900 Chr.3: 186797482-186797482 on Build GRCh38

View Full Product Details