Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 133311057-133311574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPC4
Entrez Gene ID
2239
Gene Name
glypican 4
Gene Aliases
K-glypican
UniGene
Hs.58367
Gene Chromosome Location
Chr.X: 133301036-133415177 on Build GRCh38
Transcripts
NM_001448.2

SNP Detail

SNP ID Chromosome Location
rs753598118 Chr.X: 133311494-133311494 on Build GRCh38
rs748839724 Chr.X: 133311264-133311264 on Build GRCh38
rs369641443 Chr.X: 133311222-133311222 on Build GRCh38
rs748709159 Chr.X: 133311282-133311282 on Build GRCh38
rs774268921 Chr.X: 133311554-133311554 on Build GRCh38
rs776789656 Chr.X: 133311346-133311346 on Build GRCh38
rs144650030 Chr.X: 133311318-133311318 on Build GRCh38
rs778273528 Chr.X: 133311531-133311531 on Build GRCh38
rs997760199 Chr.X: 133311085-133311085 on Build GRCh38
rs756163452 Chr.X: 133311334-133311334 on Build GRCh38
rs780257736 Chr.X: 133311401-133311401 on Build GRCh38
rs768525193 Chr.X: 133311499-133311499 on Build GRCh38
rs755737172 Chr.X: 133311241-133311241 on Build GRCh38
rs984142694 Chr.X: 133311118-133311118 on Build GRCh38
rs980660085 Chr.X: 133311468-133311468 on Build GRCh38
rs1007207927 Chr.X: 133311179-133311179 on Build GRCh38
rs778547920 Chr.X: 133311279-133311279 on Build GRCh38
rs771753995 Chr.X: 133311359-133311359 on Build GRCh38
rs1008972171 Chr.X: 133311472-133311472 on Build GRCh38
rs761804365 Chr.X: 133311428-133311428 on Build GRCh38
rs749506559 Chr.X: 133311457-133311457 on Build GRCh38
rs750191791 Chr.X: 133311440-133311440 on Build GRCh38
rs199910189 Chr.X: 133311363-133311363 on Build GRCh38
rs372298799 Chr.X: 133311239-133311239 on Build GRCh38
rs771274300 Chr.X: 133311534-133311534 on Build GRCh38
rs192130195 Chr.X: 133311164-133311164 on Build GRCh38
rs866960169 Chr.X: 133311371-133311371 on Build GRCh38
rs747361365 Chr.X: 133311533-133311533 on Build GRCh38
rs962250589 Chr.X: 133311538-133311538 on Build GRCh38
rs778934042 Chr.X: 133311124-133311124 on Build GRCh38
rs767475124 Chr.X: 133311439-133311439 on Build GRCh38
rs951545662 Chr.X: 133311349-133311349 on Build GRCh38
rs757164941 Chr.X: 133311209-133311209 on Build GRCh38
rs938963596 Chr.X: 133311480-133311480 on Build GRCh38
rs889043806 Chr.X: 133311166-133311166 on Build GRCh38
rs746128607 Chr.X: 133311544-133311544 on Build GRCh38
rs926538604 Chr.X: 133311470-133311470 on Build GRCh38
rs767772904 Chr.X: 133311469-133311470 on Build GRCh38
rs778635705 Chr.X: 133311506-133311506 on Build GRCh38
rs1011086071 Chr.X: 133311355-133311355 on Build GRCh38
rs757739421 Chr.X: 133311259-133311259 on Build GRCh38
rs1015919853 Chr.X: 133311188-133311188 on Build GRCh38
rs1052589280 Chr.X: 133311327-133311327 on Build GRCh38
rs758839911 Chr.X: 133311508-133311508 on Build GRCh38
rs747650409 Chr.X: 133311507-133311507 on Build GRCh38
rs958930784 Chr.X: 133311430-133311430 on Build GRCh38
rs991732999 Chr.X: 133311438-133311438 on Build GRCh38
rs373402688 Chr.X: 133311400-133311400 on Build GRCh38
rs776957622 Chr.X: 133311537-133311537 on Build GRCh38
rs781714136 Chr.X: 133311291-133311291 on Build GRCh38
rs928201352 Chr.X: 133311288-133311288 on Build GRCh38
rs773510836 Chr.X: 133311315-133311315 on Build GRCh38
rs1030212618 Chr.X: 133311132-133311132 on Build GRCh38
rs768233396 Chr.X: 133311270-133311270 on Build GRCh38

View Full Product Details