Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 72377330-72377861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RYBP
Entrez Gene ID
23429
Gene Name
RING1 and YY1 binding protein
Gene Aliases
AAP1, APAP-1, DEDAF, YEAF1
UniGene
Hs.7910
Gene Chromosome Location
Chr.3: 72374593-72446623 on Build GRCh38
Transcripts
NM_012234.6

SNP Detail

SNP ID Chromosome Location
rs772720422 Chr.3: 72377622-72377622 on Build GRCh38
rs192441828 Chr.3: 72377513-72377513 on Build GRCh38
rs948603557 Chr.3: 72377636-72377636 on Build GRCh38
rs535028701 Chr.3: 72377660-72377660 on Build GRCh38
rs919362144 Chr.3: 72377579-72377579 on Build GRCh38
rs77987705 Chr.3: 72377524-72377524 on Build GRCh38
rs917169589 Chr.3: 72377627-72377627 on Build GRCh38
rs549871272 Chr.3: 72377808-72377808 on Build GRCh38
rs892611199 Chr.3: 72377479-72377479 on Build GRCh38
rs1031396884 Chr.3: 72377466-72377466 on Build GRCh38
rs992928455 Chr.3: 72377795-72377795 on Build GRCh38
rs1044326785 Chr.3: 72377637-72377637 on Build GRCh38
rs374778501 Chr.3: 72377617-72377617 on Build GRCh38
rs948404651 Chr.3: 72377427-72377427 on Build GRCh38
rs1017985526 Chr.3: 72377542-72377542 on Build GRCh38
rs1026392737 Chr.3: 72377757-72377757 on Build GRCh38
rs760565823 Chr.3: 72377679-72377679 on Build GRCh38
rs548691056 Chr.3: 72377732-72377732 on Build GRCh38
rs986147494 Chr.3: 72377536-72377536 on Build GRCh38
rs4677159 Chr.3: 72377426-72377426 on Build GRCh38
rs182456741 Chr.3: 72377672-72377672 on Build GRCh38
rs566604528 Chr.3: 72377530-72377530 on Build GRCh38
rs551219499 Chr.3: 72377552-72377552 on Build GRCh38
rs889430228 Chr.3: 72377483-72377483 on Build GRCh38
rs1008193323 Chr.3: 72377696-72377696 on Build GRCh38
rs1020279804 Chr.3: 72377662-72377663 on Build GRCh38
rs542661215 Chr.3: 72377360-72377360 on Build GRCh38
rs992779186 Chr.3: 72377658-72377658 on Build GRCh38
rs904168289 Chr.3: 72377644-72377644 on Build GRCh38
rs531283100 Chr.3: 72377794-72377794 on Build GRCh38
rs764408375 Chr.3: 72377821-72377821 on Build GRCh38
rs929140402 Chr.3: 72377377-72377377 on Build GRCh38
rs753457950 Chr.3: 72377671-72377671 on Build GRCh38
rs985719925 Chr.3: 72377701-72377701 on Build GRCh38
rs1044493861 Chr.3: 72377437-72377437 on Build GRCh38
rs543931089 Chr.3: 72377512-72377512 on Build GRCh38
rs760474200 Chr.3: 72377448-72377448 on Build GRCh38
rs1009731832 Chr.3: 72377675-72377675 on Build GRCh38
rs763847872 Chr.3: 72377543-72377543 on Build GRCh38
rs1017886140 Chr.3: 72377715-72377715 on Build GRCh38
rs368965567 Chr.3: 72377359-72377360 on Build GRCh38
rs963353545 Chr.3: 72377747-72377747 on Build GRCh38
rs1034458787 Chr.3: 72377649-72377649 on Build GRCh38
rs925078485 Chr.3: 72377688-72377688 on Build GRCh38
rs186173293 Chr.3: 72377451-72377451 on Build GRCh38
rs929109424 Chr.3: 72377607-72377607 on Build GRCh38
rs1057027916 Chr.3: 72377391-72377391 on Build GRCh38
rs4677160 Chr.3: 72377750-72377750 on Build GRCh38
rs576452046 Chr.3: 72377432-72377432 on Build GRCh38
rs764715216 Chr.3: 72377683-72377683 on Build GRCh38
rs950901897 Chr.3: 72377785-72377785 on Build GRCh38
rs999868028 Chr.3: 72377464-72377464 on Build GRCh38
rs936383764 Chr.3: 72377462-72377462 on Build GRCh38
rs925143270 Chr.3: 72377435-72377435 on Build GRCh38
rs973458896 Chr.3: 72377559-72377559 on Build GRCh38
rs936535337 Chr.3: 72377691-72377691 on Build GRCh38

View Full Product Details