Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 106194546-106195108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBCK
Entrez Gene ID
93627
Gene Name
TBC1 domain containing kinase
Gene Aliases
HSPC302, IHPRF3, TBCKL
UniGene
Hs.151498
Gene Chromosome Location
Chr.4: 106044317-106316704 on Build GRCh38
Transcripts
NM_001163435.2 NM_001163436.2 NM_001163437.2 NM_001290768.1 NM_033115.4 XM_011532417.2 XM_017008846.1 XM_017008847.1 XM_017008848.1 XM_017008849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1426255 c.1897+1G>A p.? Chr.4: 106194717-106194717 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs552521682 Chr.4: 106194698-106194698 on Build GRCh38
rs751559713 Chr.4: 106194709-106194709 on Build GRCh38
rs1034198791 Chr.4: 106194748-106194748 on Build GRCh38
rs557549969 Chr.4: 106194973-106194973 on Build GRCh38
rs969723923 Chr.4: 106194949-106194949 on Build GRCh38
rs753409452 Chr.4: 106194688-106194688 on Build GRCh38
rs771383864 Chr.4: 106194750-106194750 on Build GRCh38
rs762814122 Chr.4: 106194794-106194794 on Build GRCh38
rs982436877 Chr.4: 106194890-106194890 on Build GRCh38
rs1024609098 Chr.4: 106194820-106194820 on Build GRCh38
rs979773583 Chr.4: 106194952-106194952 on Build GRCh38
rs574740376 Chr.4: 106194718-106194718 on Build GRCh38
rs566797229 Chr.4: 106195046-106195046 on Build GRCh38
rs975546107 Chr.4: 106195040-106195040 on Build GRCh38
rs756854971 Chr.4: 106194664-106194664 on Build GRCh38
rs186748985 Chr.4: 106194585-106194585 on Build GRCh38
rs1002274809 Chr.4: 106194743-106194743 on Build GRCh38
rs951298672 Chr.4: 106194839-106194839 on Build GRCh38
rs921405197 Chr.4: 106194663-106194663 on Build GRCh38
rs962449738 Chr.4: 106194939-106194939 on Build GRCh38
rs1051747947 Chr.4: 106194699-106194699 on Build GRCh38
rs559351051 Chr.4: 106194761-106194761 on Build GRCh38
rs12506062 Chr.4: 106194803-106194803 on Build GRCh38
rs963672941 Chr.4: 106194993-106194993 on Build GRCh38
rs760133780 Chr.4: 106194682-106194682 on Build GRCh38
rs149743265 Chr.4: 106194660-106194660 on Build GRCh38
rs768038053 Chr.4: 106194771-106194771 on Build GRCh38
rs191226474 Chr.4: 106194885-106194885 on Build GRCh38
rs540625784 Chr.4: 106194719-106194719 on Build GRCh38
rs1031517568 Chr.4: 106194826-106194826 on Build GRCh38
rs568776740 Chr.4: 106194652-106194652 on Build GRCh38
rs909515770 Chr.4: 106194985-106194985 on Build GRCh38
rs140488279 Chr.4: 106194892-106194892 on Build GRCh38
rs747305655 Chr.4: 106194944-106194944 on Build GRCh38
rs775013614 Chr.4: 106194668-106194668 on Build GRCh38
rs1007553360 Chr.4: 106194762-106194762 on Build GRCh38
rs779889157 Chr.4: 106194706-106194706 on Build GRCh38
rs753349180 Chr.4: 106194627-106194627 on Build GRCh38
rs778376635 Chr.4: 106194759-106194759 on Build GRCh38
rs763698674 Chr.4: 106194685-106194685 on Build GRCh38
rs951012483 Chr.4: 106194630-106194630 on Build GRCh38
rs761615755 Chr.4: 106194692-106194692 on Build GRCh38
rs375645878 Chr.4: 106194852-106194852 on Build GRCh38
rs753701206 Chr.4: 106194579-106194579 on Build GRCh38
rs551728504 Chr.4: 106194790-106194790 on Build GRCh38
rs368601581 Chr.4: 106194636-106194636 on Build GRCh38
rs530763580 Chr.4: 106194884-106194884 on Build GRCh38
rs770605372 Chr.4: 106195026-106195026 on Build GRCh38
rs146770919 Chr.4: 106194700-106194700 on Build GRCh38
rs765004275 Chr.4: 106194780-106194780 on Build GRCh38
rs374319146 Chr.4: 106194717-106194717 on Build GRCh38
rs528415950 Chr.4: 106194772-106194772 on Build GRCh38
rs749673214 Chr.4: 106194742-106194742 on Build GRCh38
rs781395512 Chr.4: 106194714-106194714 on Build GRCh38
rs909674889 Chr.4: 106194638-106194638 on Build GRCh38
rs375424316 Chr.4: 106194757-106194757 on Build GRCh38
rs546592326 Chr.4: 106194984-106194984 on Build GRCh38
rs955839657 Chr.4: 106194880-106194880 on Build GRCh38
rs778049682 Chr.4: 106194738-106194738 on Build GRCh38
rs776101351 Chr.4: 106194799-106194799 on Build GRCh38
rs768363753 Chr.4: 106194770-106194770 on Build GRCh38
rs992540792 Chr.4: 106194887-106194887 on Build GRCh38
rs371418828 Chr.4: 106194702-106194702 on Build GRCh38
rs760324508 Chr.4: 106194737-106194737 on Build GRCh38
rs754993262 Chr.4: 106194710-106194710 on Build GRCh38
rs974028051 Chr.4: 106195042-106195042 on Build GRCh38
rs745338283 Chr.4: 106194778-106194778 on Build GRCh38
rs941097179 Chr.4: 106194656-106194656 on Build GRCh38

View Full Product Details