Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69171194-69171745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB1
Entrez Gene ID
891
Gene Name
cyclin B1
Gene Aliases
CCNB
UniGene
Hs.23960
Gene Chromosome Location
Chr.5: 69167010-69178245 on Build GRCh38
Transcripts
NM_031966.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1438366 c.371C>G p.T124S Chr.5: 69171277-69171277 on Build GRCh38
7138847 c.534G>A p.L178L Chr.5: 69171440-69171440 on Build GRCh38
6448439 c.384C>T p.S128S Chr.5: 69171290-69171290 on Build GRCh38
1438367 c.449T>C p.I150T Chr.5: 69171355-69171355 on Build GRCh38
6563073 c.364-4C>A p.? Chr.5: 69171266-69171266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896594579 Chr.5: 69171279-69171279 on Build GRCh38
rs868016307 Chr.5: 69171272-69171272 on Build GRCh38
rs2069433 Chr.5: 69171680-69171680 on Build GRCh38
rs968378610 Chr.5: 69171509-69171509 on Build GRCh38
rs764386519 Chr.5: 69171465-69171465 on Build GRCh38
rs761712838 Chr.5: 69171269-69171269 on Build GRCh38
rs934514238 Chr.5: 69171236-69171236 on Build GRCh38
rs1016484804 Chr.5: 69171425-69171425 on Build GRCh38
rs78743729 Chr.5: 69171388-69171388 on Build GRCh38
rs770868247 Chr.5: 69171494-69171495 on Build GRCh38
rs113314557 Chr.5: 69171468-69171468 on Build GRCh38
rs760206291 Chr.5: 69171708-69171708 on Build GRCh38
rs764774100 Chr.5: 69171417-69171417 on Build GRCh38
rs993698379 Chr.5: 69171648-69171648 on Build GRCh38
rs573688422 Chr.5: 69171681-69171681 on Build GRCh38
rs757991804 Chr.5: 69171462-69171462 on Build GRCh38
rs1013661802 Chr.5: 69171280-69171280 on Build GRCh38
rs976922435 Chr.5: 69171515-69171515 on Build GRCh38
rs1016222787 Chr.5: 69171721-69171721 on Build GRCh38
rs758845734 Chr.5: 69171315-69171315 on Build GRCh38
rs778143464 Chr.5: 69171326-69171326 on Build GRCh38
rs762887344 Chr.5: 69171469-69171470 on Build GRCh38
rs905201698 Chr.5: 69171598-69171598 on Build GRCh38
rs371830251 Chr.5: 69171415-69171415 on Build GRCh38
rs924257399 Chr.5: 69171699-69171699 on Build GRCh38
rs773250472 Chr.5: 69171469-69171469 on Build GRCh38
rs781237721 Chr.5: 69171361-69171361 on Build GRCh38
rs1002622565 Chr.5: 69171651-69171651 on Build GRCh38
rs752344882 Chr.5: 69171435-69171435 on Build GRCh38
rs921329211 Chr.5: 69171532-69171532 on Build GRCh38
rs368715958 Chr.5: 69171233-69171233 on Build GRCh38
rs778292780 Chr.5: 69171227-69171227 on Build GRCh38
rs769916796 Chr.5: 69171461-69171462 on Build GRCh38
rs369595226 Chr.5: 69171466-69171466 on Build GRCh38
rs756173499 Chr.5: 69171477-69171477 on Build GRCh38
rs146028282 Chr.5: 69171398-69171398 on Build GRCh38
rs1012644639 Chr.5: 69171672-69171672 on Build GRCh38
rs765917246 Chr.5: 69171293-69171293 on Build GRCh38
rs180760005 Chr.5: 69171294-69171294 on Build GRCh38
rs746029551 Chr.5: 69171375-69171375 on Build GRCh38
rs1038105762 Chr.5: 69171641-69171641 on Build GRCh38
rs1007729086 Chr.5: 69171666-69171666 on Build GRCh38
rs543622757 Chr.5: 69171632-69171632 on Build GRCh38
rs748224599 Chr.5: 69171320-69171322 on Build GRCh38
rs934543969 Chr.5: 69171562-69171562 on Build GRCh38
rs759125094 Chr.5: 69171416-69171416 on Build GRCh38
rs746020758 Chr.5: 69171256-69171256 on Build GRCh38
rs754571496 Chr.5: 69171297-69171297 on Build GRCh38
rs747995329 Chr.5: 69171436-69171436 on Build GRCh38
rs1030469933 Chr.5: 69171654-69171654 on Build GRCh38
rs35708754 Chr.5: 69171232-69171233 on Build GRCh38
rs1006257397 Chr.5: 69171623-69171623 on Build GRCh38
rs747537060 Chr.5: 69171228-69171228 on Build GRCh38
rs978472900 Chr.5: 69171688-69171688 on Build GRCh38
rs774015617 Chr.5: 69171263-69171263 on Build GRCh38
rs983479696 Chr.5: 69171348-69171348 on Build GRCh38
rs769878758 Chr.5: 69171377-69171377 on Build GRCh38
rs150242295 Chr.5: 69171283-69171283 on Build GRCh38
rs766846332 Chr.5: 69171644-69171644 on Build GRCh38
rs1030438884 Chr.5: 69171437-69171437 on Build GRCh38
rs747450397 Chr.5: 69171339-69171339 on Build GRCh38
rs958450831 Chr.5: 69171223-69171223 on Build GRCh38
rs373027578 Chr.5: 69171581-69171581 on Build GRCh38
rs771949435 Chr.5: 69171395-69171395 on Build GRCh38
rs767338588 Chr.5: 69171277-69171277 on Build GRCh38
rs757776462 Chr.5: 69171351-69171351 on Build GRCh38
rs546247897 Chr.5: 69171516-69171516 on Build GRCh38
rs373737881 Chr.5: 69171469-69171469 on Build GRCh38
rs541771012 Chr.5: 69171346-69171346 on Build GRCh38
rs917056494 Chr.5: 69171583-69171583 on Build GRCh38
rs978824676 Chr.5: 69171522-69171522 on Build GRCh38
rs924588777 Chr.5: 69171545-69171545 on Build GRCh38
rs540721481 Chr.5: 69171716-69171716 on Build GRCh38
rs968869121 Chr.5: 69171687-69171687 on Build GRCh38
rs2069432 Chr.5: 69171585-69171585 on Build GRCh38
rs766479931 Chr.5: 69171412-69171412 on Build GRCh38
rs962263214 Chr.5: 69171427-69171427 on Build GRCh38
rs374353420 Chr.5: 69171286-69171286 on Build GRCh38
rs760630040 Chr.5: 69171409-69171409 on Build GRCh38
rs776697767 Chr.5: 69171242-69171242 on Build GRCh38
rs954836794 Chr.5: 69171661-69171661 on Build GRCh38
rs770115345 Chr.5: 69171259-69171259 on Build GRCh38
rs765005697 Chr.5: 69171307-69171307 on Build GRCh38
rs773141929 Chr.5: 69171401-69171401 on Build GRCh38
rs572008792 Chr.5: 69171594-69171594 on Build GRCh38
rs138977567 Chr.5: 69171526-69171526 on Build GRCh38
rs752972082 Chr.5: 69171312-69171312 on Build GRCh38
rs1045118651 Chr.5: 69171597-69171597 on Build GRCh38
rs1056912435 Chr.5: 69171653-69171653 on Build GRCh38
rs200296214 Chr.5: 69171285-69171285 on Build GRCh38
rs775834220 Chr.5: 69171386-69171386 on Build GRCh38

View Full Product Details