Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 39371522-39372099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAB2
Entrez Gene ID
1601
Gene Name
DAB2, clathrin adaptor protein
Gene Aliases
DOC-2, DOC2
UniGene
Hs.696631
Gene Chromosome Location
Chr.5: 39371674-39425233 on Build GRCh38
Transcripts
NM_001244871.1 NM_001343.3

SNP Detail

SNP ID Chromosome Location
rs959291923 Chr.5: 39371970-39371970 on Build GRCh38
rs2542708 Chr.5: 39371809-39371809 on Build GRCh38
rs769181762 Chr.5: 39371782-39371782 on Build GRCh38
rs371411060 Chr.5: 39372001-39372001 on Build GRCh38
rs909603037 Chr.5: 39371791-39371791 on Build GRCh38
rs1034921478 Chr.5: 39371661-39371661 on Build GRCh38
rs1016742975 Chr.5: 39371994-39371994 on Build GRCh38
rs566752098 Chr.5: 39371903-39371903 on Build GRCh38
rs2738242 Chr.5: 39371760-39371760 on Build GRCh38
rs1026179224 Chr.5: 39371990-39371990 on Build GRCh38
rs547596229 Chr.5: 39371740-39371740 on Build GRCh38
rs2584246 Chr.5: 39371997-39371997 on Build GRCh38
rs933700099 Chr.5: 39371843-39371843 on Build GRCh38
rs1046033 Chr.5: 39372048-39372048 on Build GRCh38
rs1034895124 Chr.5: 39371965-39371965 on Build GRCh38
rs547523654 Chr.5: 39371868-39371868 on Build GRCh38
rs911500390 Chr.5: 39371705-39371705 on Build GRCh38
rs17470667 Chr.5: 39371973-39371973 on Build GRCh38
rs938371282 Chr.5: 39371813-39371813 on Build GRCh38
rs144179892 Chr.5: 39371912-39371912 on Build GRCh38
rs762546962 Chr.5: 39371855-39371855 on Build GRCh38
rs914727258 Chr.5: 39371734-39371734 on Build GRCh38
rs942929108 Chr.5: 39371717-39371717 on Build GRCh38
rs2738241 Chr.5: 39371818-39371818 on Build GRCh38
rs531962537 Chr.5: 39371604-39371604 on Build GRCh38
rs772741786 Chr.5: 39371852-39371852 on Build GRCh38
rs14454 Chr.5: 39371766-39371766 on Build GRCh38
rs541798592 Chr.5: 39372068-39372068 on Build GRCh38
rs1021389788 Chr.5: 39371554-39371554 on Build GRCh38
rs990368946 Chr.5: 39371719-39371719 on Build GRCh38
rs555793208 Chr.5: 39372033-39372033 on Build GRCh38
rs2738240 Chr.5: 39371830-39371830 on Build GRCh38
rs990650018 Chr.5: 39371750-39371750 on Build GRCh38
rs529260595 Chr.5: 39371849-39371849 on Build GRCh38
rs918367072 Chr.5: 39372058-39372058 on Build GRCh38
rs962781273 Chr.5: 39371793-39371793 on Build GRCh38
rs761171524 Chr.5: 39372007-39372007 on Build GRCh38
rs893870923 Chr.5: 39371900-39371900 on Build GRCh38
rs897674709 Chr.5: 39371965-39371965 on Build GRCh38
rs1003588228 Chr.5: 39371922-39371922 on Build GRCh38
rs2542709 Chr.5: 39371884-39371884 on Build GRCh38
rs568575148 Chr.5: 39371801-39371801 on Build GRCh38
rs1039221883 Chr.5: 39371869-39371869 on Build GRCh38
rs1003854305 Chr.5: 39371651-39371651 on Build GRCh38
rs918261516 Chr.5: 39371821-39371821 on Build GRCh38
rs56362760 Chr.5: 39371703-39371703 on Build GRCh38
rs558696364 Chr.5: 39371942-39371942 on Build GRCh38
rs867576466 Chr.5: 39371681-39371681 on Build GRCh38
rs770601849 Chr.5: 39372060-39372060 on Build GRCh38
rs1042504871 Chr.5: 39371909-39371909 on Build GRCh38
rs769335295 Chr.5: 39371722-39371722 on Build GRCh38
rs115305120 Chr.5: 39371581-39371581 on Build GRCh38
rs943483306 Chr.5: 39371863-39371863 on Build GRCh38

View Full Product Details