Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138291827-138292321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC25C
Entrez Gene ID
995
Gene Name
cell division cycle 25C
Gene Aliases
CDC25, PPP1R60
UniGene
Hs.656
Gene Chromosome Location
Chr.5: 138285265-138338355 on Build GRCh38
Transcripts
NM_001287582.1 NM_001287583.1 NM_001318098.1 NM_001790.4 NM_022809.3 XM_005272145.3 XM_006714739.3 XM_011543759.2 XM_011543760.2 XM_011543761.2 XM_011543762.1 XM_011543763.1 XM_011543764.1 XM_017010101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
171549 c.657G>T p.E219D Chr.5: 138292075-138292075 on Build GRCh38
6469957 c.740C>T p.S247F Chr.5: 138291992-138291992 on Build GRCh38
3237197 c.731delA p.K244fs*15 Chr.5: 138292001-138292001 on Build GRCh38
5897516 c.637C>T p.R213C Chr.5: 138292095-138292095 on Build GRCh38
6169097 c.644C>A p.P215Q Chr.5: 138292088-138292088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11567989 Chr.5: 138291962-138291962 on Build GRCh38
rs386692799 Chr.5: 138291934-138291936 on Build GRCh38
rs768102789 Chr.5: 138292165-138292165 on Build GRCh38
rs759174257 Chr.5: 138292155-138292155 on Build GRCh38
rs929265474 Chr.5: 138292063-138292063 on Build GRCh38
rs866193069 Chr.5: 138292033-138292033 on Build GRCh38
rs776173691 Chr.5: 138292140-138292140 on Build GRCh38
rs145511523 Chr.5: 138292061-138292061 on Build GRCh38
rs11567991 Chr.5: 138291934-138291934 on Build GRCh38
rs374941759 Chr.5: 138292036-138292036 on Build GRCh38
rs781599523 Chr.5: 138291931-138291931 on Build GRCh38
rs771321731 Chr.5: 138291927-138291927 on Build GRCh38
rs756396023 Chr.5: 138292103-138292103 on Build GRCh38
rs182008758 Chr.5: 138292087-138292087 on Build GRCh38
rs560587670 Chr.5: 138292273-138292273 on Build GRCh38
rs184628552 Chr.5: 138291855-138291855 on Build GRCh38
rs780168277 Chr.5: 138292109-138292109 on Build GRCh38
rs148887677 Chr.5: 138292094-138292094 on Build GRCh38
rs761679863 Chr.5: 138292074-138292074 on Build GRCh38
rs767249314 Chr.5: 138291969-138291969 on Build GRCh38
rs867464017 Chr.5: 138291908-138291908 on Build GRCh38
rs760253163 Chr.5: 138291998-138291998 on Build GRCh38
rs772988065 Chr.5: 138292085-138292085 on Build GRCh38
rs759001272 Chr.5: 138292090-138292090 on Build GRCh38
rs202224563 Chr.5: 138292100-138292100 on Build GRCh38
rs933669811 Chr.5: 138292026-138292026 on Build GRCh38
rs114224600 Chr.5: 138292095-138292095 on Build GRCh38
rs748150102 Chr.5: 138292131-138292131 on Build GRCh38
rs759829429 Chr.5: 138292000-138292000 on Build GRCh38
rs778113693 Chr.5: 138292017-138292017 on Build GRCh38
rs965880642 Chr.5: 138292194-138292194 on Build GRCh38
rs200825916 Chr.5: 138292072-138292072 on Build GRCh38
rs71596567 Chr.5: 138291941-138291941 on Build GRCh38
rs764662095 Chr.5: 138292159-138292159 on Build GRCh38
rs943530849 Chr.5: 138292211-138292211 on Build GRCh38
rs752076059 Chr.5: 138292025-138292025 on Build GRCh38
rs556465778 Chr.5: 138292008-138292008 on Build GRCh38
rs750802175 Chr.5: 138292271-138292271 on Build GRCh38
rs533244388 Chr.5: 138291918-138291918 on Build GRCh38
rs1025196214 Chr.5: 138291961-138291961 on Build GRCh38
rs1046133488 Chr.5: 138292110-138292110 on Build GRCh38
rs929106923 Chr.5: 138291880-138291880 on Build GRCh38
rs770722370 Chr.5: 138292139-138292139 on Build GRCh38
rs768047514 Chr.5: 138292097-138292097 on Build GRCh38
rs750088334 Chr.5: 138291984-138291984 on Build GRCh38
rs189309721 Chr.5: 138291965-138291965 on Build GRCh38
rs746389185 Chr.5: 138291864-138291864 on Build GRCh38
rs775927964 Chr.5: 138292001-138292001 on Build GRCh38
rs1047587657 Chr.5: 138291910-138291910 on Build GRCh38
rs765926396 Chr.5: 138292088-138292088 on Build GRCh38
rs149326899 Chr.5: 138292275-138292275 on Build GRCh38
rs762455824 Chr.5: 138292163-138292163 on Build GRCh38
rs189899115 Chr.5: 138291942-138291942 on Build GRCh38
rs765981873 Chr.5: 138292002-138292002 on Build GRCh38
rs116617843 Chr.5: 138292084-138292084 on Build GRCh38
rs762801188 Chr.5: 138292032-138292032 on Build GRCh38
rs747314775 Chr.5: 138291922-138291922 on Build GRCh38
rs180773150 Chr.5: 138291955-138291955 on Build GRCh38
rs755025829 Chr.5: 138292122-138292122 on Build GRCh38
rs746155525 Chr.5: 138292034-138292034 on Build GRCh38
rs187425038 Chr.5: 138292135-138292135 on Build GRCh38
rs11567990 Chr.5: 138291936-138291936 on Build GRCh38
rs138086428 Chr.5: 138292117-138292117 on Build GRCh38
rs1018327460 Chr.5: 138292178-138292180 on Build GRCh38
rs963208355 Chr.5: 138292285-138292285 on Build GRCh38
rs548954692 Chr.5: 138292301-138292301 on Build GRCh38
rs11567988 Chr.5: 138292138-138292138 on Build GRCh38
rs1050808052 Chr.5: 138292027-138292027 on Build GRCh38
rs757723414 Chr.5: 138292031-138292031 on Build GRCh38
rs199830488 Chr.5: 138292056-138292056 on Build GRCh38
rs914786807 Chr.5: 138292181-138292181 on Build GRCh38
rs553796635 Chr.5: 138292069-138292069 on Build GRCh38
rs1032611147 Chr.5: 138292149-138292149 on Build GRCh38
rs763448312 Chr.5: 138292116-138292116 on Build GRCh38
rs764422233 Chr.5: 138291967-138291967 on Build GRCh38
rs1051281762 Chr.5: 138292282-138292282 on Build GRCh38
rs778984587 Chr.5: 138292123-138292123 on Build GRCh38
rs149693926 Chr.5: 138292029-138292029 on Build GRCh38
rs772973906 Chr.5: 138292001-138292001 on Build GRCh38
rs974884329 Chr.5: 138292227-138292227 on Build GRCh38
rs765464033 Chr.5: 138292000-138292001 on Build GRCh38
rs886818854 Chr.5: 138292290-138292290 on Build GRCh38

View Full Product Details