Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42721632-42722230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPH2
Entrez Gene ID
5961
Gene Name
peripherin 2
Gene Aliases
AOFMD, AVMD, CACD2, DS, MDBS1, PRPH, RDS, RP7, TSPAN22, rd2
UniGene
Hs.654489
Gene Chromosome Location
Chr.6: 42694509-42723024 on Build GRCh38
Transcripts
NM_000322.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4718043 c.455T>C p.M152T Chr.6: 42721880-42721880 on Build GRCh38
4533511 c.198G>A p.G66G Chr.6: 42722137-42722137 on Build GRCh38
6471217 c.555G>A p.L185L Chr.6: 42721780-42721780 on Build GRCh38
4464952 c.135C>T p.L45L Chr.6: 42722200-42722200 on Build GRCh38
188503 c.367C>T p.R123W Chr.6: 42721968-42721968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781631103 Chr.6: 42722014-42722014 on Build GRCh38
rs772861671 Chr.6: 42721961-42721961 on Build GRCh38
rs61755796 Chr.6: 42721800-42721800 on Build GRCh38
rs375823961 Chr.6: 42721897-42721897 on Build GRCh38
rs146844134 Chr.6: 42722189-42722189 on Build GRCh38
rs950303632 Chr.6: 42722011-42722011 on Build GRCh38
rs61755770 Chr.6: 42722202-42722202 on Build GRCh38
rs61755775 Chr.6: 42722086-42722086 on Build GRCh38
rs760213205 Chr.6: 42721966-42721966 on Build GRCh38
rs751049619 Chr.6: 42721739-42721739 on Build GRCh38
rs61755776 Chr.6: 42722032-42722032 on Build GRCh38
rs746940785 Chr.6: 42722198-42722198 on Build GRCh38
rs563581127 Chr.6: 42721968-42721968 on Build GRCh38
rs61755790 Chr.6: 42721828-42721830 on Build GRCh38
rs574297962 Chr.6: 42721669-42721669 on Build GRCh38
rs61755780 Chr.6: 42721914-42721914 on Build GRCh38
rs567678322 Chr.6: 42722055-42722055 on Build GRCh38
rs752700882 Chr.6: 42721727-42721727 on Build GRCh38
rs371990988 Chr.6: 42722107-42722107 on Build GRCh38
rs61755771 Chr.6: 42722199-42722199 on Build GRCh38
rs777764923 Chr.6: 42721960-42721960 on Build GRCh38
rs786205579 Chr.6: 42721838-42721838 on Build GRCh38
rs762692183 Chr.6: 42721936-42721936 on Build GRCh38
rs758276201 Chr.6: 42721729-42721729 on Build GRCh38
rs748478593 Chr.6: 42721805-42721805 on Build GRCh38
rs372456408 Chr.6: 42722188-42722188 on Build GRCh38
rs750164572 Chr.6: 42721890-42721890 on Build GRCh38
rs61755785 Chr.6: 42721877-42721877 on Build GRCh38
rs672601326 Chr.6: 42721913-42721914 on Build GRCh38
rs750076275 Chr.6: 42721769-42721769 on Build GRCh38
rs200876455 Chr.6: 42721824-42721824 on Build GRCh38
rs530135649 Chr.6: 42722065-42722065 on Build GRCh38
rs771068866 Chr.6: 42721923-42721923 on Build GRCh38
rs747987442 Chr.6: 42722187-42722187 on Build GRCh38
rs750904649 Chr.6: 42722127-42722127 on Build GRCh38
rs121918563 Chr.6: 42721781-42721781 on Build GRCh38
rs554945964 Chr.6: 42721910-42721910 on Build GRCh38
rs61755794 Chr.6: 42721817-42721817 on Build GRCh38
rs143293289 Chr.6: 42721665-42721665 on Build GRCh38
rs770029839 Chr.6: 42721930-42721930 on Build GRCh38
rs960152679 Chr.6: 42721787-42721787 on Build GRCh38
rs61755789 Chr.6: 42721835-42721835 on Build GRCh38
rs768400169 Chr.6: 42722036-42722036 on Build GRCh38
rs879352836 Chr.6: 42722151-42722151 on Build GRCh38
rs61755782 Chr.6: 42721911-42721912 on Build GRCh38
rs61755781 Chr.6: 42721913-42721913 on Build GRCh38
rs140227298 Chr.6: 42721989-42721989 on Build GRCh38
rs199572514 Chr.6: 42721871-42721871 on Build GRCh38
rs749258271 Chr.6: 42722139-42722139 on Build GRCh38
rs1008805780 Chr.6: 42722132-42722132 on Build GRCh38
rs558060514 Chr.6: 42722130-42722130 on Build GRCh38
rs753035269 Chr.6: 42721898-42721898 on Build GRCh38
rs61755788 Chr.6: 42721841-42721841 on Build GRCh38
rs769723975 Chr.6: 42721941-42721941 on Build GRCh38
rs527236098 Chr.6: 42721836-42721836 on Build GRCh38
rs749136402 Chr.6: 42722113-42722115 on Build GRCh38
rs61755784 Chr.6: 42721894-42721894 on Build GRCh38
rs375090109 Chr.6: 42722120-42722120 on Build GRCh38
rs765006899 Chr.6: 42721726-42721726 on Build GRCh38
rs76989855 Chr.6: 42721852-42721852 on Build GRCh38
rs201501819 Chr.6: 42721804-42721804 on Build GRCh38
rs868309840 Chr.6: 42722182-42722182 on Build GRCh38
rs758924894 Chr.6: 42722205-42722205 on Build GRCh38
rs980078719 Chr.6: 42721704-42721704 on Build GRCh38
rs61755791 Chr.6: 42721827-42721827 on Build GRCh38
rs61755795 Chr.6: 42721802-42721802 on Build GRCh38
rs757778182 Chr.6: 42722001-42722001 on Build GRCh38
rs148513859 Chr.6: 42721927-42721927 on Build GRCh38
rs904908608 Chr.6: 42721661-42721661 on Build GRCh38
rs781029409 Chr.6: 42721762-42721762 on Build GRCh38
rs935103151 Chr.6: 42721873-42721873 on Build GRCh38
rs772157862 Chr.6: 42722066-42722066 on Build GRCh38
rs672601327 Chr.6: 42721872-42721874 on Build GRCh38
rs61755774 Chr.6: 42722133-42722133 on Build GRCh38
rs61729453 Chr.6: 42722197-42722197 on Build GRCh38
rs574930557 Chr.6: 42721933-42721933 on Build GRCh38
rs774210091 Chr.6: 42721954-42721954 on Build GRCh38
rs776805706 Chr.6: 42721719-42721719 on Build GRCh38
rs63749073 Chr.6: 42722133-42722137 on Build GRCh38
rs751480327 Chr.6: 42721858-42721858 on Build GRCh38
rs879134790 Chr.6: 42721666-42721666 on Build GRCh38
rs61755786 Chr.6: 42721873-42721875 on Build GRCh38
rs61755787 Chr.6: 42721866-42721866 on Build GRCh38
rs751946505 Chr.6: 42722098-42722098 on Build GRCh38
rs780175779 Chr.6: 42722131-42722131 on Build GRCh38
rs747893076 Chr.6: 42722064-42722064 on Build GRCh38
rs369556021 Chr.6: 42722185-42722185 on Build GRCh38
rs749097332 Chr.6: 42722111-42722111 on Build GRCh38
rs200009675 Chr.6: 42722023-42722023 on Build GRCh38
rs755239769 Chr.6: 42722149-42722149 on Build GRCh38
rs769939935 Chr.6: 42721851-42721851 on Build GRCh38
rs781256236 Chr.6: 42721926-42721926 on Build GRCh38
rs376120359 Chr.6: 42721732-42721732 on Build GRCh38
rs61755777 Chr.6: 42721978-42721980 on Build GRCh38
rs121918561 Chr.6: 42721981-42721983 on Build GRCh38
rs759752477 Chr.6: 42721837-42721837 on Build GRCh38
rs150695654 Chr.6: 42722035-42722035 on Build GRCh38
rs1011270423 Chr.6: 42721659-42721659 on Build GRCh38
rs62645926 Chr.6: 42721784-42721784 on Build GRCh38
rs200978519 Chr.6: 42721900-42721900 on Build GRCh38
rs62645929 Chr.6: 42721756-42721757 on Build GRCh38
rs146703538 Chr.6: 42721881-42721881 on Build GRCh38
rs7764439 Chr.6: 42722017-42722017 on Build GRCh38
rs768473101 Chr.6: 42721938-42721938 on Build GRCh38
rs61755779 Chr.6: 42721959-42721959 on Build GRCh38
rs766542243 Chr.6: 42721713-42721713 on Build GRCh38
rs756746825 Chr.6: 42721755-42721755 on Build GRCh38
rs780590186 Chr.6: 42721764-42721764 on Build GRCh38
rs373113428 Chr.6: 42721723-42721723 on Build GRCh38
rs543703718 Chr.6: 42721955-42721955 on Build GRCh38
rs61755793 Chr.6: 42721820-42721820 on Build GRCh38
rs773254206 Chr.6: 42722070-42722070 on Build GRCh38
rs760311433 Chr.6: 42722082-42722082 on Build GRCh38
rs6928781 Chr.6: 42721673-42721673 on Build GRCh38
rs755449912 Chr.6: 42721788-42721788 on Build GRCh38
rs993714311 Chr.6: 42721780-42721780 on Build GRCh38
rs779414078 Chr.6: 42721798-42721798 on Build GRCh38
rs761275918 Chr.6: 42721867-42721867 on Build GRCh38
rs139936445 Chr.6: 42722083-42722083 on Build GRCh38
rs368257452 Chr.6: 42722085-42722085 on Build GRCh38
rs532319562 Chr.6: 42721986-42721986 on Build GRCh38
rs527236097 Chr.6: 42721925-42721925 on Build GRCh38
rs62645928 Chr.6: 42721756-42721758 on Build GRCh38
rs61755792 Chr.6: 42721821-42721821 on Build GRCh38
rs61755783 Chr.6: 42721911-42721911 on Build GRCh38
rs61755773 Chr.6: 42722134-42722136 on Build GRCh38
rs753507738 Chr.6: 42721972-42721972 on Build GRCh38
rs61755778 Chr.6: 42721964-42721964 on Build GRCh38
rs143096101 Chr.6: 42721828-42721828 on Build GRCh38

View Full Product Details