Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87518548-87519084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RARS2
Entrez Gene ID
57038
Gene Name
arginyl-tRNA synthetase 2, mitochondrial
Gene Aliases
ArgRS, DALRD2, PCH6, PRO1992, RARSL
UniGene
Hs.485910
Gene Chromosome Location
Chr.6: 87513744-87590032 on Build GRCh38
Transcripts
NM_001318785.1 NM_020320.4 NR_134857.1 XM_011535948.2 XM_011535949.2 XM_017011072.1 XM_017011073.1 XM_017011074.1 XM_017011075.1 XM_017011076.1 XM_017011077.1 XM_017011078.1 XM_017011079.1 XM_017011080.1 XM_017011081.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5787071 c.1305+10G>C p.? Chr.6: 87518814-87518814 on Build GRCh38
6107593 c.1293A>T p.A431A Chr.6: 87518836-87518836 on Build GRCh38
6763013 c.1287C>T p.L429L Chr.6: 87518842-87518842 on Build GRCh38
5507542 c.1288G>A p.A430T Chr.6: 87518841-87518841 on Build GRCh38
6020417 c.1273G>C p.E425Q Chr.6: 87518856-87518856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375589432 Chr.6: 87518747-87518747 on Build GRCh38
rs751019907 Chr.6: 87518750-87518750 on Build GRCh38
rs1057518531 Chr.6: 87518804-87518804 on Build GRCh38
rs182823716 Chr.6: 87518751-87518751 on Build GRCh38
rs1024293332 Chr.6: 87519030-87519030 on Build GRCh38
rs924027192 Chr.6: 87518865-87518865 on Build GRCh38
rs764280940 Chr.6: 87518583-87518586 on Build GRCh38
rs764711096 Chr.6: 87518641-87518641 on Build GRCh38
rs759331139 Chr.6: 87518639-87518639 on Build GRCh38
rs757736377 Chr.6: 87518603-87518603 on Build GRCh38
rs192821560 Chr.6: 87518842-87518842 on Build GRCh38
rs755167895 Chr.6: 87518789-87518789 on Build GRCh38
rs374122988 Chr.6: 87518805-87518805 on Build GRCh38
rs539646866 Chr.6: 87519028-87519028 on Build GRCh38
rs766014854 Chr.6: 87518907-87518907 on Build GRCh38
rs577289150 Chr.6: 87518746-87518746 on Build GRCh38
rs143076225 Chr.6: 87518883-87518883 on Build GRCh38
rs751992245 Chr.6: 87518780-87518780 on Build GRCh38
rs767767123 Chr.6: 87518875-87518875 on Build GRCh38
rs202092389 Chr.6: 87518682-87518682 on Build GRCh38
rs201596237 Chr.6: 87518941-87518941 on Build GRCh38
rs370940168 Chr.6: 87518644-87518644 on Build GRCh38
rs749385331 Chr.6: 87518908-87518908 on Build GRCh38
rs767004280 Chr.6: 87518866-87518866 on Build GRCh38
rs199584519 Chr.6: 87518798-87518798 on Build GRCh38
rs773434032 Chr.6: 87518742-87518742 on Build GRCh38
rs778121766 Chr.6: 87518581-87518581 on Build GRCh38
rs139564081 Chr.6: 87518635-87518635 on Build GRCh38
rs202090254 Chr.6: 87518986-87518989 on Build GRCh38
rs188174893 Chr.6: 87518770-87518770 on Build GRCh38
rs142443912 Chr.6: 87518934-87518934 on Build GRCh38
rs771658534 Chr.6: 87518876-87518876 on Build GRCh38
rs896630672 Chr.6: 87519011-87519011 on Build GRCh38
rs946822680 Chr.6: 87518882-87518882 on Build GRCh38
rs779295415 Chr.6: 87518669-87518669 on Build GRCh38
rs147844153 Chr.6: 87518679-87518679 on Build GRCh38
rs779288448 Chr.6: 87518606-87518606 on Build GRCh38
rs751904499 Chr.6: 87518872-87518872 on Build GRCh38
rs1056912836 Chr.6: 87519002-87519002 on Build GRCh38
rs540253901 Chr.6: 87518578-87518578 on Build GRCh38
rs144447777 Chr.6: 87518678-87518678 on Build GRCh38
rs1036068925 Chr.6: 87518960-87518960 on Build GRCh38
rs377000350 Chr.6: 87518677-87518677 on Build GRCh38
rs41273291 Chr.6: 87518926-87518926 on Build GRCh38
rs1027330571 Chr.6: 87519032-87519032 on Build GRCh38
rs756545365 Chr.6: 87518926-87518927 on Build GRCh38
rs755687141 Chr.6: 87518579-87518579 on Build GRCh38
rs922879443 Chr.6: 87518855-87518855 on Build GRCh38
rs757728606 Chr.6: 87518668-87518668 on Build GRCh38
rs777781045 Chr.6: 87518886-87518886 on Build GRCh38
rs775209523 Chr.6: 87518812-87518812 on Build GRCh38
rs754411280 Chr.6: 87518587-87518587 on Build GRCh38
rs932650639 Chr.6: 87518869-87518869 on Build GRCh38
rs750816674 Chr.6: 87518671-87518671 on Build GRCh38
rs764875815 Chr.6: 87518847-87518847 on Build GRCh38
rs895104861 Chr.6: 87519022-87519022 on Build GRCh38
rs551756663 Chr.6: 87518665-87518665 on Build GRCh38
rs1002590565 Chr.6: 87518981-87518981 on Build GRCh38
rs767150990 Chr.6: 87518640-87518640 on Build GRCh38
rs3734908 Chr.6: 87518806-87518806 on Build GRCh38
rs1012320735 Chr.6: 87519026-87519026 on Build GRCh38
rs752396603 Chr.6: 87518643-87518643 on Build GRCh38
rs774339242 Chr.6: 87518698-87518698 on Build GRCh38
rs780493627 Chr.6: 87518615-87518615 on Build GRCh38
rs144988947 Chr.6: 87518707-87518707 on Build GRCh38
rs144896612 Chr.6: 87518711-87518711 on Build GRCh38
rs373600402 Chr.6: 87518612-87518612 on Build GRCh38
rs768826734 Chr.6: 87518624-87518624 on Build GRCh38
rs763667779 Chr.6: 87518732-87518732 on Build GRCh38
rs776208986 Chr.6: 87518841-87518841 on Build GRCh38
rs772875883 Chr.6: 87518848-87518848 on Build GRCh38
rs374214191 Chr.6: 87518693-87518693 on Build GRCh38
rs772358117 Chr.6: 87518614-87518614 on Build GRCh38
rs575650808 Chr.6: 87518970-87518970 on Build GRCh38
rs991448116 Chr.6: 87518736-87518736 on Build GRCh38
rs745765361 Chr.6: 87518810-87518810 on Build GRCh38
rs573594453 Chr.6: 87519058-87519058 on Build GRCh38
rs562751470 Chr.6: 87518743-87518743 on Build GRCh38
rs751624778 Chr.6: 87518589-87518598 on Build GRCh38
rs372092585 Chr.6: 87518833-87518833 on Build GRCh38
rs760327372 Chr.6: 87518727-87518727 on Build GRCh38
rs775295739 Chr.6: 87518718-87518718 on Build GRCh38
rs968433432 Chr.6: 87518782-87518782 on Build GRCh38
rs200492384 Chr.6: 87518649-87518649 on Build GRCh38
rs750873299 Chr.6: 87518827-87518827 on Build GRCh38
rs755363278 Chr.6: 87518874-87518874 on Build GRCh38
rs149046384 Chr.6: 87518850-87518850 on Build GRCh38
rs779393765 Chr.6: 87518995-87518995 on Build GRCh38
rs141460433 Chr.6: 87518694-87518694 on Build GRCh38
rs778799689 Chr.6: 87518809-87518809 on Build GRCh38
rs758894784 Chr.6: 87518676-87518676 on Build GRCh38
rs753312969 Chr.6: 87518655-87518655 on Build GRCh38
rs768352579 Chr.6: 87518839-87518839 on Build GRCh38
rs772989345 Chr.6: 87518628-87518628 on Build GRCh38
rs187815721 Chr.6: 87518632-87518632 on Build GRCh38
rs768019443 Chr.6: 87518791-87518791 on Build GRCh38
rs1012438012 Chr.6: 87519024-87519024 on Build GRCh38
rs756206300 Chr.6: 87518807-87518807 on Build GRCh38
rs558353837 Chr.6: 87519046-87519046 on Build GRCh38
rs747075912 Chr.6: 87518620-87518620 on Build GRCh38

View Full Product Details