Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116880571-116880933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFX6
Entrez Gene ID
222546
Gene Name
regulatory factor X6
Gene Aliases
MTCHRS, MTFS, RFXDC1, dJ955L16.1
UniGene
Hs.352276
Gene Chromosome Location
Chr.6: 116877213-116932163 on Build GRCh38
Transcripts
NM_173560.3 XM_011535589.1 XM_017010477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3619753 c.495C>T p.T165T Chr.6: 116880658-116880658 on Build GRCh38
1697798 c.408A>T p.E136D Chr.6: 116880571-116880571 on Build GRCh38
5404213 c.412G>A p.V138I Chr.6: 116880575-116880575 on Build GRCh38
5951176 c.479C>T p.P160L Chr.6: 116880642-116880642 on Build GRCh38
4760491 c.504G>C p.K168N Chr.6: 116880667-116880667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769409570 Chr.6: 116880607-116880607 on Build GRCh38
rs761820367 Chr.6: 116880893-116880893 on Build GRCh38
rs745575226 Chr.6: 116880678-116880678 on Build GRCh38
rs373152553 Chr.6: 116880616-116880616 on Build GRCh38
rs199687281 Chr.6: 116880688-116880688 on Build GRCh38
rs751139970 Chr.6: 116880706-116880706 on Build GRCh38
rs1041420610 Chr.6: 116880836-116880836 on Build GRCh38
rs951641567 Chr.6: 116880821-116880822 on Build GRCh38
rs559897961 Chr.6: 116880813-116880813 on Build GRCh38
rs149957510 Chr.6: 116880705-116880705 on Build GRCh38
rs769171416 Chr.6: 116880675-116880675 on Build GRCh38
rs965882346 Chr.6: 116880594-116880594 on Build GRCh38
rs938341286 Chr.6: 116880663-116880663 on Build GRCh38
rs142041542 Chr.6: 116880818-116880818 on Build GRCh38
rs994247579 Chr.6: 116880603-116880603 on Build GRCh38
rs1037452168 Chr.6: 116880712-116880712 on Build GRCh38
rs984058497 Chr.6: 116880679-116880679 on Build GRCh38
rs372661046 Chr.6: 116880808-116880809 on Build GRCh38
rs762557693 Chr.6: 116880691-116880691 on Build GRCh38
rs746133716 Chr.6: 116880631-116880631 on Build GRCh38
rs898660305 Chr.6: 116880770-116880770 on Build GRCh38
rs530843099 Chr.6: 116880907-116880907 on Build GRCh38
rs541223818 Chr.6: 116880787-116880787 on Build GRCh38
rs777620596 Chr.6: 116880619-116880619 on Build GRCh38
rs762166593 Chr.6: 116880682-116880682 on Build GRCh38
rs754657962 Chr.6: 116880746-116880746 on Build GRCh38
rs772316570 Chr.6: 116880651-116880651 on Build GRCh38
rs778438467 Chr.6: 116880771-116880771 on Build GRCh38
rs563328137 Chr.6: 116880862-116880862 on Build GRCh38
rs760704729 Chr.6: 116880657-116880657 on Build GRCh38
rs775585977 Chr.6: 116880652-116880652 on Build GRCh38
rs57167027 Chr.6: 116880822-116880822 on Build GRCh38

View Full Product Details